Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7741075-7741583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD209
Entrez Gene ID
30835
Gene Name
CD209 molecule
Gene Aliases
CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1
UniGene
Hs.278694
Gene Chromosome Location
Chr.19: 7739993-7747609 on Build GRCh38
Transcripts
NM_001144893.1 NM_001144894.1 NM_001144895.1 NM_001144896.1 NM_001144897.1 NM_001144899.1 NM_021155.3 NR_026692.1 XM_005272472.4

SNP Detail

SNP ID Chromosome Location
rs963854728 Chr.19: 7741237-7741237 on Build GRCh38
rs796475184 Chr.19: 7741346-7741346 on Build GRCh38
rs535167552 Chr.19: 7741492-7741492 on Build GRCh38
rs565643352 Chr.19: 7741515-7741515 on Build GRCh38
rs1053674937 Chr.19: 7741277-7741277 on Build GRCh38
rs879618257 Chr.19: 7741470-7741470 on Build GRCh38
rs768293088 Chr.19: 7741327-7741327 on Build GRCh38
rs772896241 Chr.19: 7741358-7741358 on Build GRCh38
rs374694844 Chr.19: 7741239-7741239 on Build GRCh38
rs563294494 Chr.19: 7741538-7741538 on Build GRCh38
rs11465406 Chr.19: 7741344-7741344 on Build GRCh38
rs546385583 Chr.19: 7741180-7741180 on Build GRCh38
rs758293186 Chr.19: 7741141-7741141 on Build GRCh38
rs144176155 Chr.19: 7741198-7741198 on Build GRCh38
rs982158408 Chr.19: 7741420-7741420 on Build GRCh38
rs534340413 Chr.19: 7741276-7741276 on Build GRCh38
rs11465410 Chr.19: 7741193-7741193 on Build GRCh38
rs953672919 Chr.19: 7741207-7741207 on Build GRCh38
rs113936914 Chr.19: 7741192-7741192 on Build GRCh38
rs548839611 Chr.19: 7741498-7741498 on Build GRCh38
rs560547049 Chr.19: 7741118-7741118 on Build GRCh38
rs536492795 Chr.19: 7741491-7741492 on Build GRCh38
rs893659269 Chr.19: 7741316-7741316 on Build GRCh38
rs949488915 Chr.19: 7741241-7741241 on Build GRCh38
rs556742276 Chr.19: 7741319-7741319 on Build GRCh38
rs386806502 Chr.19: 7741191-7741193 on Build GRCh38
rs563534836 Chr.19: 7741448-7741448 on Build GRCh38
rs970804728 Chr.19: 7741414-7741414 on Build GRCh38
rs543231457 Chr.19: 7741434-7741434 on Build GRCh38
rs397897390 Chr.19: 7741345-7741345 on Build GRCh38
rs11465405 Chr.19: 7741453-7741453 on Build GRCh38
rs1053621492 Chr.19: 7741165-7741165 on Build GRCh38
rs867217755 Chr.19: 7741219-7741219 on Build GRCh38
rs201094015 Chr.19: 7741343-7741343 on Build GRCh38
rs549990133 Chr.19: 7741238-7741238 on Build GRCh38
rs1009331904 Chr.19: 7741190-7741190 on Build GRCh38
rs532944619 Chr.19: 7741220-7741220 on Build GRCh38
rs572423438 Chr.19: 7741365-7741365 on Build GRCh38
rs905142903 Chr.19: 7741115-7741115 on Build GRCh38
rs529094961 Chr.19: 7741472-7741472 on Build GRCh38
rs1046417524 Chr.19: 7741110-7741110 on Build GRCh38
rs754714381 Chr.19: 7741117-7741117 on Build GRCh38
rs11465411 Chr.19: 7741191-7741191 on Build GRCh38
rs539640019 Chr.19: 7741318-7741318 on Build GRCh38
rs919364392 Chr.19: 7741240-7741240 on Build GRCh38
rs909393873 Chr.19: 7741226-7741226 on Build GRCh38
rs11260028 Chr.19: 7741333-7741333 on Build GRCh38
rs762968999 Chr.19: 7741364-7741364 on Build GRCh38
rs190106388 Chr.19: 7741254-7741254 on Build GRCh38
rs11465412 Chr.19: 7741184-7741184 on Build GRCh38
rs11465408 Chr.19: 7741268-7741268 on Build GRCh38
rs562097599 Chr.19: 7741197-7741197 on Build GRCh38
rs748005219 Chr.19: 7741137-7741137 on Build GRCh38
rs556116854 Chr.19: 7741357-7741357 on Build GRCh38
rs760255942 Chr.19: 7741424-7741424 on Build GRCh38
rs564574231 Chr.19: 7741213-7741213 on Build GRCh38
rs997890618 Chr.19: 7741330-7741330 on Build GRCh38
rs12460694 Chr.19: 7741369-7741369 on Build GRCh38
rs867765175 Chr.19: 7741106-7741106 on Build GRCh38
rs35705945 Chr.19: 7741221-7741222 on Build GRCh38
rs533515835 Chr.19: 7741408-7741408 on Build GRCh38
rs766190869 Chr.19: 7741529-7741529 on Build GRCh38
rs1016950053 Chr.19: 7741353-7741353 on Build GRCh38
rs11465409 Chr.19: 7741200-7741200 on Build GRCh38
rs945306378 Chr.19: 7741492-7741492 on Build GRCh38
rs529216188 Chr.19: 7741253-7741253 on Build GRCh38
rs181874569 Chr.19: 7741331-7741331 on Build GRCh38
rs187334590 Chr.19: 7741243-7741243 on Build GRCh38
rs564562150 Chr.19: 7741384-7741384 on Build GRCh38
rs776109765 Chr.19: 7741296-7741296 on Build GRCh38
rs571047910 Chr.19: 7741289-7741289 on Build GRCh38
rs778958009 Chr.19: 7741129-7741129 on Build GRCh38
rs73921550 Chr.19: 7741541-7741541 on Build GRCh38
rs900967601 Chr.19: 7741536-7741536 on Build GRCh38
rs557387594 Chr.19: 7741287-7741287 on Build GRCh38

View Full Product Details