Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8302685-8303110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD320
Entrez Gene ID
51293
Gene Name
CD320 molecule
Gene Aliases
8D6, 8D6A, TCBLR
UniGene
Hs.558499
Gene Chromosome Location
Chr.19: 8302127-8308356 on Build GRCh38
Transcripts
NM_001165895.1 NM_016579.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1711737 c.626_627CC>TT p.P209L Chr.19: 8302856-8302857 on Build GRCh38
1711738 c.577C>T p.L193F Chr.19: 8302906-8302906 on Build GRCh38
4981936 c.532G>A p.A178T Chr.19: 8302951-8302951 on Build GRCh38
4132802 c.658G>A p.G220R Chr.19: 8302825-8302825 on Build GRCh38
1711739 c.520_521CC>TT p.P174L Chr.19: 8302962-8302963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543136189 Chr.19: 8302739-8302739 on Build GRCh38
rs115039587 Chr.19: 8303028-8303028 on Build GRCh38
rs767894834 Chr.19: 8302761-8302765 on Build GRCh38
rs143803542 Chr.19: 8302964-8302964 on Build GRCh38
rs755718429 Chr.19: 8302818-8302818 on Build GRCh38
rs749530258 Chr.19: 8302865-8302865 on Build GRCh38
rs964182059 Chr.19: 8303052-8303052 on Build GRCh38
rs368024906 Chr.19: 8302962-8302962 on Build GRCh38
rs2336573 Chr.19: 8302825-8302825 on Build GRCh38
rs754469537 Chr.19: 8302931-8302932 on Build GRCh38
rs528401782 Chr.19: 8302746-8302746 on Build GRCh38
rs1011293712 Chr.19: 8303029-8303029 on Build GRCh38
rs958385780 Chr.19: 8302732-8302732 on Build GRCh38
rs989778116 Chr.19: 8303040-8303040 on Build GRCh38
rs753410306 Chr.19: 8302906-8302907 on Build GRCh38
rs79658260 Chr.19: 8302961-8302961 on Build GRCh38
rs772010099 Chr.19: 8302766-8302766 on Build GRCh38
rs759992081 Chr.19: 8302880-8302880 on Build GRCh38
rs149721174 Chr.19: 8302802-8302802 on Build GRCh38
rs754818625 Chr.19: 8302955-8302955 on Build GRCh38
rs776070970 Chr.19: 8302895-8302895 on Build GRCh38
rs957741164 Chr.19: 8303029-8303030 on Build GRCh38
rs188427014 Chr.19: 8302995-8302995 on Build GRCh38
rs780100188 Chr.19: 8302847-8302847 on Build GRCh38
rs748946125 Chr.19: 8302736-8302736 on Build GRCh38
rs746913945 Chr.19: 8302848-8302848 on Build GRCh38
rs1021300319 Chr.19: 8303043-8303043 on Build GRCh38
rs558056448 Chr.19: 8302977-8302977 on Build GRCh38
rs774612614 Chr.19: 8302963-8302963 on Build GRCh38
rs545972688 Chr.19: 8303021-8303021 on Build GRCh38
rs764838647 Chr.19: 8302987-8302987 on Build GRCh38
rs574401407 Chr.19: 8302714-8302714 on Build GRCh38
rs202203481 Chr.19: 8302937-8302937 on Build GRCh38
rs201027249 Chr.19: 8303012-8303012 on Build GRCh38
rs778739908 Chr.19: 8302991-8302991 on Build GRCh38
rs1033817603 Chr.19: 8303006-8303006 on Build GRCh38
rs551738826 Chr.19: 8302774-8302774 on Build GRCh38
rs747753907 Chr.19: 8303024-8303024 on Build GRCh38
rs146328123 Chr.19: 8302836-8302836 on Build GRCh38
rs761030759 Chr.19: 8302907-8302907 on Build GRCh38
rs777259220 Chr.19: 8302982-8302982 on Build GRCh38
rs778793240 Chr.19: 8302744-8302744 on Build GRCh38
rs148167974 Chr.19: 8302972-8302972 on Build GRCh38
rs778887130 Chr.19: 8302828-8302828 on Build GRCh38
rs770855698 Chr.19: 8302741-8302741 on Build GRCh38
rs753363084 Chr.19: 8302917-8302917 on Build GRCh38
rs967442273 Chr.19: 8303072-8303072 on Build GRCh38
rs374451374 Chr.19: 8302944-8302944 on Build GRCh38
rs114265368 Chr.19: 8302998-8302998 on Build GRCh38
rs146900609 Chr.19: 8302974-8302974 on Build GRCh38
rs774524619 Chr.19: 8302878-8302878 on Build GRCh38
rs529629965 Chr.19: 8302932-8302932 on Build GRCh38
rs766740010 Chr.19: 8302804-8302804 on Build GRCh38
rs762005169 Chr.19: 8302796-8302796 on Build GRCh38
rs559637771 Chr.19: 8302709-8302709 on Build GRCh38
rs566090996 Chr.19: 8302912-8302912 on Build GRCh38
rs377252284 Chr.19: 8302850-8302850 on Build GRCh38
rs760066815 Chr.19: 8302808-8302808 on Build GRCh38
rs767770100 Chr.19: 8302889-8302889 on Build GRCh38
rs779644500 Chr.19: 8302940-8302940 on Build GRCh38
rs906320781 Chr.19: 8303004-8303004 on Build GRCh38
rs750093192 Chr.19: 8302991-8302991 on Build GRCh38
rs201082304 Chr.19: 8302816-8302816 on Build GRCh38
rs763091865 Chr.19: 8302803-8302803 on Build GRCh38
rs772500448 Chr.19: 8302968-8302968 on Build GRCh38
rs753120890 Chr.19: 8302817-8302817 on Build GRCh38
rs763720075 Chr.19: 8302911-8302911 on Build GRCh38
rs769747738 Chr.19: 8302797-8302797 on Build GRCh38
rs550390075 Chr.19: 8302849-8302849 on Build GRCh38
rs530872669 Chr.19: 8302753-8302753 on Build GRCh38
rs374222979 Chr.19: 8302734-8302734 on Build GRCh38
rs959137935 Chr.19: 8302787-8302787 on Build GRCh38
rs368089898 Chr.19: 8302826-8302826 on Build GRCh38
rs139075923 Chr.19: 8302870-8302870 on Build GRCh38
rs761471053 Chr.19: 8302986-8302986 on Build GRCh38
rs372678225 Chr.19: 8302928-8302928 on Build GRCh38
rs200298268 Chr.19: 8303026-8303026 on Build GRCh38
rs374436174 Chr.19: 8303011-8303011 on Build GRCh38
rs746125718 Chr.19: 8303027-8303027 on Build GRCh38
rs766215870 Chr.19: 8302997-8302997 on Build GRCh38
rs758108600 Chr.19: 8302841-8302841 on Build GRCh38
rs147690674 Chr.19: 8302811-8302811 on Build GRCh38

View Full Product Details