Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8364018-8364521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL4
Entrez Gene ID
51129
Gene Name
angiopoietin like 4
Gene Aliases
ARP4, FIAF, HARP, HFARP, NL2, PGAR, TGQTL, UNQ171, pp1158
UniGene
Hs.9613
Gene Chromosome Location
Chr.19: 8364127-8374375 on Build GRCh38
Transcripts
NM_001039667.2 NM_139314.2 NR_104213.1 XM_005272484.3 XM_005272485.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6385316 c.190A>G p.S64G Chr.19: 8364511-8364511 on Build GRCh38
7355429 c.118G>A p.E40K Chr.19: 8364439-8364439 on Build GRCh38
4656332 c.188G>A p.R63H Chr.19: 8364509-8364509 on Build GRCh38
1631166 c.106G>T p.A36S Chr.19: 8364427-8364427 on Build GRCh38
6436575 c.34A>G p.M12V Chr.19: 8364355-8364355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760847575 Chr.19: 8364375-8364375 on Build GRCh38
rs765373026 Chr.19: 8364402-8364402 on Build GRCh38
rs967106746 Chr.19: 8364058-8364058 on Build GRCh38
rs186754194 Chr.19: 8364444-8364444 on Build GRCh38
rs901683775 Chr.19: 8364238-8364238 on Build GRCh38
rs1037990584 Chr.19: 8364236-8364236 on Build GRCh38
rs570089916 Chr.19: 8364068-8364068 on Build GRCh38
rs116843064 Chr.19: 8364439-8364439 on Build GRCh38
rs555162667 Chr.19: 8364121-8364121 on Build GRCh38
rs754020483 Chr.19: 8364289-8364289 on Build GRCh38
rs1012821736 Chr.19: 8364089-8364089 on Build GRCh38
rs997347875 Chr.19: 8364291-8364291 on Build GRCh38
rs776244357 Chr.19: 8364334-8364334 on Build GRCh38
rs768253869 Chr.19: 8364329-8364329 on Build GRCh38
rs373435482 Chr.19: 8364206-8364206 on Build GRCh38
rs753864074 Chr.19: 8364388-8364388 on Build GRCh38
rs980968097 Chr.19: 8364367-8364367 on Build GRCh38
rs4076317 Chr.19: 8364115-8364115 on Build GRCh38
rs757440743 Chr.19: 8364297-8364297 on Build GRCh38
rs866788681 Chr.19: 8364495-8364495 on Build GRCh38
rs374923101 Chr.19: 8364305-8364305 on Build GRCh38
rs948267922 Chr.19: 8364278-8364278 on Build GRCh38
rs866272907 Chr.19: 8364341-8364341 on Build GRCh38
rs970881685 Chr.19: 8364354-8364354 on Build GRCh38
rs766397887 Chr.19: 8364353-8364353 on Build GRCh38
rs1005829925 Chr.19: 8364323-8364323 on Build GRCh38
rs906931140 Chr.19: 8364105-8364105 on Build GRCh38
rs913024943 Chr.19: 8364054-8364054 on Build GRCh38
rs922718704 Chr.19: 8364481-8364481 on Build GRCh38
rs762127484 Chr.19: 8364053-8364053 on Build GRCh38
rs764188393 Chr.19: 8364386-8364386 on Build GRCh38
rs11544611 Chr.19: 8364301-8364301 on Build GRCh38
rs776996548 Chr.19: 8364396-8364396 on Build GRCh38
rs1022747478 Chr.19: 8364096-8364096 on Build GRCh38
rs769637836 Chr.19: 8364465-8364465 on Build GRCh38
rs757481474 Chr.19: 8364398-8364398 on Build GRCh38
rs895373925 Chr.19: 8364263-8364263 on Build GRCh38
rs569539753 Chr.19: 8364081-8364081 on Build GRCh38
rs541624403 Chr.19: 8364387-8364387 on Build GRCh38
rs997400079 Chr.19: 8364106-8364106 on Build GRCh38
rs974891921 Chr.19: 8364042-8364042 on Build GRCh38
rs780654441 Chr.19: 8364441-8364441 on Build GRCh38
rs953051314 Chr.19: 8364123-8364123 on Build GRCh38
rs964963416 Chr.19: 8364401-8364401 on Build GRCh38
rs1015958266 Chr.19: 8364150-8364150 on Build GRCh38
rs576293992 Chr.19: 8364336-8364336 on Build GRCh38
rs978451160 Chr.19: 8364085-8364085 on Build GRCh38
rs574811101 Chr.19: 8364142-8364142 on Build GRCh38
rs763423702 Chr.19: 8364161-8364161 on Build GRCh38
rs774154527 Chr.19: 8364493-8364493 on Build GRCh38
rs981023950 Chr.19: 8364193-8364193 on Build GRCh38
rs918105445 Chr.19: 8364175-8364175 on Build GRCh38
rs370009734 Chr.19: 8364205-8364205 on Build GRCh38
rs747597003 Chr.19: 8364462-8364462 on Build GRCh38
rs537310571 Chr.19: 8364327-8364327 on Build GRCh38
rs964606868 Chr.19: 8364171-8364171 on Build GRCh38
rs199968545 Chr.19: 8364421-8364421 on Build GRCh38
rs150590640 Chr.19: 8364298-8364298 on Build GRCh38
rs565710166 Chr.19: 8364212-8364212 on Build GRCh38
rs372653113 Chr.19: 8364051-8364051 on Build GRCh38
rs539078315 Chr.19: 8364079-8364079 on Build GRCh38
rs945497646 Chr.19: 8364232-8364232 on Build GRCh38
rs772634881 Chr.19: 8364312-8364312 on Build GRCh38
rs926847231 Chr.19: 8364368-8364368 on Build GRCh38
rs376357875 Chr.19: 8364477-8364477 on Build GRCh38
rs746583639 Chr.19: 8364328-8364328 on Build GRCh38
rs780416714 Chr.19: 8364324-8364324 on Build GRCh38
rs1047698759 Chr.19: 8364070-8364070 on Build GRCh38
rs532722600 Chr.19: 8364459-8364459 on Build GRCh38
rs972618841 Chr.19: 8364169-8364169 on Build GRCh38
rs762653036 Chr.19: 8364352-8364352 on Build GRCh38
rs11544612 Chr.19: 8364208-8364208 on Build GRCh38
rs993269648 Chr.19: 8364344-8364344 on Build GRCh38
rs761583091 Chr.19: 8364335-8364335 on Build GRCh38
rs1025526193 Chr.19: 8364346-8364346 on Build GRCh38
rs913954371 Chr.19: 8364226-8364226 on Build GRCh38
rs558824775 Chr.19: 8364080-8364080 on Build GRCh38
rs770601171 Chr.19: 8364482-8364482 on Build GRCh38
rs923345662 Chr.19: 8364049-8364049 on Build GRCh38
rs759490625 Chr.19: 8364503-8364503 on Build GRCh38
rs773147922 Chr.19: 8364349-8364349 on Build GRCh38
rs1041911598 Chr.19: 8364233-8364233 on Build GRCh38
rs751429634 Chr.19: 8364362-8364362 on Build GRCh38
rs35137994 Chr.19: 8364182-8364182 on Build GRCh38

View Full Product Details