Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9959932-9960428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL5A3
Entrez Gene ID
50509
Gene Name
collagen type V alpha 3
Gene Aliases
-
UniGene
Hs.235368
Gene Chromosome Location
Chr.19: 9959561-10010532 on Build GRCh38
Transcripts
NM_015719.3 XM_011528042.2 XM_017026849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1189863 c.5226C>A p.C1742* Chr.19: 9960423-9960423 on Build GRCh38
5424946 c.5221G>A p.V1741I Chr.19: 9960428-9960428 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772807518 Chr.19: 9960283-9960283 on Build GRCh38
rs912956202 Chr.19: 9960136-9960136 on Build GRCh38
rs200938535 Chr.19: 9960366-9960366 on Build GRCh38
rs1013768576 Chr.19: 9960313-9960313 on Build GRCh38
rs868455616 Chr.19: 9960354-9960354 on Build GRCh38
rs1004624727 Chr.19: 9960189-9960189 on Build GRCh38
rs974306909 Chr.19: 9960044-9960044 on Build GRCh38
rs528095148 Chr.19: 9960221-9960221 on Build GRCh38
rs1042955958 Chr.19: 9960130-9960130 on Build GRCh38
rs552187023 Chr.19: 9960202-9960202 on Build GRCh38
rs551238510 Chr.19: 9960124-9960124 on Build GRCh38
rs762390903 Chr.19: 9960388-9960388 on Build GRCh38
rs768467338 Chr.19: 9959967-9959967 on Build GRCh38
rs73020030 Chr.19: 9960276-9960276 on Build GRCh38
rs369674786 Chr.19: 9960406-9960406 on Build GRCh38
rs868487597 Chr.19: 9960018-9960018 on Build GRCh38
rs546825998 Chr.19: 9960265-9960265 on Build GRCh38
rs370477121 Chr.19: 9960332-9960332 on Build GRCh38
rs35522591 Chr.19: 9959964-9959965 on Build GRCh38
rs772473205 Chr.19: 9960368-9960368 on Build GRCh38
rs749160193 Chr.19: 9959956-9959956 on Build GRCh38
rs75339267 Chr.19: 9960363-9960363 on Build GRCh38
rs992816587 Chr.19: 9960047-9960047 on Build GRCh38
rs995106502 Chr.19: 9960239-9960239 on Build GRCh38
rs913055160 Chr.19: 9960102-9960102 on Build GRCh38
rs115689201 Chr.19: 9960016-9960016 on Build GRCh38
rs386806739 Chr.19: 9960276-9960278 on Build GRCh38
rs182768899 Chr.19: 9960281-9960281 on Build GRCh38
rs570393410 Chr.19: 9960216-9960216 on Build GRCh38
rs996095172 Chr.19: 9960253-9960253 on Build GRCh38
rs1043396896 Chr.19: 9959958-9959958 on Build GRCh38
rs943927769 Chr.19: 9960166-9960166 on Build GRCh38
rs961569550 Chr.19: 9960041-9960041 on Build GRCh38
rs988890838 Chr.19: 9960092-9960092 on Build GRCh38
rs765724915 Chr.19: 9960391-9960391 on Build GRCh38
rs1015766132 Chr.19: 9960031-9960031 on Build GRCh38
rs374926094 Chr.19: 9960403-9960403 on Build GRCh38
rs1035484810 Chr.19: 9960286-9960286 on Build GRCh38
rs371498623 Chr.19: 9960402-9960402 on Build GRCh38
rs539576929 Chr.19: 9960336-9960336 on Build GRCh38
rs563370293 Chr.19: 9960133-9960133 on Build GRCh38
rs979439259 Chr.19: 9960113-9960113 on Build GRCh38
rs10406152 Chr.19: 9960387-9960387 on Build GRCh38
rs916835806 Chr.19: 9960080-9960080 on Build GRCh38
rs1030054046 Chr.19: 9959989-9959989 on Build GRCh38
rs201950607 Chr.19: 9959981-9959981 on Build GRCh38
rs530651438 Chr.19: 9960135-9960135 on Build GRCh38
rs1023864284 Chr.19: 9960335-9960335 on Build GRCh38
rs935393574 Chr.19: 9960121-9960121 on Build GRCh38
rs573174477 Chr.19: 9959960-9959960 on Build GRCh38
rs758738260 Chr.19: 9960399-9960399 on Build GRCh38
rs1051718074 Chr.19: 9960376-9960376 on Build GRCh38
rs562249517 Chr.19: 9960013-9960013 on Build GRCh38
rs969342702 Chr.19: 9960082-9960082 on Build GRCh38
rs988525619 Chr.19: 9960131-9960131 on Build GRCh38
rs576683390 Chr.19: 9960077-9960077 on Build GRCh38
rs375617796 Chr.19: 9960374-9960374 on Build GRCh38
rs970060042 Chr.19: 9960069-9960069 on Build GRCh38
rs1039558909 Chr.19: 9960186-9960186 on Build GRCh38
rs368612762 Chr.19: 9960383-9960383 on Build GRCh38
rs889129 Chr.19: 9960278-9960278 on Build GRCh38
rs925332380 Chr.19: 9960118-9960118 on Build GRCh38
rs556540201 Chr.19: 9959992-9959992 on Build GRCh38
rs778574580 Chr.19: 9960237-9960237 on Build GRCh38
rs369236503 Chr.19: 9960400-9960400 on Build GRCh38
rs775752009 Chr.19: 9960373-9960373 on Build GRCh38
rs374836707 Chr.19: 9960393-9960393 on Build GRCh38
rs1051872550 Chr.19: 9960161-9960161 on Build GRCh38
rs1027671033 Chr.19: 9960052-9960052 on Build GRCh38
rs749890637 Chr.19: 9960255-9960255 on Build GRCh38
rs965843247 Chr.19: 9960155-9960155 on Build GRCh38
rs188181882 Chr.19: 9960289-9960289 on Build GRCh38

View Full Product Details