Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9966443-9966971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL5A3
Entrez Gene ID
50509
Gene Name
collagen type V alpha 3
Gene Aliases
-
UniGene
Hs.235368
Gene Chromosome Location
Chr.19: 9959561-10010532 on Build GRCh38
Transcripts
NM_015719.3 XM_011528042.2 XM_017026849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3756245 c.4462G>C p.A1488P Chr.19: 9966743-9966743 on Build GRCh38
4619096 c.4576G>C p.E1526Q Chr.19: 9966629-9966629 on Build GRCh38
4073690 c.4592G>A p.R1531Q Chr.19: 9966613-9966613 on Build GRCh38
990330 c.4471G>A p.E1491K Chr.19: 9966734-9966734 on Build GRCh38
3692284 c.4566G>A p.S1522S Chr.19: 9966639-9966639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045727182 Chr.19: 9966789-9966789 on Build GRCh38
rs763206379 Chr.19: 9966494-9966494 on Build GRCh38
rs771222862 Chr.19: 9966652-9966652 on Build GRCh38
rs762054437 Chr.19: 9966518-9966518 on Build GRCh38
rs756176856 Chr.19: 9966519-9966519 on Build GRCh38
rs747211641 Chr.19: 9966475-9966475 on Build GRCh38
rs560024591 Chr.19: 9966912-9966912 on Build GRCh38
rs1028667843 Chr.19: 9966898-9966898 on Build GRCh38
rs1027315262 Chr.19: 9966649-9966649 on Build GRCh38
rs767139457 Chr.19: 9966517-9966517 on Build GRCh38
rs766096921 Chr.19: 9966495-9966495 on Build GRCh38
rs751202808 Chr.19: 9966503-9966503 on Build GRCh38
rs752303045 Chr.19: 9966718-9966718 on Build GRCh38
rs1009637251 Chr.19: 9966842-9966842 on Build GRCh38
rs1036517451 Chr.19: 9966780-9966780 on Build GRCh38
rs1014280993 Chr.19: 9966490-9966490 on Build GRCh38
rs757031604 Chr.19: 9966539-9966539 on Build GRCh38
rs1024873374 Chr.19: 9966532-9966532 on Build GRCh38
rs768803819 Chr.19: 9966613-9966613 on Build GRCh38
rs980741820 Chr.19: 9966553-9966553 on Build GRCh38
rs952386109 Chr.19: 9966911-9966911 on Build GRCh38
rs975770249 Chr.19: 9966662-9966662 on Build GRCh38
rs1050245430 Chr.19: 9966522-9966522 on Build GRCh38
rs574434275 Chr.19: 9966864-9966864 on Build GRCh38
rs778717136 Chr.19: 9966549-9966549 on Build GRCh38
rs1034461758 Chr.19: 9966691-9966691 on Build GRCh38
rs748217274 Chr.19: 9966630-9966630 on Build GRCh38
rs781320467 Chr.19: 9966624-9966624 on Build GRCh38
rs763088219 Chr.19: 9966473-9966474 on Build GRCh38
rs563370281 Chr.19: 9966472-9966472 on Build GRCh38
rs1000917709 Chr.19: 9966555-9966555 on Build GRCh38
rs1027261401 Chr.19: 9966930-9966931 on Build GRCh38
rs570072354 Chr.19: 9966787-9966787 on Build GRCh38
rs993508361 Chr.19: 9966481-9966482 on Build GRCh38
rs774752117 Chr.19: 9966638-9966638 on Build GRCh38
rs904611669 Chr.19: 9966502-9966502 on Build GRCh38
rs762315981 Chr.19: 9966486-9966486 on Build GRCh38
rs752249793 Chr.19: 9966518-9966518 on Build GRCh38
rs753737838 Chr.19: 9966727-9966727 on Build GRCh38
rs937580773 Chr.19: 9966477-9966477 on Build GRCh38
rs757156012 Chr.19: 9966734-9966734 on Build GRCh38
rs777861514 Chr.19: 9966757-9966757 on Build GRCh38
rs66715357 Chr.19: 9966820-9966820 on Build GRCh38
rs959143879 Chr.19: 9966702-9966702 on Build GRCh38
rs939742407 Chr.19: 9966924-9966924 on Build GRCh38
rs759329516 Chr.19: 9966678-9966678 on Build GRCh38
rs868593655 Chr.19: 9966548-9966548 on Build GRCh38
rs753704267 Chr.19: 9966527-9966527 on Build GRCh38
rs187502272 Chr.19: 9966469-9966469 on Build GRCh38
rs529042269 Chr.19: 9966719-9966719 on Build GRCh38
rs759215406 Chr.19: 9966510-9966510 on Build GRCh38
rs897732268 Chr.19: 9966877-9966877 on Build GRCh38
rs956509114 Chr.19: 9966596-9966596 on Build GRCh38
rs749772615 Chr.19: 9966768-9966768 on Build GRCh38
rs778581474 Chr.19: 9966735-9966735 on Build GRCh38
rs1005064639 Chr.19: 9966487-9966487 on Build GRCh38
rs770337172 Chr.19: 9966635-9966635 on Build GRCh38
rs748516330 Chr.19: 9966657-9966657 on Build GRCh38
rs201058798 Chr.19: 9966489-9966489 on Build GRCh38
rs1008797937 Chr.19: 9966603-9966603 on Build GRCh38
rs561653974 Chr.19: 9966674-9966674 on Build GRCh38
rs534195118 Chr.19: 9966824-9966824 on Build GRCh38
rs748268752 Chr.19: 9966747-9966747 on Build GRCh38
rs3745582 Chr.19: 9966482-9966482 on Build GRCh38
rs1019298477 Chr.19: 9966626-9966626 on Build GRCh38
rs76626564 Chr.19: 9966918-9966918 on Build GRCh38
rs763616893 Chr.19: 9966726-9966726 on Build GRCh38
rs776638360 Chr.19: 9966476-9966476 on Build GRCh38
rs764095590 Chr.19: 9966478-9966481 on Build GRCh38
rs528109510 Chr.19: 9966556-9966556 on Build GRCh38
rs73003436 Chr.19: 9966786-9966786 on Build GRCh38
rs774719655 Chr.19: 9966676-9966676 on Build GRCh38
rs773563954 Chr.19: 9966636-9966636 on Build GRCh38
rs980518142 Chr.19: 9966685-9966685 on Build GRCh38
rs970652729 Chr.19: 9966547-9966547 on Build GRCh38
rs920939158 Chr.19: 9966671-9966671 on Build GRCh38
rs774449371 Chr.19: 9966486-9966487 on Build GRCh38
rs939773737 Chr.19: 9966764-9966764 on Build GRCh38
rs763478169 Chr.19: 9966642-9966642 on Build GRCh38
rs747206052 Chr.19: 9966564-9966564 on Build GRCh38
rs192738745 Chr.19: 9966794-9966794 on Build GRCh38
rs3745583 Chr.19: 9966523-9966523 on Build GRCh38
rs769893805 Chr.19: 9966753-9966753 on Build GRCh38
rs3745584 Chr.19: 9966743-9966743 on Build GRCh38
rs975319851 Chr.19: 9966887-9966887 on Build GRCh38
rs765731246 Chr.19: 9966488-9966488 on Build GRCh38
rs972334856 Chr.19: 9966524-9966524 on Build GRCh38
rs1001285992 Chr.19: 9966816-9966816 on Build GRCh38
rs538420438 Chr.19: 9966878-9966878 on Build GRCh38
rs556086242 Chr.19: 9966859-9966859 on Build GRCh38
rs767264180 Chr.19: 9966686-9966686 on Build GRCh38
rs987954158 Chr.19: 9966612-9966612 on Build GRCh38
rs771019887 Chr.19: 9966832-9966832 on Build GRCh38
rs546443206 Chr.19: 9966641-9966641 on Build GRCh38
rs965628589 Chr.19: 9966862-9966862 on Build GRCh38

View Full Product Details