Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10085978-10086469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf66
Entrez Gene ID
55337
Gene Name
chromosome 19 open reading frame 66
Gene Aliases
RyDEN
UniGene
Hs.655613
Gene Chromosome Location
Chr.19: 10086119-10093252 on Build GRCh38
Transcripts
NM_001308277.1 NM_018381.3 XM_011528121.1 XM_011528122.1

SNP Detail

SNP ID Chromosome Location
rs1054111607 Chr.19: 10086117-10086117 on Build GRCh38
rs867635587 Chr.19: 10086033-10086033 on Build GRCh38
rs1029168446 Chr.19: 10086053-10086053 on Build GRCh38
rs923043647 Chr.19: 10086356-10086356 on Build GRCh38
rs987431314 Chr.19: 10086246-10086247 on Build GRCh38
rs999423201 Chr.19: 10086151-10086151 on Build GRCh38
rs761757913 Chr.19: 10086386-10086386 on Build GRCh38
rs767712921 Chr.19: 10086418-10086418 on Build GRCh38
rs552683675 Chr.19: 10086040-10086040 on Build GRCh38
rs1042583288 Chr.19: 10086174-10086174 on Build GRCh38
rs113654885 Chr.19: 10086027-10086027 on Build GRCh38
rs577390135 Chr.19: 10086219-10086219 on Build GRCh38
rs998805497 Chr.19: 10086000-10086000 on Build GRCh38
rs551004979 Chr.19: 10086421-10086421 on Build GRCh38
rs998356493 Chr.19: 10086190-10086190 on Build GRCh38
rs1016997710 Chr.19: 10086074-10086074 on Build GRCh38
rs927223554 Chr.19: 10086103-10086103 on Build GRCh38
rs772957680 Chr.19: 10086401-10086401 on Build GRCh38
rs1030367941 Chr.19: 10086206-10086206 on Build GRCh38
rs374201119 Chr.19: 10086353-10086353 on Build GRCh38
rs937041339 Chr.19: 10086107-10086107 on Build GRCh38
rs537378466 Chr.19: 10086058-10086058 on Build GRCh38
rs866235886 Chr.19: 10086170-10086170 on Build GRCh38
rs1051258674 Chr.19: 10086377-10086377 on Build GRCh38
rs1016860747 Chr.19: 10086239-10086239 on Build GRCh38
rs985643925 Chr.19: 10086070-10086070 on Build GRCh38
rs528324973 Chr.19: 10086267-10086267 on Build GRCh38
rs558065274 Chr.19: 10086009-10086009 on Build GRCh38
rs562187238 Chr.19: 10086352-10086352 on Build GRCh38
rs544711339 Chr.19: 10086032-10086033 on Build GRCh38
rs760849361 Chr.19: 10086223-10086223 on Build GRCh38
rs972707743 Chr.19: 10086080-10086080 on Build GRCh38
rs927390263 Chr.19: 10086319-10086319 on Build GRCh38
rs1056925105 Chr.19: 10086180-10086180 on Build GRCh38
rs534121488 Chr.19: 10086035-10086035 on Build GRCh38
rs564218080 Chr.19: 10086244-10086244 on Build GRCh38
rs1025481771 Chr.19: 10086263-10086263 on Build GRCh38
rs1019817534 Chr.19: 10086268-10086268 on Build GRCh38
rs992617466 Chr.19: 10086347-10086347 on Build GRCh38
rs945758019 Chr.19: 10086171-10086171 on Build GRCh38
rs540408167 Chr.19: 10086326-10086326 on Build GRCh38
rs981728902 Chr.19: 10086090-10086090 on Build GRCh38
rs769523931 Chr.19: 10086389-10086389 on Build GRCh38
rs556501613 Chr.19: 10086156-10086156 on Build GRCh38
rs949998257 Chr.19: 10086089-10086089 on Build GRCh38
rs945891654 Chr.19: 10086403-10086403 on Build GRCh38
rs557495444 Chr.19: 10086175-10086175 on Build GRCh38
rs760748514 Chr.19: 10086258-10086258 on Build GRCh38
rs904155652 Chr.19: 10086225-10086225 on Build GRCh38
rs879808268 Chr.19: 10086285-10086285 on Build GRCh38
rs567008896 Chr.19: 10086013-10086013 on Build GRCh38
rs761689385 Chr.19: 10086075-10086075 on Build GRCh38
rs1041596763 Chr.19: 10086406-10086406 on Build GRCh38
rs989835040 Chr.19: 10086314-10086314 on Build GRCh38
rs1028716637 Chr.19: 10086255-10086255 on Build GRCh38
rs574172603 Chr.19: 10086083-10086083 on Build GRCh38
rs952703592 Chr.19: 10086292-10086292 on Build GRCh38
rs890026566 Chr.19: 10086218-10086218 on Build GRCh38
rs958808414 Chr.19: 10086341-10086341 on Build GRCh38
rs759819693 Chr.19: 10086411-10086411 on Build GRCh38
rs927389877 Chr.19: 10086172-10086172 on Build GRCh38
rs541445169 Chr.19: 10086086-10086086 on Build GRCh38
rs958584718 Chr.19: 10086300-10086300 on Build GRCh38
rs965614246 Chr.19: 10086275-10086275 on Build GRCh38
rs946007092 Chr.19: 10086209-10086209 on Build GRCh38
rs895752336 Chr.19: 10086007-10086007 on Build GRCh38
rs927224679 Chr.19: 10086291-10086291 on Build GRCh38
rs972554123 Chr.19: 10086079-10086079 on Build GRCh38
rs971669073 Chr.19: 10086279-10086279 on Build GRCh38
rs994173734 Chr.19: 10086259-10086259 on Build GRCh38
rs934177106 Chr.19: 10086367-10086367 on Build GRCh38

View Full Product Details