Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10086849-10087354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf66
Entrez Gene ID
55337
Gene Name
chromosome 19 open reading frame 66
Gene Aliases
RyDEN
UniGene
Hs.655613
Gene Chromosome Location
Chr.19: 10086119-10093252 on Build GRCh38
Transcripts
NM_001308277.1 NM_018381.3 XM_011528121.1 XM_011528122.1 XM_017026934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
990380 c.190G>A p.A64T Chr.19: 10087295-10087295 on Build GRCh38
4404113 c.159A>C p.K53N Chr.19: 10087264-10087264 on Build GRCh38
990379 c.180C>T p.N60N Chr.19: 10087285-10087285 on Build GRCh38
5648036 c.124G>A p.V42M Chr.19: 10087031-10087031 on Build GRCh38
223969 c.148G>A p.V50I Chr.19: 10087253-10087253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759951116 Chr.19: 10087326-10087326 on Build GRCh38
rs975743757 Chr.19: 10086979-10086979 on Build GRCh38
rs773313777 Chr.19: 10087168-10087168 on Build GRCh38
rs1007382892 Chr.19: 10087156-10087156 on Build GRCh38
rs776766427 Chr.19: 10086937-10086937 on Build GRCh38
rs763624368 Chr.19: 10087313-10087313 on Build GRCh38
rs371273567 Chr.19: 10086912-10086912 on Build GRCh38
rs373427175 Chr.19: 10086929-10086929 on Build GRCh38
rs377121282 Chr.19: 10087071-10087071 on Build GRCh38
rs907169504 Chr.19: 10087085-10087085 on Build GRCh38
rs367666774 Chr.19: 10086955-10086955 on Build GRCh38
rs779955311 Chr.19: 10087286-10087288 on Build GRCh38
rs1054924224 Chr.19: 10087143-10087143 on Build GRCh38
rs758753002 Chr.19: 10087181-10087181 on Build GRCh38
rs998742600 Chr.19: 10087150-10087150 on Build GRCh38
rs747259503 Chr.19: 10087067-10087067 on Build GRCh38
rs752402517 Chr.19: 10087107-10087107 on Build GRCh38
rs765364092 Chr.19: 10087079-10087079 on Build GRCh38
rs950412742 Chr.19: 10087199-10087199 on Build GRCh38
rs777489589 Chr.19: 10086882-10086883 on Build GRCh38
rs751951044 Chr.19: 10087089-10087089 on Build GRCh38
rs780928721 Chr.19: 10086914-10086915 on Build GRCh38
rs748328640 Chr.19: 10087288-10087288 on Build GRCh38
rs745686607 Chr.19: 10087270-10087270 on Build GRCh38
rs770645927 Chr.19: 10086914-10086914 on Build GRCh38
rs757102628 Chr.19: 10086908-10086909 on Build GRCh38
rs750889944 Chr.19: 10086988-10086988 on Build GRCh38
rs376452273 Chr.19: 10086897-10086897 on Build GRCh38
rs544762485 Chr.19: 10087051-10087051 on Build GRCh38
rs774664890 Chr.19: 10087321-10087321 on Build GRCh38
rs754631429 Chr.19: 10087014-10087014 on Build GRCh38
rs748078962 Chr.19: 10086903-10086903 on Build GRCh38
rs998869969 Chr.19: 10087140-10087140 on Build GRCh38
rs374047761 Chr.19: 10087027-10087027 on Build GRCh38
rs780635544 Chr.19: 10087021-10087021 on Build GRCh38
rs777241925 Chr.19: 10087227-10087227 on Build GRCh38
rs779853424 Chr.19: 10086881-10086881 on Build GRCh38
rs749175062 Chr.19: 10087033-10087033 on Build GRCh38
rs997400780 Chr.19: 10087187-10087187 on Build GRCh38
rs751223122 Chr.19: 10087013-10087013 on Build GRCh38
rs778628779 Chr.19: 10087267-10087267 on Build GRCh38
rs763536514 Chr.19: 10087084-10087084 on Build GRCh38
rs540368072 Chr.19: 10087110-10087110 on Build GRCh38
rs773049741 Chr.19: 10086907-10086907 on Build GRCh38
rs560581119 Chr.19: 10087052-10087052 on Build GRCh38
rs749262992 Chr.19: 10087232-10087232 on Build GRCh38
rs562830935 Chr.19: 10087080-10087080 on Build GRCh38
rs766805025 Chr.19: 10087012-10087012 on Build GRCh38
rs771694772 Chr.19: 10087283-10087283 on Build GRCh38
rs903118672 Chr.19: 10087130-10087130 on Build GRCh38
rs773398219 Chr.19: 10087297-10087297 on Build GRCh38
rs200493899 Chr.19: 10087094-10087094 on Build GRCh38
rs764118380 Chr.19: 10086934-10086934 on Build GRCh38
rs759453711 Chr.19: 10087123-10087123 on Build GRCh38
rs770052476 Chr.19: 10087296-10087296 on Build GRCh38
rs774164300 Chr.19: 10087058-10087058 on Build GRCh38
rs368279366 Chr.19: 10087285-10087285 on Build GRCh38
rs963083041 Chr.19: 10087180-10087180 on Build GRCh38
rs999376515 Chr.19: 10086872-10086872 on Build GRCh38
rs766893959 Chr.19: 10087086-10087086 on Build GRCh38
rs780734067 Chr.19: 10086901-10086901 on Build GRCh38
rs567174163 Chr.19: 10087158-10087158 on Build GRCh38
rs754646972 Chr.19: 10087091-10087091 on Build GRCh38
rs751117105 Chr.19: 10086896-10086901 on Build GRCh38
rs769720043 Chr.19: 10086905-10086905 on Build GRCh38
rs370833713 Chr.19: 10087072-10087072 on Build GRCh38
rs967276319 Chr.19: 10086876-10086876 on Build GRCh38
rs986195235 Chr.19: 10087174-10087174 on Build GRCh38
rs1017329391 Chr.19: 10087176-10087176 on Build GRCh38
rs747682860 Chr.19: 10087022-10087022 on Build GRCh38
rs527942731 Chr.19: 10087286-10087286 on Build GRCh38
rs930231116 Chr.19: 10087247-10087247 on Build GRCh38
rs745632374 Chr.19: 10086915-10086915 on Build GRCh38
rs920056828 Chr.19: 10087222-10087222 on Build GRCh38
rs757181284 Chr.19: 10087243-10087243 on Build GRCh38
rs769772858 Chr.19: 10087090-10087109 on Build GRCh38
rs762281881 Chr.19: 10086954-10086954 on Build GRCh38
rs375793952 Chr.19: 10086982-10086982 on Build GRCh38
rs941498351 Chr.19: 10087048-10087048 on Build GRCh38
rs767213644 Chr.19: 10087093-10087093 on Build GRCh38
rs984489052 Chr.19: 10087219-10087219 on Build GRCh38
rs2227286 Chr.19: 10087083-10087083 on Build GRCh38
rs893232695 Chr.19: 10087146-10087146 on Build GRCh38
rs561825144 Chr.19: 10087011-10087011 on Build GRCh38
rs771458344 Chr.19: 10087320-10087320 on Build GRCh38
rs370331075 Chr.19: 10087024-10087024 on Build GRCh38
rs762750518 Chr.19: 10086908-10086908 on Build GRCh38
rs369693356 Chr.19: 10086902-10086902 on Build GRCh38
rs774196130 Chr.19: 10087096-10087096 on Build GRCh38
rs959137478 Chr.19: 10087309-10087309 on Build GRCh38
rs1030418352 Chr.19: 10087151-10087151 on Build GRCh38
rs775711032 Chr.19: 10086915-10086915 on Build GRCh38
rs1026235898 Chr.19: 10087189-10087189 on Build GRCh38
rs768077028 Chr.19: 10086900-10086900 on Build GRCh38
rs768908147 Chr.19: 10087070-10087070 on Build GRCh38

View Full Product Details