Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10092758-10093264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL6
Entrez Gene ID
83854
Gene Name
angiopoietin like 6
Gene Aliases
AGF, ARP5
UniGene
Hs.712850
Gene Chromosome Location
Chr.19: 10092337-10106194 on Build GRCh38
Transcripts
NM_001321411.1 NM_031917.2 XM_005260091.3 XM_011528347.2 XM_011528348.2 XM_011528349.2 XM_011528350.2 XM_017027347.1 NM_001308277.1 NM_018381.3 XM_011528121.1 XM_017026934.1

Gene Symbol
C19orf66
Entrez Gene ID
55337
Gene Name
chromosome 19 open reading frame 66
Gene Aliases
RyDEN
UniGene
Hs.655613
Gene Chromosome Location
Chr.19: 10086119-10093252 on Build GRCh38
Transcripts
NM_001321411.1 NM_031917.2 XM_005260091.3 XM_011528347.2 XM_011528348.2 XM_011528349.2 XM_011528350.2 XM_017027347.1 NM_001308277.1 NM_018381.3 XM_011528121.1 XM_017026934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599667 c.1239C>G p.Y413* Chr.19: 10092763-10092763 on Build GRCh38
6849018 c.1244G>A p.R415Q Chr.19: 10092758-10092758 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs569429706 Chr.19: 10092874-10092874 on Build GRCh38
rs755916692 Chr.19: 10093130-10093130 on Build GRCh38
rs989955180 Chr.19: 10092948-10092948 on Build GRCh38
rs200075423 Chr.19: 10092800-10092800 on Build GRCh38
rs371039233 Chr.19: 10092790-10092790 on Build GRCh38
rs765746562 Chr.19: 10092831-10092832 on Build GRCh38
rs745849563 Chr.19: 10092795-10092795 on Build GRCh38
rs903297975 Chr.19: 10093215-10093215 on Build GRCh38
rs994027468 Chr.19: 10093065-10093065 on Build GRCh38
rs951534534 Chr.19: 10093083-10093083 on Build GRCh38
rs777887335 Chr.19: 10092983-10092983 on Build GRCh38
rs946935587 Chr.19: 10092978-10092978 on Build GRCh38
rs558036247 Chr.19: 10092893-10092893 on Build GRCh38
rs375476177 Chr.19: 10092812-10092812 on Build GRCh38
rs551292743 Chr.19: 10092805-10092805 on Build GRCh38
rs745608694 Chr.19: 10093119-10093119 on Build GRCh38
rs1046906734 Chr.19: 10093069-10093069 on Build GRCh38
rs1044611 Chr.19: 10093185-10093185 on Build GRCh38
rs765443316 Chr.19: 10092787-10092787 on Build GRCh38
rs915876975 Chr.19: 10093139-10093140 on Build GRCh38
rs914289394 Chr.19: 10092965-10092965 on Build GRCh38
rs540047609 Chr.19: 10092879-10092879 on Build GRCh38
rs967510650 Chr.19: 10093217-10093217 on Build GRCh38
rs535015105 Chr.19: 10093219-10093219 on Build GRCh38
rs911257762 Chr.19: 10093018-10093018 on Build GRCh38
rs561692672 Chr.19: 10093033-10093033 on Build GRCh38
rs535087245 Chr.19: 10093155-10093155 on Build GRCh38
rs1002825237 Chr.19: 10093103-10093103 on Build GRCh38
rs982935293 Chr.19: 10093100-10093100 on Build GRCh38
rs528562847 Chr.19: 10093085-10093085 on Build GRCh38
rs771366968 Chr.19: 10092787-10092788 on Build GRCh38
rs924215319 Chr.19: 10092988-10092988 on Build GRCh38
rs777600596 Chr.19: 10092788-10092788 on Build GRCh38
rs1034265961 Chr.19: 10093114-10093114 on Build GRCh38
rs1051055681 Chr.19: 10093017-10093017 on Build GRCh38
rs756878062 Chr.19: 10092791-10092791 on Build GRCh38
rs898560629 Chr.19: 10093050-10093050 on Build GRCh38
rs998703469 Chr.19: 10092872-10092872 on Build GRCh38
rs756481281 Chr.19: 10092785-10092785 on Build GRCh38
rs866109968 Chr.19: 10092886-10092886 on Build GRCh38
rs934208322 Chr.19: 10092994-10092994 on Build GRCh38
rs189443545 Chr.19: 10093220-10093220 on Build GRCh38
rs778479876 Chr.19: 10092793-10092793 on Build GRCh38
rs1045790057 Chr.19: 10093193-10093193 on Build GRCh38
rs758594440 Chr.19: 10092858-10092858 on Build GRCh38
rs924079668 Chr.19: 10093236-10093236 on Build GRCh38
rs958608137 Chr.19: 10092896-10092896 on Build GRCh38
rs752838801 Chr.19: 10092836-10092836 on Build GRCh38
rs1038475687 Chr.19: 10093043-10093043 on Build GRCh38
rs570220793 Chr.19: 10092959-10092959 on Build GRCh38
rs534369126 Chr.19: 10093024-10093024 on Build GRCh38
rs1033976161 Chr.19: 10092887-10092887 on Build GRCh38
rs139370588 Chr.19: 10093101-10093101 on Build GRCh38
rs759925335 Chr.19: 10092787-10092790 on Build GRCh38
rs777219035 Chr.19: 10092786-10092787 on Build GRCh38

View Full Product Details