Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10096330-10096788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL6
Entrez Gene ID
83854
Gene Name
angiopoietin like 6
Gene Aliases
AGF, ARP5
UniGene
Hs.712850
Gene Chromosome Location
Chr.19: 10092337-10106194 on Build GRCh38
Transcripts
NM_001321411.1 NM_031917.2 XM_005260091.3 XM_011528347.2 XM_011528348.2 XM_011528349.2 XM_011528350.2 XM_017027347.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4140200 c.212G>T p.R71L Chr.19: 10096352-10096352 on Build GRCh38
4880324 c.136G>A p.G46S Chr.19: 10096428-10096428 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs962036607 Chr.19: 10096672-10096672 on Build GRCh38
rs74178192 Chr.19: 10096714-10096714 on Build GRCh38
rs776870275 Chr.19: 10096405-10096405 on Build GRCh38
rs953632169 Chr.19: 10096652-10096652 on Build GRCh38
rs931019018 Chr.19: 10096595-10096595 on Build GRCh38
rs752442516 Chr.19: 10096612-10096612 on Build GRCh38
rs769120820 Chr.19: 10096396-10096396 on Build GRCh38
rs896781163 Chr.19: 10096642-10096642 on Build GRCh38
rs531203056 Chr.19: 10096614-10096614 on Build GRCh38
rs181109012 Chr.19: 10096733-10096733 on Build GRCh38
rs972134275 Chr.19: 10096444-10096444 on Build GRCh38
rs776178141 Chr.19: 10096352-10096352 on Build GRCh38
rs917995516 Chr.19: 10096678-10096678 on Build GRCh38
rs925083378 Chr.19: 10096540-10096540 on Build GRCh38
rs1009308504 Chr.19: 10096605-10096605 on Build GRCh38
rs985298052 Chr.19: 10096717-10096717 on Build GRCh38
rs560892001 Chr.19: 10096427-10096427 on Build GRCh38
rs1048163957 Chr.19: 10096625-10096626 on Build GRCh38
rs1053343270 Chr.19: 10096569-10096569 on Build GRCh38
rs949227445 Chr.19: 10096459-10096459 on Build GRCh38
rs572134737 Chr.19: 10096402-10096402 on Build GRCh38
rs932514643 Chr.19: 10096542-10096542 on Build GRCh38
rs772872134 Chr.19: 10096452-10096452 on Build GRCh38
rs751565560 Chr.19: 10096549-10096549 on Build GRCh38
rs972165238 Chr.19: 10096676-10096676 on Build GRCh38
rs960684849 Chr.19: 10096428-10096428 on Build GRCh38
rs746538090 Chr.19: 10096407-10096407 on Build GRCh38
rs1033913810 Chr.19: 10096641-10096641 on Build GRCh38
rs1032456431 Chr.19: 10096681-10096681 on Build GRCh38
rs896835993 Chr.19: 10096589-10096589 on Build GRCh38
rs564817179 Chr.19: 10096665-10096665 on Build GRCh38
rs372470045 Chr.19: 10096538-10096538 on Build GRCh38
rs762498031 Chr.19: 10096460-10096460 on Build GRCh38
rs1045414937 Chr.19: 10096474-10096474 on Build GRCh38
rs936357188 Chr.19: 10096544-10096544 on Build GRCh38
rs1029673499 Chr.19: 10096650-10096650 on Build GRCh38
rs1036720141 Chr.19: 10096359-10096359 on Build GRCh38
rs10408244 Chr.19: 10096680-10096680 on Build GRCh38
rs560311003 Chr.19: 10096400-10096400 on Build GRCh38
rs550598317 Chr.19: 10096422-10096422 on Build GRCh38
rs376808065 Chr.19: 10096372-10096372 on Build GRCh38
rs767432273 Chr.19: 10096577-10096577 on Build GRCh38
rs532199359 Chr.19: 10096666-10096666 on Build GRCh38
rs997682086 Chr.19: 10096632-10096632 on Build GRCh38
rs184926120 Chr.19: 10096758-10096758 on Build GRCh38
rs764827031 Chr.19: 10096445-10096445 on Build GRCh38
rs912500842 Chr.19: 10096555-10096555 on Build GRCh38
rs113716218 Chr.19: 10096365-10096365 on Build GRCh38
rs1002404787 Chr.19: 10096632-10096633 on Build GRCh38
rs765845435 Chr.19: 10096461-10096461 on Build GRCh38
rs979542849 Chr.19: 10096467-10096467 on Build GRCh38
rs188290430 Chr.19: 10096630-10096630 on Build GRCh38
rs993670995 Chr.19: 10096395-10096395 on Build GRCh38
rs943702834 Chr.19: 10096564-10096564 on Build GRCh38
rs771122754 Chr.19: 10096651-10096651 on Build GRCh38
rs965232486 Chr.19: 10096740-10096740 on Build GRCh38
rs926529923 Chr.19: 10096517-10096517 on Build GRCh38
rs972629286 Chr.19: 10096743-10096743 on Build GRCh38
rs992184775 Chr.19: 10096431-10096431 on Build GRCh38
rs369675860 Chr.19: 10096408-10096408 on Build GRCh38
rs754898032 Chr.19: 10096574-10096574 on Build GRCh38
rs967231178 Chr.19: 10096449-10096449 on Build GRCh38
rs953992303 Chr.19: 10096704-10096704 on Build GRCh38
rs1021049166 Chr.19: 10096448-10096448 on Build GRCh38
rs1040797541 Chr.19: 10096621-10096621 on Build GRCh38

View Full Product Details