Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10102440-10102983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL6
Entrez Gene ID
83854
Gene Name
angiopoietin like 6
Gene Aliases
AGF, ARP5
UniGene
Hs.712850
Gene Chromosome Location
Chr.19: 10092337-10106194 on Build GRCh38
Transcripts
NM_001321411.1 NM_031917.2 XM_005260091.3 XM_011528347.2 XM_011528348.2 XM_011528349.2 XM_011528350.2 XM_017027347.1

SNP Detail

SNP ID Chromosome Location
rs182440152 Chr.19: 10102892-10102892 on Build GRCh38
rs902349523 Chr.19: 10102826-10102826 on Build GRCh38
rs985275043 Chr.19: 10102721-10102721 on Build GRCh38
rs917808218 Chr.19: 10102761-10102761 on Build GRCh38
rs980367910 Chr.19: 10102802-10102802 on Build GRCh38
rs557818794 Chr.19: 10102912-10102912 on Build GRCh38
rs76589705 Chr.19: 10102708-10102708 on Build GRCh38
rs1032142230 Chr.19: 10102834-10102834 on Build GRCh38
rs1054663611 Chr.19: 10102614-10102614 on Build GRCh38
rs953705225 Chr.19: 10102654-10102654 on Build GRCh38
rs1014091664 Chr.19: 10102505-10102505 on Build GRCh38
rs946179243 Chr.19: 10102832-10102832 on Build GRCh38
rs1042012045 Chr.19: 10102835-10102835 on Build GRCh38
rs1029085440 Chr.19: 10102718-10102718 on Build GRCh38
rs764627600 Chr.19: 10102490-10102490 on Build GRCh38
rs571416425 Chr.19: 10102595-10102595 on Build GRCh38
rs547564982 Chr.19: 10102663-10102663 on Build GRCh38
rs993696032 Chr.19: 10102916-10102916 on Build GRCh38
rs972907545 Chr.19: 10102733-10102733 on Build GRCh38
rs117484638 Chr.19: 10102911-10102911 on Build GRCh38
rs949026816 Chr.19: 10102561-10102561 on Build GRCh38
rs902056674 Chr.19: 10102841-10102841 on Build GRCh38
rs144713562 Chr.19: 10102735-10102735 on Build GRCh38
rs972390664 Chr.19: 10102547-10102547 on Build GRCh38
rs972492254 Chr.19: 10102885-10102885 on Build GRCh38
rs770074594 Chr.19: 10102817-10102818 on Build GRCh38
rs888064564 Chr.19: 10102787-10102787 on Build GRCh38
rs370636606 Chr.19: 10102812-10102812 on Build GRCh38
rs561002744 Chr.19: 10102467-10102467 on Build GRCh38
rs374706422 Chr.19: 10102868-10102868 on Build GRCh38
rs938255719 Chr.19: 10102933-10102933 on Build GRCh38
rs896475698 Chr.19: 10102943-10102943 on Build GRCh38
rs564007863 Chr.19: 10102790-10102790 on Build GRCh38
rs976194034 Chr.19: 10102465-10102465 on Build GRCh38
rs953924526 Chr.19: 10102854-10102854 on Build GRCh38
rs887759445 Chr.19: 10102470-10102470 on Build GRCh38
rs940654384 Chr.19: 10102537-10102537 on Build GRCh38
rs8109578 Chr.19: 10102478-10102478 on Build GRCh38
rs575397454 Chr.19: 10102585-10102585 on Build GRCh38
rs1044808664 Chr.19: 10102589-10102589 on Build GRCh38
rs1002245662 Chr.19: 10102471-10102471 on Build GRCh38
rs949452869 Chr.19: 10102956-10102956 on Build GRCh38
rs984924681 Chr.19: 10102905-10102905 on Build GRCh38
rs577508361 Chr.19: 10102827-10102827 on Build GRCh38
rs909132852 Chr.19: 10102528-10102528 on Build GRCh38
rs544337041 Chr.19: 10102824-10102824 on Build GRCh38
rs1016139025 Chr.19: 10102726-10102726 on Build GRCh38
rs755307702 Chr.19: 10102612-10102612 on Build GRCh38
rs1010511249 Chr.19: 10102681-10102681 on Build GRCh38
rs917610253 Chr.19: 10102551-10102551 on Build GRCh38
rs970652114 Chr.19: 10102942-10102942 on Build GRCh38
rs566100979 Chr.19: 10102665-10102665 on Build GRCh38
rs531460933 Chr.19: 10102944-10102944 on Build GRCh38
rs972017251 Chr.19: 10102755-10102755 on Build GRCh38
rs985107357 Chr.19: 10102664-10102664 on Build GRCh38
rs953211184 Chr.19: 10102479-10102479 on Build GRCh38
rs1024853558 Chr.19: 10102934-10102934 on Build GRCh38
rs1020943444 Chr.19: 10102682-10102682 on Build GRCh38
rs542190183 Chr.19: 10102693-10102693 on Build GRCh38
rs1019248469 Chr.19: 10102875-10102875 on Build GRCh38
rs73922324 Chr.19: 10102729-10102729 on Build GRCh38
rs893290857 Chr.19: 10102633-10102633 on Build GRCh38
rs78665538 Chr.19: 10102872-10102872 on Build GRCh38
rs985798520 Chr.19: 10102874-10102875 on Build GRCh38
rs929453318 Chr.19: 10102893-10102893 on Build GRCh38
rs1053926914 Chr.19: 10102819-10102819 on Build GRCh38
rs753087281 Chr.19: 10102818-10102818 on Build GRCh38
rs984602128 Chr.19: 10102480-10102480 on Build GRCh38
rs779303052 Chr.19: 10102719-10102719 on Build GRCh38
rs549528557 Chr.19: 10102497-10102497 on Build GRCh38
rs889116086 Chr.19: 10102881-10102881 on Build GRCh38
rs758858995 Chr.19: 10102825-10102825 on Build GRCh38
rs532517962 Chr.19: 10102600-10102600 on Build GRCh38

View Full Product Details