Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10106131-10106620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL6
Entrez Gene ID
83854
Gene Name
angiopoietin like 6
Gene Aliases
AGF, ARP5
UniGene
Hs.712850
Gene Chromosome Location
Chr.19: 10092337-10106194 on Build GRCh38
Transcripts
XM_011528347.2 NM_020230.5 NM_001040664.2 NM_001198690.1

Gene Symbol
PPAN
Entrez Gene ID
56342
Gene Name
peter pan homolog (Drosophila)
Gene Aliases
BXDC3, SSF, SSF-1, SSF1, SSF2
UniGene
Hs.14468
Gene Chromosome Location
Chr.19: 10106223-10111299 on Build GRCh38
Transcripts
XM_011528347.2 NM_020230.5 NM_001040664.2 NM_001198690.1

Gene Symbol
PPAN-P2RY11
Entrez Gene ID
692312
Gene Name
PPAN-P2RY11 readthrough
Gene Aliases
-
UniGene
Hs.14468
Gene Chromosome Location
Chr.19: 10106223-10115389 on Build GRCh38
Transcripts
XM_011528347.2 NM_020230.5 NM_001040664.2 NM_001198690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6303635 c.19-4G>A p.? Chr.19: 10106497-10106497 on Build GRCh38
7242353 c.30G>A p.Q10Q Chr.19: 10106512-10106512 on Build GRCh38
1287321 c.80delC p.A27fs*44 Chr.19: 10106562-10106562 on Build GRCh38
6303636 c.19-4G>A p.? Chr.19: 10106497-10106497 on Build GRCh38
1480502 c.67G>C p.E23Q Chr.19: 10106549-10106549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891472876 Chr.19: 10106271-10106271 on Build GRCh38
rs770172587 Chr.19: 10106407-10106407 on Build GRCh38
rs953907519 Chr.19: 10106506-10106506 on Build GRCh38
rs11559190 Chr.19: 10106572-10106572 on Build GRCh38
rs771593184 Chr.19: 10106566-10106566 on Build GRCh38
rs567384448 Chr.19: 10106303-10106303 on Build GRCh38
rs867448059 Chr.19: 10106299-10106299 on Build GRCh38
rs781326420 Chr.19: 10106597-10106597 on Build GRCh38
rs371223447 Chr.19: 10106599-10106599 on Build GRCh38
rs748481680 Chr.19: 10106389-10106389 on Build GRCh38
rs767389658 Chr.19: 10106442-10106442 on Build GRCh38
rs755035078 Chr.19: 10106524-10106524 on Build GRCh38
rs556089220 Chr.19: 10106344-10106344 on Build GRCh38
rs985166863 Chr.19: 10106578-10106578 on Build GRCh38
rs756229840 Chr.19: 10106309-10106309 on Build GRCh38
rs948572382 Chr.19: 10106214-10106214 on Build GRCh38
rs566696390 Chr.19: 10106259-10106259 on Build GRCh38
rs925061860 Chr.19: 10106319-10106319 on Build GRCh38
rs1054861239 Chr.19: 10106457-10106457 on Build GRCh38
rs925808278 Chr.19: 10106413-10106413 on Build GRCh38
rs944956383 Chr.19: 10106205-10106205 on Build GRCh38
rs542820412 Chr.19: 10106465-10106465 on Build GRCh38
rs948878951 Chr.19: 10106414-10106414 on Build GRCh38
rs555510856 Chr.19: 10106546-10106546 on Build GRCh38
rs758704411 Chr.19: 10106488-10106488 on Build GRCh38
rs373410012 Chr.19: 10106458-10106458 on Build GRCh38
rs984335806 Chr.19: 10106153-10106153 on Build GRCh38
rs758992938 Chr.19: 10106560-10106560 on Build GRCh38
rs746069216 Chr.19: 10106364-10106364 on Build GRCh38
rs373033480 Chr.19: 10106405-10106405 on Build GRCh38
rs935783782 Chr.19: 10106419-10106419 on Build GRCh38
rs762590771 Chr.19: 10106421-10106421 on Build GRCh38
rs780354576 Chr.19: 10106490-10106490 on Build GRCh38
rs775333581 Chr.19: 10106569-10106569 on Build GRCh38
rs966398268 Chr.19: 10106468-10106468 on Build GRCh38
rs980589460 Chr.19: 10106394-10106394 on Build GRCh38
rs369983672 Chr.19: 10106424-10106424 on Build GRCh38
rs187146844 Chr.19: 10106178-10106178 on Build GRCh38
rs551378309 Chr.19: 10106250-10106250 on Build GRCh38
rs1049254030 Chr.19: 10106554-10106554 on Build GRCh38
rs755052688 Chr.19: 10106592-10106592 on Build GRCh38
rs917315634 Chr.19: 10106176-10106176 on Build GRCh38
rs537856657 Chr.19: 10106313-10106313 on Build GRCh38
rs953829900 Chr.19: 10106361-10106361 on Build GRCh38
rs375786293 Chr.19: 10106509-10106509 on Build GRCh38
rs867284680 Chr.19: 10106273-10106273 on Build GRCh38
rs2290686 Chr.19: 10106301-10106301 on Build GRCh38
rs74178193 Chr.19: 10106237-10106237 on Build GRCh38
rs756658300 Chr.19: 10106601-10106601 on Build GRCh38
rs761319840 Chr.19: 10106362-10106363 on Build GRCh38
rs145490435 Chr.19: 10106423-10106423 on Build GRCh38
rs572538168 Chr.19: 10106158-10106158 on Build GRCh38
rs199958570 Chr.19: 10106384-10106384 on Build GRCh38
rs1040677808 Chr.19: 10106403-10106403 on Build GRCh38
rs961377278 Chr.19: 10106366-10106366 on Build GRCh38
rs868791919 Chr.19: 10106559-10106559 on Build GRCh38
rs1032021287 Chr.19: 10106484-10106484 on Build GRCh38
rs533306747 Chr.19: 10106240-10106240 on Build GRCh38
rs1053014298 Chr.19: 10106262-10106262 on Build GRCh38
rs993584064 Chr.19: 10106253-10106254 on Build GRCh38
rs2290685 Chr.19: 10106295-10106295 on Build GRCh38
rs979196160 Chr.19: 10106308-10106308 on Build GRCh38
rs766612887 Chr.19: 10106310-10106310 on Build GRCh38
rs900165027 Chr.19: 10106522-10106522 on Build GRCh38
rs73009299 Chr.19: 10106159-10106159 on Build GRCh38
rs970189463 Chr.19: 10106392-10106392 on Build GRCh38
rs1040065680 Chr.19: 10106473-10106473 on Build GRCh38
rs866508542 Chr.19: 10106497-10106497 on Build GRCh38
rs374860278 Chr.19: 10106557-10106557 on Build GRCh38
rs758614227 Chr.19: 10106368-10106368 on Build GRCh38
rs779031052 Chr.19: 10106350-10106350 on Build GRCh38
rs1020860711 Chr.19: 10106464-10106464 on Build GRCh38
rs1049647300 Chr.19: 10106257-10106257 on Build GRCh38
rs780263009 Chr.19: 10106379-10106379 on Build GRCh38
rs753687473 Chr.19: 10106574-10106574 on Build GRCh38
rs773955600 Chr.19: 10106558-10106558 on Build GRCh38
rs908584160 Chr.19: 10106157-10106157 on Build GRCh38
rs751806988 Chr.19: 10106587-10106587 on Build GRCh38
rs749597901 Chr.19: 10106552-10106552 on Build GRCh38
rs370147388 Chr.19: 10106540-10106540 on Build GRCh38
rs753986549 Chr.19: 10106377-10106377 on Build GRCh38
rs770123257 Chr.19: 10106545-10106545 on Build GRCh38
rs1033480152 Chr.19: 10106276-10106276 on Build GRCh38
rs750728403 Chr.19: 10106469-10106469 on Build GRCh38
rs760644168 Chr.19: 10106570-10106570 on Build GRCh38
rs573733797 Chr.19: 10106556-10106556 on Build GRCh38
rs759021092 Chr.19: 10106436-10106436 on Build GRCh38
rs766729415 Chr.19: 10106586-10106586 on Build GRCh38
rs561739565 Chr.19: 10106163-10106163 on Build GRCh38
rs781598891 Chr.19: 10106536-10106536 on Build GRCh38
rs1028531433 Chr.19: 10106306-10106306 on Build GRCh38
rs141612097 Chr.19: 10106239-10106239 on Build GRCh38
rs772891041 Chr.19: 10106408-10106408 on Build GRCh38
rs761656993 Chr.19: 10106584-10106584 on Build GRCh38
rs544188416 Chr.19: 10106219-10106219 on Build GRCh38
rs1002304897 Chr.19: 10106455-10106455 on Build GRCh38

View Full Product Details