Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10115605-10116107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3G
Entrez Gene ID
8666
Gene Name
eukaryotic translation initiation factor 3 subunit G
Gene Aliases
EIF3-P42, EIF3S4, eIF3-delta, eIF3-p44
UniGene
Hs.529059
Gene Chromosome Location
Chr.19: 10115014-10119923 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
990425 c.621G>T p.Q207H Chr.19: 10116049-10116049 on Build GRCh38
4277279 c.840+3G>A p.? Chr.19: 10115681-10115681 on Build GRCh38
4277278 c.840+6C>T p.? Chr.19: 10115678-10115678 on Build GRCh38
3528087 c.777C>T p.F259F Chr.19: 10115747-10115747 on Build GRCh38
5028490 c.724C>T p.R242C Chr.19: 10115800-10115800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs71188883 Chr.19: 10115665-10115668 on Build GRCh38
rs374215335 Chr.19: 10115853-10115853 on Build GRCh38
rs17850344 Chr.19: 10115732-10115732 on Build GRCh38
rs149009824 Chr.19: 10116047-10116047 on Build GRCh38
rs766409457 Chr.19: 10115779-10115779 on Build GRCh38
rs140952996 Chr.19: 10115819-10115819 on Build GRCh38
rs770681828 Chr.19: 10116012-10116012 on Build GRCh38
rs778229939 Chr.19: 10115677-10115677 on Build GRCh38
rs112317953 Chr.19: 10116077-10116077 on Build GRCh38
rs773964286 Chr.19: 10115960-10115960 on Build GRCh38
rs148928911 Chr.19: 10115949-10115951 on Build GRCh38
rs1009104725 Chr.19: 10115886-10115886 on Build GRCh38
rs769120649 Chr.19: 10115941-10115941 on Build GRCh38
rs771061841 Chr.19: 10115705-10115705 on Build GRCh38
rs754831371 Chr.19: 10115810-10115810 on Build GRCh38
rs150759321 Chr.19: 10115676-10115679 on Build GRCh38
rs530986361 Chr.19: 10115628-10115628 on Build GRCh38
rs770201819 Chr.19: 10115695-10115695 on Build GRCh38
rs760367729 Chr.19: 10115662-10115662 on Build GRCh38
rs759591490 Chr.19: 10115963-10115963 on Build GRCh38
rs370607828 Chr.19: 10116031-10116031 on Build GRCh38
rs774251071 Chr.19: 10115716-10115716 on Build GRCh38
rs371439478 Chr.19: 10115992-10115992 on Build GRCh38
rs756797715 Chr.19: 10115668-10115668 on Build GRCh38
rs201999477 Chr.19: 10115939-10115939 on Build GRCh38
rs778713206 Chr.19: 10116080-10116080 on Build GRCh38
rs776511542 Chr.19: 10115759-10115759 on Build GRCh38
rs767456129 Chr.19: 10115916-10115916 on Build GRCh38
rs754922352 Chr.19: 10115872-10115872 on Build GRCh38
rs772631789 Chr.19: 10115838-10115838 on Build GRCh38
rs757556891 Chr.19: 10115923-10115923 on Build GRCh38
rs916532401 Chr.19: 10115878-10115878 on Build GRCh38
rs748451179 Chr.19: 10115952-10115952 on Build GRCh38
rs74555430 Chr.19: 10115845-10115845 on Build GRCh38
rs1050049016 Chr.19: 10115963-10115963 on Build GRCh38
rs767951767 Chr.19: 10115656-10115656 on Build GRCh38
rs868284931 Chr.19: 10115809-10115809 on Build GRCh38
rs375499220 Chr.19: 10115964-10115964 on Build GRCh38
rs763721672 Chr.19: 10115663-10115663 on Build GRCh38
rs763211394 Chr.19: 10116037-10116037 on Build GRCh38
rs761565482 Chr.19: 10115834-10115834 on Build GRCh38
rs781660556 Chr.19: 10115950-10115950 on Build GRCh38
rs752411883 Chr.19: 10115919-10115919 on Build GRCh38
rs757557182 Chr.19: 10115951-10115953 on Build GRCh38
rs564599932 Chr.19: 10115637-10115637 on Build GRCh38
rs763113798 Chr.19: 10115647-10115647 on Build GRCh38
rs192366729 Chr.19: 10115985-10115985 on Build GRCh38
rs772956400 Chr.19: 10115659-10115661 on Build GRCh38
rs764778081 Chr.19: 10115859-10115859 on Build GRCh38
rs758233095 Chr.19: 10115681-10115681 on Build GRCh38
rs199542850 Chr.19: 10115762-10115762 on Build GRCh38
rs746781564 Chr.19: 10115690-10115690 on Build GRCh38
rs776280151 Chr.19: 10115746-10115746 on Build GRCh38
rs766352700 Chr.19: 10115924-10115924 on Build GRCh38
rs886250382 Chr.19: 10115642-10115642 on Build GRCh38
rs947940226 Chr.19: 10115889-10115889 on Build GRCh38
rs755540229 Chr.19: 10115995-10115995 on Build GRCh38
rs771542095 Chr.19: 10116014-10116014 on Build GRCh38
rs751953776 Chr.19: 10115849-10115849 on Build GRCh38
rs200368684 Chr.19: 10115731-10115731 on Build GRCh38
rs146113547 Chr.19: 10115937-10115937 on Build GRCh38
rs765425850 Chr.19: 10115664-10115665 on Build GRCh38
rs762550205 Chr.19: 10115865-10115865 on Build GRCh38
rs780148640 Chr.19: 10115991-10115991 on Build GRCh38
rs201796922 Chr.19: 10115648-10115648 on Build GRCh38
rs778156862 Chr.19: 10116009-10116009 on Build GRCh38
rs753369453 Chr.19: 10115859-10115860 on Build GRCh38
rs763889903 Chr.19: 10115971-10115971 on Build GRCh38
rs142974473 Chr.19: 10115774-10115774 on Build GRCh38
rs770074103 Chr.19: 10116003-10116003 on Build GRCh38
rs766578639 Chr.19: 10116042-10116042 on Build GRCh38
rs375266384 Chr.19: 10116061-10116061 on Build GRCh38
rs904468043 Chr.19: 10115862-10115863 on Build GRCh38
rs199504887 Chr.19: 10115837-10115837 on Build GRCh38
rs374292482 Chr.19: 10116067-10116067 on Build GRCh38
rs753166561 Chr.19: 10115667-10115668 on Build GRCh38
rs780039345 Chr.19: 10115684-10115684 on Build GRCh38
rs368114638 Chr.19: 10115904-10115904 on Build GRCh38
rs374203842 Chr.19: 10115879-10115896 on Build GRCh38
rs147076389 Chr.19: 10116052-10116052 on Build GRCh38
rs117516437 Chr.19: 10115903-10115903 on Build GRCh38
rs776867477 Chr.19: 10115662-10115663 on Build GRCh38
rs758579235 Chr.19: 10115973-10115973 on Build GRCh38
rs150270880 Chr.19: 10116000-10116000 on Build GRCh38
rs759907311 Chr.19: 10115653-10115653 on Build GRCh38
rs758688972 Chr.19: 10115810-10115812 on Build GRCh38
rs144308529 Chr.19: 10115765-10115765 on Build GRCh38
rs200490441 Chr.19: 10116010-10116010 on Build GRCh38
rs755709143 Chr.19: 10115857-10115858 on Build GRCh38
rs1056224917 Chr.19: 10115877-10115877 on Build GRCh38
rs1048477123 Chr.19: 10115650-10115650 on Build GRCh38
rs746033497 Chr.19: 10115929-10115929 on Build GRCh38
rs760391314 Chr.19: 10115970-10115970 on Build GRCh38
rs760011858 Chr.19: 10115667-10115670 on Build GRCh38
rs557339769 Chr.19: 10115898-10115898 on Build GRCh38
rs371210294 Chr.19: 10115851-10115851 on Build GRCh38
rs774932193 Chr.19: 10115651-10115651 on Build GRCh38
rs188476640 Chr.19: 10115738-10115738 on Build GRCh38
rs375004089 Chr.19: 10115633-10115633 on Build GRCh38
rs112076821 Chr.19: 10115823-10115823 on Build GRCh38
rs1018700772 Chr.19: 10115892-10115892 on Build GRCh38
rs569846791 Chr.19: 10115816-10115816 on Build GRCh38
rs757386242 Chr.19: 10115830-10115830 on Build GRCh38
rs952182350 Chr.19: 10115842-10115842 on Build GRCh38
rs888697172 Chr.19: 10115998-10115998 on Build GRCh38
rs753414953 Chr.19: 10115667-10115667 on Build GRCh38
rs775982765 Chr.19: 10115843-10115843 on Build GRCh38
rs750397804 Chr.19: 10115678-10115678 on Build GRCh38
rs761323264 Chr.19: 10115750-10115750 on Build GRCh38
rs375316477 Chr.19: 10116072-10116072 on Build GRCh38
rs766644786 Chr.19: 10115663-10115664 on Build GRCh38
rs542533375 Chr.19: 10115897-10115897 on Build GRCh38
rs370993731 Chr.19: 10115953-10115953 on Build GRCh38
rs764671159 Chr.19: 10115847-10115848 on Build GRCh38
rs775888897 Chr.19: 10115721-10115721 on Build GRCh38
rs530819290 Chr.19: 10116026-10116026 on Build GRCh38
rs575445249 Chr.19: 10115899-10115899 on Build GRCh38
rs369841100 Chr.19: 10115720-10115720 on Build GRCh38
rs550273947 Chr.19: 10115860-10115877 on Build GRCh38
rs999830510 Chr.19: 10115711-10115711 on Build GRCh38
rs750633234 Chr.19: 10115972-10115972 on Build GRCh38
rs781549844 Chr.19: 10115999-10115999 on Build GRCh38
rs548473194 Chr.19: 10116013-10116013 on Build GRCh38
rs1043391593 Chr.19: 10115891-10115891 on Build GRCh38
rs574722029 Chr.19: 10115873-10115873 on Build GRCh38
rs549483404 Chr.19: 10115636-10115636 on Build GRCh38
rs772746591 Chr.19: 10115956-10115956 on Build GRCh38
rs372065668 Chr.19: 10116016-10116016 on Build GRCh38
rs1019087864 Chr.19: 10115925-10115925 on Build GRCh38
rs377152440 Chr.19: 10115848-10115849 on Build GRCh38
rs771915899 Chr.19: 10115629-10115629 on Build GRCh38
rs367686709 Chr.19: 10115643-10115643 on Build GRCh38
rs28510792 Chr.19: 10116025-10116025 on Build GRCh38
rs753028203 Chr.19: 10115659-10115659 on Build GRCh38
rs895723640 Chr.19: 10115664-10115664 on Build GRCh38
rs146486140 Chr.19: 10115700-10115700 on Build GRCh38
rs765927181 Chr.19: 10115870-10115870 on Build GRCh38
rs772839578 Chr.19: 10115863-10115863 on Build GRCh38
rs760613402 Chr.19: 10116022-10116022 on Build GRCh38
rs751469081 Chr.19: 10115780-10115780 on Build GRCh38
rs117805948 Chr.19: 10115717-10115717 on Build GRCh38
rs535149710 Chr.19: 10116078-10116078 on Build GRCh38
rs760753735 Chr.19: 10115663-10115666 on Build GRCh38
rs776820864 Chr.19: 10115850-10115850 on Build GRCh38
rs751709107 Chr.19: 10116049-10116049 on Build GRCh38
rs752604243 Chr.19: 10115967-10115967 on Build GRCh38
rs778674406 Chr.19: 10116064-10116064 on Build GRCh38
rs935955265 Chr.19: 10115962-10115962 on Build GRCh38

View Full Product Details