Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10116716-10117207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3G
Entrez Gene ID
8666
Gene Name
eukaryotic translation initiation factor 3 subunit G
Gene Aliases
EIF3-P42, EIF3S4, eIF3-delta, eIF3-p44
UniGene
Hs.529059
Gene Chromosome Location
Chr.19: 10115014-10119923 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5590737 c.498C>T p.T166T Chr.19: 10116897-10116897 on Build GRCh38
6414119 c.574G>T p.E192* Chr.19: 10116821-10116821 on Build GRCh38
5520931 c.406G>T p.D136Y Chr.19: 10116989-10116989 on Build GRCh38
6403962 c.322T>A p.S108T Chr.19: 10117167-10117167 on Build GRCh38
3528088 c.336C>T p.P112P Chr.19: 10117153-10117153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025098381 Chr.19: 10116818-10116819 on Build GRCh38
rs750780486 Chr.19: 10116873-10116873 on Build GRCh38
rs545769118 Chr.19: 10116760-10116760 on Build GRCh38
rs746516276 Chr.19: 10116785-10116785 on Build GRCh38
rs763384052 Chr.19: 10117010-10117010 on Build GRCh38
rs549623816 Chr.19: 10117021-10117021 on Build GRCh38
rs749122290 Chr.19: 10117037-10117037 on Build GRCh38
rs763322919 Chr.19: 10117068-10117068 on Build GRCh38
rs561764575 Chr.19: 10116844-10116844 on Build GRCh38
rs961381126 Chr.19: 10116903-10116903 on Build GRCh38
rs141171264 Chr.19: 10117150-10117150 on Build GRCh38
rs369083840 Chr.19: 10117060-10117060 on Build GRCh38
rs900979934 Chr.19: 10117048-10117048 on Build GRCh38
rs368376665 Chr.19: 10116894-10116894 on Build GRCh38
rs774554344 Chr.19: 10117078-10117078 on Build GRCh38
rs112649117 Chr.19: 10117051-10117051 on Build GRCh38
rs763462020 Chr.19: 10117110-10117110 on Build GRCh38
rs768886941 Chr.19: 10116986-10116986 on Build GRCh38
rs777767578 Chr.19: 10117027-10117027 on Build GRCh38
rs770781857 Chr.19: 10117042-10117042 on Build GRCh38
rs148192612 Chr.19: 10116975-10116975 on Build GRCh38
rs766800494 Chr.19: 10117077-10117077 on Build GRCh38
rs757111239 Chr.19: 10117126-10117126 on Build GRCh38
rs1028007124 Chr.19: 10116826-10116826 on Build GRCh38
rs370039447 Chr.19: 10117173-10117173 on Build GRCh38
rs936726045 Chr.19: 10117005-10117005 on Build GRCh38
rs566842898 Chr.19: 10117001-10117001 on Build GRCh38
rs376735435 Chr.19: 10117014-10117014 on Build GRCh38
rs922208764 Chr.19: 10117012-10117012 on Build GRCh38
rs767148811 Chr.19: 10117084-10117084 on Build GRCh38
rs143486896 Chr.19: 10116833-10116833 on Build GRCh38
rs142682262 Chr.19: 10117174-10117174 on Build GRCh38
rs748023262 Chr.19: 10116797-10116797 on Build GRCh38
rs745548190 Chr.19: 10116766-10116766 on Build GRCh38
rs965292110 Chr.19: 10116795-10116795 on Build GRCh38
rs145896417 Chr.19: 10116963-10116963 on Build GRCh38
rs371843551 Chr.19: 10117000-10117000 on Build GRCh38
rs961171414 Chr.19: 10117152-10117152 on Build GRCh38
rs544307400 Chr.19: 10116882-10116882 on Build GRCh38
rs781570961 Chr.19: 10117142-10117142 on Build GRCh38
rs888351041 Chr.19: 10117102-10117102 on Build GRCh38
rs767761417 Chr.19: 10116821-10116821 on Build GRCh38
rs747274062 Chr.19: 10117047-10117047 on Build GRCh38
rs1054189173 Chr.19: 10117008-10117008 on Build GRCh38
rs772957450 Chr.19: 10116805-10116805 on Build GRCh38
rs764693573 Chr.19: 10116759-10116759 on Build GRCh38
rs745645967 Chr.19: 10117141-10117141 on Build GRCh38
rs1009438775 Chr.19: 10116741-10116741 on Build GRCh38
rs748326626 Chr.19: 10117147-10117147 on Build GRCh38
rs569020571 Chr.19: 10117135-10117135 on Build GRCh38
rs776823093 Chr.19: 10116995-10116995 on Build GRCh38
rs765737865 Chr.19: 10116871-10116871 on Build GRCh38
rs150720892 Chr.19: 10117155-10117155 on Build GRCh38
rs376758358 Chr.19: 10117091-10117091 on Build GRCh38
rs775495349 Chr.19: 10116827-10116827 on Build GRCh38
rs139174106 Chr.19: 10117171-10117171 on Build GRCh38
rs1047846088 Chr.19: 10116738-10116738 on Build GRCh38
rs190019354 Chr.19: 10117063-10117063 on Build GRCh38
rs776558448 Chr.19: 10116791-10116791 on Build GRCh38
rs185092700 Chr.19: 10116846-10116846 on Build GRCh38
rs368540466 Chr.19: 10117022-10117022 on Build GRCh38
rs757797322 Chr.19: 10116870-10116870 on Build GRCh38
rs770564468 Chr.19: 10116966-10116966 on Build GRCh38
rs770821575 Chr.19: 10117149-10117150 on Build GRCh38
rs372463482 Chr.19: 10116783-10116783 on Build GRCh38
rs11558272 Chr.19: 10117136-10117136 on Build GRCh38
rs759241458 Chr.19: 10117079-10117079 on Build GRCh38
rs756633803 Chr.19: 10116751-10116751 on Build GRCh38
rs756201398 Chr.19: 10116999-10116999 on Build GRCh38
rs974767860 Chr.19: 10117006-10117008 on Build GRCh38
rs369663424 Chr.19: 10117103-10117103 on Build GRCh38
rs200081419 Chr.19: 10116912-10116912 on Build GRCh38
rs528984502 Chr.19: 10116804-10116804 on Build GRCh38
rs377318103 Chr.19: 10117114-10117114 on Build GRCh38
rs748121308 Chr.19: 10116929-10116929 on Build GRCh38
rs771325052 Chr.19: 10117156-10117156 on Build GRCh38
rs777497675 Chr.19: 10116802-10116802 on Build GRCh38
rs143183271 Chr.19: 10117117-10117117 on Build GRCh38
rs201238271 Chr.19: 10117098-10117098 on Build GRCh38
rs751110674 Chr.19: 10116909-10116909 on Build GRCh38
rs533246672 Chr.19: 10116948-10116948 on Build GRCh38
rs756106816 Chr.19: 10117023-10117023 on Build GRCh38
rs766599531 Chr.19: 10117013-10117013 on Build GRCh38
rs1045318368 Chr.19: 10116997-10116997 on Build GRCh38
rs192747251 Chr.19: 10116740-10116740 on Build GRCh38
rs372765542 Chr.19: 10117043-10117043 on Build GRCh38
rs540508075 Chr.19: 10116786-10116786 on Build GRCh38
rs564068319 Chr.19: 10116765-10116765 on Build GRCh38
rs573098250 Chr.19: 10116781-10116781 on Build GRCh38
rs775793343 Chr.19: 10116973-10116973 on Build GRCh38
rs757707392 Chr.19: 10116763-10116763 on Build GRCh38
rs746905307 Chr.19: 10117126-10117128 on Build GRCh38
rs773787173 Chr.19: 10117064-10117064 on Build GRCh38
rs983508264 Chr.19: 10117036-10117036 on Build GRCh38
rs759722830 Chr.19: 10116815-10116815 on Build GRCh38
rs976719050 Chr.19: 10117004-10117004 on Build GRCh38
rs750820266 Chr.19: 10117007-10117007 on Build GRCh38
rs777784165 Chr.19: 10117006-10117006 on Build GRCh38
rs200494795 Chr.19: 10116933-10116933 on Build GRCh38
rs908318457 Chr.19: 10116897-10116897 on Build GRCh38
rs761979960 Chr.19: 10117062-10117062 on Build GRCh38

View Full Product Details