Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10928876-10929366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf52
Entrez Gene ID
90580
Gene Name
chromosome 19 open reading frame 52
Gene Aliases
-
UniGene
Hs.164026
Gene Chromosome Location
Chr.19: 10928748-10930241 on Build GRCh38
Transcripts
NM_138358.2 NM_024029.4

Gene Symbol
YIPF2
Entrez Gene ID
78992
Gene Name
Yip1 domain family member 2
Gene Aliases
FinGER2
UniGene
Hs.164026
Gene Chromosome Location
Chr.19: 10922185-10929012 on Build GRCh38
Transcripts
NM_138358.2 NM_024029.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7242513 c.69G>C p.A23A Chr.19: 10928891-10928891 on Build GRCh38
4955760 c.120G>T p.R40R Chr.19: 10929039-10929039 on Build GRCh38
6374783 c.235A>T p.S79C Chr.19: 10929154-10929154 on Build GRCh38
4960327 c.274G>A p.G92R Chr.19: 10929193-10929193 on Build GRCh38
6418980 c.373G>A p.V125I Chr.19: 10929292-10929292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972795751 Chr.19: 10928976-10928976 on Build GRCh38
rs551153094 Chr.19: 10929159-10929159 on Build GRCh38
rs1044531871 Chr.19: 10929157-10929157 on Build GRCh38
rs903576605 Chr.19: 10929115-10929115 on Build GRCh38
rs769474996 Chr.19: 10929002-10929002 on Build GRCh38
rs759198868 Chr.19: 10928897-10928897 on Build GRCh38
rs897179089 Chr.19: 10929198-10929198 on Build GRCh38
rs764585177 Chr.19: 10928900-10928900 on Build GRCh38
rs964975979 Chr.19: 10928978-10928978 on Build GRCh38
rs762745240 Chr.19: 10929007-10929007 on Build GRCh38
rs749280252 Chr.19: 10929229-10929229 on Build GRCh38
rs952735447 Chr.19: 10929211-10929211 on Build GRCh38
rs1042131862 Chr.19: 10929099-10929099 on Build GRCh38
rs1008266532 Chr.19: 10928923-10928923 on Build GRCh38
rs200318904 Chr.19: 10929329-10929329 on Build GRCh38
rs373414405 Chr.19: 10928925-10928925 on Build GRCh38
rs755111488 Chr.19: 10929318-10929318 on Build GRCh38
rs762303431 Chr.19: 10929091-10929091 on Build GRCh38
rs777646969 Chr.19: 10929336-10929336 on Build GRCh38
rs773960098 Chr.19: 10929036-10929036 on Build GRCh38
rs752972395 Chr.19: 10929327-10929327 on Build GRCh38
rs767440748 Chr.19: 10929321-10929321 on Build GRCh38
rs533053035 Chr.19: 10929133-10929133 on Build GRCh38
rs1027081356 Chr.19: 10929207-10929207 on Build GRCh38
rs775027123 Chr.19: 10928938-10928958 on Build GRCh38
rs757088126 Chr.19: 10929342-10929342 on Build GRCh38
rs370473955 Chr.19: 10929027-10929027 on Build GRCh38
rs752026693 Chr.19: 10929311-10929311 on Build GRCh38
rs116937805 Chr.19: 10929048-10929048 on Build GRCh38
rs780985635 Chr.19: 10928996-10928996 on Build GRCh38
rs775269582 Chr.19: 10929256-10929256 on Build GRCh38
rs771702181 Chr.19: 10929254-10929254 on Build GRCh38
rs775250686 Chr.19: 10929181-10929181 on Build GRCh38
rs771000885 Chr.19: 10928999-10928999 on Build GRCh38
rs779512392 Chr.19: 10928916-10928916 on Build GRCh38
rs755953874 Chr.19: 10929201-10929201 on Build GRCh38
rs765505019 Chr.19: 10929095-10929095 on Build GRCh38
rs774409982 Chr.19: 10929247-10929247 on Build GRCh38
rs777369006 Chr.19: 10929202-10929202 on Build GRCh38
rs768822045 Chr.19: 10928983-10928983 on Build GRCh38
rs750954172 Chr.19: 10929100-10929100 on Build GRCh38
rs980235010 Chr.19: 10929021-10929021 on Build GRCh38
rs549144776 Chr.19: 10929308-10929308 on Build GRCh38
rs527929447 Chr.19: 10929296-10929296 on Build GRCh38
rs1019636250 Chr.19: 10929197-10929197 on Build GRCh38
rs945018907 Chr.19: 10929082-10929082 on Build GRCh38
rs750887649 Chr.19: 10928920-10928920 on Build GRCh38
rs11538076 Chr.19: 10929241-10929241 on Build GRCh38
rs748615198 Chr.19: 10929334-10929346 on Build GRCh38
rs1009743376 Chr.19: 10928943-10928943 on Build GRCh38
rs956296950 Chr.19: 10928895-10928895 on Build GRCh38
rs758834737 Chr.19: 10929118-10929118 on Build GRCh38
rs766918046 Chr.19: 10929309-10929309 on Build GRCh38
rs900881790 Chr.19: 10929259-10929259 on Build GRCh38
rs770573684 Chr.19: 10929035-10929035 on Build GRCh38
rs752262540 Chr.19: 10929083-10929083 on Build GRCh38
rs762492490 Chr.19: 10929249-10929249 on Build GRCh38
rs927572143 Chr.19: 10929038-10929038 on Build GRCh38
rs763468858 Chr.19: 10929304-10929304 on Build GRCh38
rs751586374 Chr.19: 10929134-10929134 on Build GRCh38
rs936511059 Chr.19: 10929125-10929125 on Build GRCh38
rs113408828 Chr.19: 10929024-10929024 on Build GRCh38
rs566222619 Chr.19: 10929290-10929290 on Build GRCh38
rs760187177 Chr.19: 10929269-10929269 on Build GRCh38
rs747231999 Chr.19: 10928972-10928972 on Build GRCh38
rs748308054 Chr.19: 10929191-10929191 on Build GRCh38
rs1034925273 Chr.19: 10929223-10929223 on Build GRCh38
rs765637963 Chr.19: 10929283-10929283 on Build GRCh38
rs938932170 Chr.19: 10929042-10929042 on Build GRCh38
rs745429855 Chr.19: 10929017-10929017 on Build GRCh38
rs781222031 Chr.19: 10929004-10929004 on Build GRCh38
rs917554295 Chr.19: 10929079-10929079 on Build GRCh38
rs749145423 Chr.19: 10929338-10929338 on Build GRCh38
rs755025765 Chr.19: 10929143-10929143 on Build GRCh38
rs759284814 Chr.19: 10929047-10929047 on Build GRCh38
rs754184331 Chr.19: 10928902-10928902 on Build GRCh38
rs913455030 Chr.19: 10929270-10929270 on Build GRCh38
rs780049386 Chr.19: 10928966-10928966 on Build GRCh38
rs1028055153 Chr.19: 10929317-10929317 on Build GRCh38
rs936190717 Chr.19: 10929217-10929217 on Build GRCh38
rs544209641 Chr.19: 10928894-10928894 on Build GRCh38
rs972620783 Chr.19: 10929072-10929072 on Build GRCh38
rs892598670 Chr.19: 10928918-10928918 on Build GRCh38
rs952870906 Chr.19: 10928992-10928993 on Build GRCh38
rs757541992 Chr.19: 10928912-10928912 on Build GRCh38
rs980337108 Chr.19: 10929216-10929216 on Build GRCh38

View Full Product Details