Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11451436-11451937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL3
Entrez Gene ID
1995
Gene Name
ELAV like neuron-specific RNA binding protein 3
Gene Aliases
HUC, HUCL, PLE21
UniGene
Hs.1701
Gene Chromosome Location
Chr.19: 11451328-11480994 on Build GRCh38
Transcripts
NM_001420.3 NM_032281.2

SNP Detail

SNP ID Chromosome Location
rs1020824895 Chr.19: 11451494-11451494 on Build GRCh38
rs983504717 Chr.19: 11451691-11451691 on Build GRCh38
rs370492835 Chr.19: 11451535-11451535 on Build GRCh38
rs886483034 Chr.19: 11451822-11451822 on Build GRCh38
rs955841875 Chr.19: 11451884-11451884 on Build GRCh38
rs1016333621 Chr.19: 11451844-11451845 on Build GRCh38
rs878940568 Chr.19: 11451482-11451482 on Build GRCh38
rs1004440541 Chr.19: 11451806-11451806 on Build GRCh38
rs988987770 Chr.19: 11451888-11451888 on Build GRCh38
rs950770452 Chr.19: 11451668-11451668 on Build GRCh38
rs995717027 Chr.19: 11451846-11451846 on Build GRCh38
rs571280193 Chr.19: 11451802-11451802 on Build GRCh38
rs900468322 Chr.19: 11451764-11451764 on Build GRCh38
rs985504633 Chr.19: 11451824-11451824 on Build GRCh38
rs369400019 Chr.19: 11451480-11451492 on Build GRCh38
rs934395124 Chr.19: 11451802-11451802 on Build GRCh38
rs893296052 Chr.19: 11451804-11451805 on Build GRCh38
rs77049617 Chr.19: 11451492-11451492 on Build GRCh38
rs561905305 Chr.19: 11451860-11451860 on Build GRCh38
rs973996355 Chr.19: 11451722-11451722 on Build GRCh38
rs375969105 Chr.19: 11451480-11451494 on Build GRCh38
rs915574311 Chr.19: 11451589-11451589 on Build GRCh38
rs928286808 Chr.19: 11451800-11451801 on Build GRCh38
rs200985797 Chr.19: 11451461-11451461 on Build GRCh38
rs906921133 Chr.19: 11451609-11451609 on Build GRCh38
rs934403693 Chr.19: 11451614-11451614 on Build GRCh38
rs994839922 Chr.19: 11451773-11451773 on Build GRCh38
rs974438451 Chr.19: 11451588-11451588 on Build GRCh38
rs1030303056 Chr.19: 11451667-11451667 on Build GRCh38
rs554385350 Chr.19: 11451629-11451629 on Build GRCh38
rs963510068 Chr.19: 11451845-11451845 on Build GRCh38
rs969694396 Chr.19: 11451855-11451855 on Build GRCh38
rs981069496 Chr.19: 11451730-11451730 on Build GRCh38
rs925681450 Chr.19: 11451862-11451862 on Build GRCh38
rs965107216 Chr.19: 11451479-11451479 on Build GRCh38
rs908393059 Chr.19: 11451543-11451544 on Build GRCh38
rs1030397789 Chr.19: 11451493-11451493 on Build GRCh38
rs182533486 Chr.19: 11451660-11451660 on Build GRCh38
rs35932742 Chr.19: 11451867-11451868 on Build GRCh38
rs138354612 Chr.19: 11451644-11451644 on Build GRCh38
rs544926982 Chr.19: 11451810-11451810 on Build GRCh38
rs543370794 Chr.19: 11451798-11451798 on Build GRCh38
rs886557117 Chr.19: 11451804-11451804 on Build GRCh38
rs564976823 Chr.19: 11451803-11451803 on Build GRCh38
rs948676051 Chr.19: 11451728-11451728 on Build GRCh38
rs34915442 Chr.19: 11451825-11451826 on Build GRCh38
rs1017601175 Chr.19: 11451478-11451478 on Build GRCh38
rs1015380641 Chr.19: 11451695-11451695 on Build GRCh38
rs893346976 Chr.19: 11451634-11451634 on Build GRCh38
rs1045417648 Chr.19: 11451595-11451595 on Build GRCh38
rs1039222634 Chr.19: 11451808-11451809 on Build GRCh38
rs950823031 Chr.19: 11451494-11451495 on Build GRCh38
rs140160574 Chr.19: 11451801-11451802 on Build GRCh38
rs1022840365 Chr.19: 11451851-11451851 on Build GRCh38
rs948411051 Chr.19: 11451590-11451590 on Build GRCh38
rs560277500 Chr.19: 11451841-11451841 on Build GRCh38
rs576483144 Chr.19: 11451694-11451694 on Build GRCh38
rs542073024 Chr.19: 11451809-11451809 on Build GRCh38
rs947565642 Chr.19: 11451805-11451805 on Build GRCh38
rs1012244112 Chr.19: 11451467-11451467 on Build GRCh38
rs944954532 Chr.19: 11451651-11451651 on Build GRCh38
rs1053321517 Chr.19: 11451624-11451624 on Build GRCh38
rs535690633 Chr.19: 11451889-11451889 on Build GRCh38
rs539389362 Chr.19: 11451617-11451617 on Build GRCh38
rs944862436 Chr.19: 11451893-11451893 on Build GRCh38
rs373766267 Chr.19: 11451567-11451567 on Build GRCh38
rs981395763 Chr.19: 11451885-11451885 on Build GRCh38
rs578247953 Chr.19: 11451613-11451613 on Build GRCh38
rs1027735326 Chr.19: 11451799-11451799 on Build GRCh38
rs373036949 Chr.19: 11451487-11451487 on Build GRCh38
rs982551533 Chr.19: 11451495-11451498 on Build GRCh38
rs976179986 Chr.19: 11451488-11451489 on Build GRCh38
rs879016342 Chr.19: 11451480-11451480 on Build GRCh38
rs962899409 Chr.19: 11451709-11451709 on Build GRCh38
rs574176645 Chr.19: 11451604-11451604 on Build GRCh38
rs567379211 Chr.19: 11451533-11451533 on Build GRCh38
rs770710524 Chr.19: 11451464-11451464 on Build GRCh38
rs998183032 Chr.19: 11451792-11451792 on Build GRCh38
rs1039262185 Chr.19: 11451911-11451911 on Build GRCh38
rs992674649 Chr.19: 11451835-11451835 on Build GRCh38
rs997787118 Chr.19: 11451809-11451810 on Build GRCh38
rs922217145 Chr.19: 11451915-11451915 on Build GRCh38
rs954091441 Chr.19: 11451812-11451812 on Build GRCh38

View Full Product Details