Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11451947-11452466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL3
Entrez Gene ID
1995
Gene Name
ELAV like neuron-specific RNA binding protein 3
Gene Aliases
HUC, HUCL, PLE21
UniGene
Hs.1701
Gene Chromosome Location
Chr.19: 11451328-11480994 on Build GRCh38
Transcripts
NM_001420.3 NM_032281.2

SNP Detail

SNP ID Chromosome Location
rs1014030325 Chr.19: 11452222-11452222 on Build GRCh38
rs539323398 Chr.19: 11451971-11451971 on Build GRCh38
rs775751854 Chr.19: 11452178-11452178 on Build GRCh38
rs955692587 Chr.19: 11452310-11452310 on Build GRCh38
rs576859480 Chr.19: 11452291-11452291 on Build GRCh38
rs968834890 Chr.19: 11452270-11452270 on Build GRCh38
rs555892783 Chr.19: 11452242-11452242 on Build GRCh38
rs940953083 Chr.19: 11452333-11452333 on Build GRCh38
rs747422325 Chr.19: 11452104-11452108 on Build GRCh38
rs974939774 Chr.19: 11452177-11452177 on Build GRCh38
rs933522048 Chr.19: 11452292-11452292 on Build GRCh38
rs150329191 Chr.19: 11452110-11452111 on Build GRCh38
rs911631769 Chr.19: 11452152-11452152 on Build GRCh38
rs948126353 Chr.19: 11452410-11452410 on Build GRCh38
rs76880598 Chr.19: 11452181-11452181 on Build GRCh38
rs554136591 Chr.19: 11451984-11451984 on Build GRCh38
rs746283996 Chr.19: 11452103-11452103 on Build GRCh38
rs191775580 Chr.19: 11452309-11452309 on Build GRCh38
rs758494814 Chr.19: 11452174-11452174 on Build GRCh38
rs969977520 Chr.19: 11452035-11452035 on Build GRCh38
rs769976972 Chr.19: 11452166-11452166 on Build GRCh38
rs560142919 Chr.19: 11452314-11452314 on Build GRCh38
rs1012334560 Chr.19: 11452146-11452146 on Build GRCh38
rs952202381 Chr.19: 11452404-11452404 on Build GRCh38
rs988447801 Chr.19: 11452220-11452220 on Build GRCh38
rs543554643 Chr.19: 11452183-11452183 on Build GRCh38
rs1056551188 Chr.19: 11452201-11452201 on Build GRCh38
rs974889143 Chr.19: 11452357-11452357 on Build GRCh38
rs1000091095 Chr.19: 11452297-11452297 on Build GRCh38
rs1049796056 Chr.19: 11452182-11452182 on Build GRCh38
rs766572253 Chr.19: 11452243-11452243 on Build GRCh38
rs940694021 Chr.19: 11452185-11452185 on Build GRCh38
rs1032658459 Chr.19: 11452050-11452050 on Build GRCh38
rs749352649 Chr.19: 11452390-11452390 on Build GRCh38
rs149256658 Chr.19: 11452032-11452032 on Build GRCh38
rs143597166 Chr.19: 11452358-11452358 on Build GRCh38
rs905725343 Chr.19: 11452281-11452281 on Build GRCh38
rs757604112 Chr.19: 11452015-11452015 on Build GRCh38
rs768262774 Chr.19: 11452105-11452106 on Build GRCh38
rs1035631008 Chr.19: 11452213-11452215 on Build GRCh38
rs899202548 Chr.19: 11452385-11452385 on Build GRCh38
rs999802453 Chr.19: 11452048-11452048 on Build GRCh38
rs186495240 Chr.19: 11452257-11452257 on Build GRCh38
rs896610302 Chr.19: 11452208-11452208 on Build GRCh38
rs1004813429 Chr.19: 11452049-11452049 on Build GRCh38
rs781435684 Chr.19: 11452033-11452033 on Build GRCh38
rs1010355467 Chr.19: 11452311-11452311 on Build GRCh38
rs536397658 Chr.19: 11452009-11452009 on Build GRCh38
rs751780406 Chr.19: 11452291-11452294 on Build GRCh38
rs988546776 Chr.19: 11452141-11452141 on Build GRCh38
rs183587442 Chr.19: 11452332-11452332 on Build GRCh38
rs955912470 Chr.19: 11452099-11452099 on Build GRCh38
rs566383889 Chr.19: 11452003-11452003 on Build GRCh38
rs965811550 Chr.19: 11452334-11452334 on Build GRCh38
rs1052002561 Chr.19: 11452016-11452016 on Build GRCh38
rs1051949973 Chr.19: 11452299-11452300 on Build GRCh38
rs955959635 Chr.19: 11452218-11452220 on Build GRCh38
rs530582137 Chr.19: 11451985-11451985 on Build GRCh38
rs1029142501 Chr.19: 11452378-11452378 on Build GRCh38

View Full Product Details