Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11452746-11453248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL3
Entrez Gene ID
1995
Gene Name
ELAV like neuron-specific RNA binding protein 3
Gene Aliases
HUC, HUCL, PLE21
UniGene
Hs.1701
Gene Chromosome Location
Chr.19: 11451328-11480994 on Build GRCh38
Transcripts
NM_001420.3 NM_032281.2

SNP Detail

SNP ID Chromosome Location
rs969360235 Chr.19: 11453186-11453186 on Build GRCh38
rs1043292170 Chr.19: 11453072-11453072 on Build GRCh38
rs532116460 Chr.19: 11452795-11452795 on Build GRCh38
rs913027058 Chr.19: 11453197-11453197 on Build GRCh38
rs945930244 Chr.19: 11453042-11453042 on Build GRCh38
rs962632923 Chr.19: 11452918-11452918 on Build GRCh38
rs776544004 Chr.19: 11452902-11452902 on Build GRCh38
rs961783859 Chr.19: 11453171-11453171 on Build GRCh38
rs779987842 Chr.19: 11452835-11452835 on Build GRCh38
rs948323081 Chr.19: 11453007-11453007 on Build GRCh38
rs958385282 Chr.19: 11453195-11453195 on Build GRCh38
rs1008489493 Chr.19: 11453224-11453224 on Build GRCh38
rs537130912 Chr.19: 11452998-11452998 on Build GRCh38
rs78076958 Chr.19: 11452835-11452835 on Build GRCh38
rs763546092 Chr.19: 11453193-11453193 on Build GRCh38
rs923222657 Chr.19: 11453219-11453219 on Build GRCh38
rs554859375 Chr.19: 11452881-11452881 on Build GRCh38
rs536992960 Chr.19: 11452869-11452869 on Build GRCh38
rs908060232 Chr.19: 11452768-11452769 on Build GRCh38
rs757915974 Chr.19: 11453120-11453120 on Build GRCh38
rs1054373935 Chr.19: 11452770-11452770 on Build GRCh38
rs1013759149 Chr.19: 11453146-11453146 on Build GRCh38
rs549937322 Chr.19: 11452789-11452789 on Build GRCh38
rs192366977 Chr.19: 11453065-11453065 on Build GRCh38
rs948065732 Chr.19: 11452947-11452947 on Build GRCh38
rs558590483 Chr.19: 11453054-11453054 on Build GRCh38
rs553375976 Chr.19: 11453136-11453136 on Build GRCh38
rs992442188 Chr.19: 11452926-11452926 on Build GRCh38
rs1056951048 Chr.19: 11452853-11452853 on Build GRCh38
rs906591924 Chr.19: 11452846-11452846 on Build GRCh38
rs952150907 Chr.19: 11452903-11452903 on Build GRCh38
rs534660764 Chr.19: 11453088-11453088 on Build GRCh38
rs948108690 Chr.19: 11452834-11452835 on Build GRCh38
rs543455540 Chr.19: 11453190-11453190 on Build GRCh38
rs1025186018 Chr.19: 11453159-11453159 on Build GRCh38
rs1054723278 Chr.19: 11452986-11452986 on Build GRCh38
rs542936485 Chr.19: 11453198-11453198 on Build GRCh38
rs980757475 Chr.19: 11453013-11453013 on Build GRCh38
rs865879081 Chr.19: 11453066-11453066 on Build GRCh38
rs752413167 Chr.19: 11453112-11453112 on Build GRCh38
rs891691898 Chr.19: 11453027-11453027 on Build GRCh38
rs1015484355 Chr.19: 11453135-11453135 on Build GRCh38
rs569940739 Chr.19: 11452956-11452956 on Build GRCh38
rs757125510 Chr.19: 11452847-11452847 on Build GRCh38
rs1009889458 Chr.19: 11452778-11452778 on Build GRCh38
rs754879241 Chr.19: 11453193-11453193 on Build GRCh38
rs778759174 Chr.19: 11452896-11452896 on Build GRCh38
rs759470826 Chr.19: 11452980-11452980 on Build GRCh38
rs1040275631 Chr.19: 11453202-11453202 on Build GRCh38
rs1050043884 Chr.19: 11453102-11453102 on Build GRCh38
rs188926056 Chr.19: 11452818-11452818 on Build GRCh38
rs1006372070 Chr.19: 11453093-11453093 on Build GRCh38
rs985399118 Chr.19: 11452908-11452908 on Build GRCh38
rs771524803 Chr.19: 11452985-11452985 on Build GRCh38
rs764970946 Chr.19: 11453085-11453085 on Build GRCh38
rs981464148 Chr.19: 11453194-11453194 on Build GRCh38
rs1040327425 Chr.19: 11453044-11453044 on Build GRCh38
rs931624197 Chr.19: 11453225-11453225 on Build GRCh38
rs868608846 Chr.19: 11453051-11453051 on Build GRCh38
rs371526572 Chr.19: 11452864-11452864 on Build GRCh38
rs917890117 Chr.19: 11452946-11452946 on Build GRCh38
rs891740187 Chr.19: 11452774-11452774 on Build GRCh38
rs544515244 Chr.19: 11453188-11453188 on Build GRCh38
rs1015164965 Chr.19: 11452909-11452909 on Build GRCh38
rs374445012 Chr.19: 11452791-11452791 on Build GRCh38
rs1010153244 Chr.19: 11452865-11452865 on Build GRCh38
rs890160115 Chr.19: 11453104-11453104 on Build GRCh38
rs138155859 Chr.19: 11452793-11452793 on Build GRCh38
rs980786360 Chr.19: 11452948-11452948 on Build GRCh38
rs1028240693 Chr.19: 11453193-11453194 on Build GRCh38
rs925412832 Chr.19: 11452950-11452950 on Build GRCh38
rs904172492 Chr.19: 11452895-11452895 on Build GRCh38
rs114348792 Chr.19: 11453196-11453196 on Build GRCh38

View Full Product Details