Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11454261-11454769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL3
Entrez Gene ID
1995
Gene Name
ELAV like neuron-specific RNA binding protein 3
Gene Aliases
HUC, HUCL, PLE21
UniGene
Hs.1701
Gene Chromosome Location
Chr.19: 11451328-11480994 on Build GRCh38
Transcripts
NM_001420.3 NM_032281.2 XM_011527778.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1390366 c.988G>A p.V330M Chr.19: 11454642-11454642 on Build GRCh38
7252077 c.1026C>A p.I342I Chr.19: 11454604-11454604 on Build GRCh38
6149672 c.909G>T p.L303L Chr.19: 11454721-11454721 on Build GRCh38
1390367 c.945C>A p.V315V Chr.19: 11454685-11454685 on Build GRCh38
3734547 c.1037A>G p.N346S Chr.19: 11454593-11454593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049783848 Chr.19: 11454703-11454703 on Build GRCh38
rs966855065 Chr.19: 11454450-11454451 on Build GRCh38
rs989649393 Chr.19: 11454592-11454592 on Build GRCh38
rs757745976 Chr.19: 11454567-11454567 on Build GRCh38
rs1016073271 Chr.19: 11454437-11454437 on Build GRCh38
rs776585368 Chr.19: 11454599-11454599 on Build GRCh38
rs926361415 Chr.19: 11454577-11454577 on Build GRCh38
rs553002178 Chr.19: 11454746-11454746 on Build GRCh38
rs145408161 Chr.19: 11454576-11454576 on Build GRCh38
rs754575890 Chr.19: 11454627-11454627 on Build GRCh38
rs747056035 Chr.19: 11454730-11454730 on Build GRCh38
rs747153913 Chr.19: 11454583-11454583 on Build GRCh38
rs936882896 Chr.19: 11454452-11454469 on Build GRCh38
rs765748795 Chr.19: 11454622-11454622 on Build GRCh38
rs768572703 Chr.19: 11454595-11454595 on Build GRCh38
rs748089092 Chr.19: 11454472-11454473 on Build GRCh38
rs764677546 Chr.19: 11454637-11454637 on Build GRCh38
rs925424662 Chr.19: 11454450-11454457 on Build GRCh38
rs191710732 Chr.19: 11454571-11454571 on Build GRCh38
rs761571357 Chr.19: 11454603-11454603 on Build GRCh38
rs758637215 Chr.19: 11454698-11454698 on Build GRCh38
rs1024655825 Chr.19: 11454449-11454449 on Build GRCh38
rs145003649 Chr.19: 11454556-11454556 on Build GRCh38
rs889769517 Chr.19: 11454317-11454317 on Build GRCh38
rs1007789478 Chr.19: 11454416-11454416 on Build GRCh38
rs139974609 Chr.19: 11454529-11454529 on Build GRCh38
rs1050033568 Chr.19: 11454314-11454314 on Build GRCh38
rs547037629 Chr.19: 11454412-11454412 on Build GRCh38
rs749641214 Chr.19: 11454547-11454547 on Build GRCh38
rs199635144 Chr.19: 11454742-11454742 on Build GRCh38
rs753076296 Chr.19: 11454527-11454527 on Build GRCh38
rs778955437 Chr.19: 11454570-11454570 on Build GRCh38
rs750136144 Chr.19: 11454517-11454517 on Build GRCh38
rs552353483 Chr.19: 11454328-11454328 on Build GRCh38
rs1049833660 Chr.19: 11454408-11454408 on Build GRCh38
rs778492575 Chr.19: 11454530-11454530 on Build GRCh38
rs199860514 Chr.19: 11454604-11454604 on Build GRCh38
rs945149656 Chr.19: 11454288-11454288 on Build GRCh38
rs148710112 Chr.19: 11454643-11454643 on Build GRCh38
rs911450726 Chr.19: 11454469-11454470 on Build GRCh38
rs142289115 Chr.19: 11454694-11454694 on Build GRCh38
rs766524991 Chr.19: 11454302-11454302 on Build GRCh38
rs768770817 Chr.19: 11454736-11454736 on Build GRCh38
rs779645608 Chr.19: 11454523-11454523 on Build GRCh38
rs994268769 Chr.19: 11454368-11454368 on Build GRCh38
rs760668510 Chr.19: 11454499-11454499 on Build GRCh38
rs757797512 Chr.19: 11454521-11454521 on Build GRCh38
rs963554329 Chr.19: 11454467-11454467 on Build GRCh38
rs369778910 Chr.19: 11454399-11454399 on Build GRCh38
rs35334288 Chr.19: 11454470-11454471 on Build GRCh38
rs764834247 Chr.19: 11454509-11454509 on Build GRCh38
rs977908007 Chr.19: 11454450-11454453 on Build GRCh38
rs767566028 Chr.19: 11454470-11454473 on Build GRCh38
rs568484090 Chr.19: 11454493-11454493 on Build GRCh38
rs925508709 Chr.19: 11454298-11454298 on Build GRCh38
rs1008154193 Chr.19: 11454321-11454321 on Build GRCh38
rs558761025 Chr.19: 11454649-11454649 on Build GRCh38
rs200375391 Chr.19: 11454338-11454338 on Build GRCh38
rs878986385 Chr.19: 11454748-11454748 on Build GRCh38
rs991785064 Chr.19: 11454435-11454435 on Build GRCh38
rs926413565 Chr.19: 11454296-11454296 on Build GRCh38
rs775255481 Chr.19: 11454497-11454497 on Build GRCh38
rs1013542667 Chr.19: 11454737-11454737 on Build GRCh38
rs901135573 Chr.19: 11454396-11454396 on Build GRCh38
rs112277943 Chr.19: 11454309-11454309 on Build GRCh38
rs113632486 Chr.19: 11454292-11454292 on Build GRCh38
rs746022996 Chr.19: 11454697-11454697 on Build GRCh38
rs753613748 Chr.19: 11454501-11454501 on Build GRCh38
rs140647234 Chr.19: 11454744-11454744 on Build GRCh38
rs1043041667 Chr.19: 11454378-11454378 on Build GRCh38
rs200383181 Chr.19: 11454500-11454500 on Build GRCh38
rs761661246 Chr.19: 11454502-11454502 on Build GRCh38
rs985605661 Chr.19: 11454468-11454469 on Build GRCh38
rs780204044 Chr.19: 11454718-11454718 on Build GRCh38
rs147698039 Chr.19: 11454580-11454580 on Build GRCh38
rs1053984953 Chr.19: 11454316-11454316 on Build GRCh38
rs201043909 Chr.19: 11454616-11454616 on Build GRCh38

View Full Product Details