Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 33380619-33381123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEBPG
Entrez Gene ID
1054
Gene Name
CCAAT/enhancer binding protein gamma
Gene Aliases
GPE1BP, IG/EBP-1
UniGene
Hs.429666
Gene Chromosome Location
Chr.19: 33373669-33382686 on Build GRCh38
Transcripts
NM_001252296.1 NM_001806.3

SNP Detail

SNP ID Chromosome Location
rs775676947 Chr.19: 33380818-33380818 on Build GRCh38
rs1037068024 Chr.19: 33381088-33381091 on Build GRCh38
rs764451283 Chr.19: 33380999-33381000 on Build GRCh38
rs141142953 Chr.19: 33381075-33381079 on Build GRCh38
rs928889791 Chr.19: 33380914-33380914 on Build GRCh38
rs999768546 Chr.19: 33380732-33380732 on Build GRCh38
rs961546591 Chr.19: 33380748-33380748 on Build GRCh38
rs959295349 Chr.19: 33380826-33380828 on Build GRCh38
rs1037766158 Chr.19: 33381052-33381052 on Build GRCh38
rs890770015 Chr.19: 33380693-33380693 on Build GRCh38
rs931988957 Chr.19: 33381068-33381068 on Build GRCh38
rs995356394 Chr.19: 33380811-33380811 on Build GRCh38
rs957807185 Chr.19: 33380751-33380751 on Build GRCh38
rs986201562 Chr.19: 33381075-33381093 on Build GRCh38
rs751076475 Chr.19: 33381063-33381066 on Build GRCh38
rs984066491 Chr.19: 33381008-33381008 on Build GRCh38
rs775810212 Chr.19: 33380774-33380774 on Build GRCh38
rs1027202013 Chr.19: 33380816-33380816 on Build GRCh38
rs189373610 Chr.19: 33380825-33380825 on Build GRCh38
rs763705880 Chr.19: 33381000-33381000 on Build GRCh38
rs565745321 Chr.19: 33380988-33380988 on Build GRCh38
rs116563073 Chr.19: 33381036-33381036 on Build GRCh38
rs34825094 Chr.19: 33380810-33380811 on Build GRCh38
rs764533636 Chr.19: 33380908-33380908 on Build GRCh38
rs938826804 Chr.19: 33380959-33380959 on Build GRCh38
rs916273848 Chr.19: 33380821-33380821 on Build GRCh38
rs1030747627 Chr.19: 33380746-33380746 on Build GRCh38
rs529367654 Chr.19: 33380675-33380675 on Build GRCh38
rs954533199 Chr.19: 33380684-33380684 on Build GRCh38
rs575842021 Chr.19: 33380668-33380670 on Build GRCh38
rs942150340 Chr.19: 33381039-33381039 on Build GRCh38
rs377189716 Chr.19: 33380700-33380700 on Build GRCh38
rs1051646197 Chr.19: 33381092-33381092 on Build GRCh38
rs375846958 Chr.19: 33381082-33381086 on Build GRCh38
rs77928542 Chr.19: 33381096-33381096 on Build GRCh38
rs917940900 Chr.19: 33380887-33380887 on Build GRCh38
rs989133626 Chr.19: 33380782-33380782 on Build GRCh38
rs528711140 Chr.19: 33380989-33380989 on Build GRCh38
rs77871339 Chr.19: 33381060-33381060 on Build GRCh38
rs980351788 Chr.19: 33380882-33380882 on Build GRCh38
rs990674472 Chr.19: 33380839-33380839 on Build GRCh38
rs75664808 Chr.19: 33381054-33381054 on Build GRCh38

View Full Product Details