Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 33771335-33771844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST8
Entrez Gene ID
64377
Gene Name
carbohydrate sulfotransferase 8
Gene Aliases
GALNAC4ST1, GalNAc4ST, PSS3
UniGene
Hs.165724
Gene Chromosome Location
Chr.19: 33621955-33773509 on Build GRCh38
Transcripts
NM_001127895.1 NM_001127896.1 NM_022467.3 XM_011527222.1 XM_011527224.1 XM_017027143.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6150569 c.150G>T p.R50S Chr.19: 33771432-33771432 on Build GRCh38
6629030 c.149G>T p.R50M Chr.19: 33771431-33771431 on Build GRCh38
6629028 c.131G>A p.G44E Chr.19: 33771413-33771413 on Build GRCh38
3981672 c.168+1G>T p.? Chr.19: 33771451-33771451 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775070377 Chr.19: 33771379-33771379 on Build GRCh38
rs900203461 Chr.19: 33771814-33771814 on Build GRCh38
rs756838397 Chr.19: 33771420-33771420 on Build GRCh38
rs8112884 Chr.19: 33771544-33771544 on Build GRCh38
rs995924717 Chr.19: 33771819-33771819 on Build GRCh38
rs896357783 Chr.19: 33771535-33771535 on Build GRCh38
rs911626871 Chr.19: 33771718-33771718 on Build GRCh38
rs900255818 Chr.19: 33771612-33771612 on Build GRCh38
rs985782613 Chr.19: 33771702-33771702 on Build GRCh38
rs1004787819 Chr.19: 33771566-33771566 on Build GRCh38
rs911672852 Chr.19: 33771448-33771448 on Build GRCh38
rs763630529 Chr.19: 33771486-33771486 on Build GRCh38
rs1032072270 Chr.19: 33771579-33771579 on Build GRCh38
rs866170073 Chr.19: 33771752-33771752 on Build GRCh38
rs988305129 Chr.19: 33771737-33771737 on Build GRCh38
rs905141215 Chr.19: 33771799-33771799 on Build GRCh38
rs765524936 Chr.19: 33771483-33771483 on Build GRCh38
rs59181704 Chr.19: 33771695-33771695 on Build GRCh38
rs1054885751 Chr.19: 33771529-33771529 on Build GRCh38
rs542211031 Chr.19: 33771740-33771740 on Build GRCh38
rs373932408 Chr.19: 33771450-33771450 on Build GRCh38
rs770347420 Chr.19: 33771478-33771478 on Build GRCh38
rs920222183 Chr.19: 33771413-33771413 on Build GRCh38
rs543532748 Chr.19: 33771682-33771682 on Build GRCh38
rs974270359 Chr.19: 33771684-33771684 on Build GRCh38
rs150528629 Chr.19: 33771467-33771467 on Build GRCh38
rs577801599 Chr.19: 33771680-33771680 on Build GRCh38
rs747859086 Chr.19: 33771372-33771372 on Build GRCh38
rs572359168 Chr.19: 33771494-33771494 on Build GRCh38
rs913696423 Chr.19: 33771774-33771774 on Build GRCh38
rs1052315703 Chr.19: 33771455-33771455 on Build GRCh38
rs923865770 Chr.19: 33771776-33771776 on Build GRCh38
rs1035502301 Chr.19: 33771543-33771543 on Build GRCh38
rs551683292 Chr.19: 33771786-33771786 on Build GRCh38
rs750997956 Chr.19: 33771409-33771409 on Build GRCh38
rs564772614 Chr.19: 33771651-33771651 on Build GRCh38
rs373322550 Chr.19: 33771459-33771459 on Build GRCh38
rs1015726345 Chr.19: 33771613-33771613 on Build GRCh38
rs376789824 Chr.19: 33771451-33771451 on Build GRCh38
rs369757128 Chr.19: 33771408-33771408 on Build GRCh38
rs1042737586 Chr.19: 33771557-33771557 on Build GRCh38
rs949265446 Chr.19: 33771768-33771768 on Build GRCh38
rs189355962 Chr.19: 33771378-33771378 on Build GRCh38
rs767511543 Chr.19: 33771422-33771422 on Build GRCh38
rs944507816 Chr.19: 33771736-33771736 on Build GRCh38
rs983000462 Chr.19: 33771363-33771363 on Build GRCh38
rs964860118 Chr.19: 33771389-33771389 on Build GRCh38
rs375546489 Chr.19: 33771383-33771383 on Build GRCh38
rs1030492128 Chr.19: 33771616-33771616 on Build GRCh38
rs371107498 Chr.19: 33771471-33771471 on Build GRCh38
rs776443865 Chr.19: 33771673-33771673 on Build GRCh38
rs565103445 Chr.19: 33771815-33771815 on Build GRCh38
rs760543482 Chr.19: 33771569-33771569 on Build GRCh38
rs917756179 Chr.19: 33771766-33771766 on Build GRCh38
rs774072457 Chr.19: 33771489-33771489 on Build GRCh38
rs559386868 Chr.19: 33771753-33771753 on Build GRCh38
rs3810362 Chr.19: 33771484-33771484 on Build GRCh38
rs902158189 Chr.19: 33771565-33771565 on Build GRCh38
rs905172824 Chr.19: 33771508-33771508 on Build GRCh38
rs528434904 Chr.19: 33771779-33771779 on Build GRCh38
rs902211131 Chr.19: 33771366-33771366 on Build GRCh38
rs60463284 Chr.19: 33771514-33771514 on Build GRCh38
rs963204490 Chr.19: 33771683-33771683 on Build GRCh38
rs974303156 Chr.19: 33771388-33771388 on Build GRCh38
rs771847472 Chr.19: 33771437-33771437 on Build GRCh38
rs755905971 Chr.19: 33771447-33771447 on Build GRCh38
rs746392212 Chr.19: 33771476-33771476 on Build GRCh38
rs775825098 Chr.19: 33771480-33771480 on Build GRCh38
rs1043638055 Chr.19: 33771795-33771795 on Build GRCh38
rs955087434 Chr.19: 33771659-33771659 on Build GRCh38
rs369106366 Chr.19: 33771456-33771456 on Build GRCh38
rs866764582 Chr.19: 33771614-33771614 on Build GRCh38
rs767137491 Chr.19: 33771495-33771495 on Build GRCh38
rs921532155 Chr.19: 33771688-33771688 on Build GRCh38
rs573590809 Chr.19: 33771722-33771722 on Build GRCh38
rs1042293320 Chr.19: 33771775-33771775 on Build GRCh38
rs759856653 Chr.19: 33771367-33771367 on Build GRCh38

View Full Product Details