Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35143115-35143632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD7
Entrez Gene ID
53822
Gene Name
FXYD domain containing ion transport regulator 7
Gene Aliases
-
UniGene
Hs.134729
Gene Chromosome Location
Chr.19: 35143250-35154302 on Build GRCh38
Transcripts
NM_022006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
395713 c.23C>A p.P8H Chr.19: 35143356-35143356 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886666139 Chr.19: 35143513-35143513 on Build GRCh38
rs988522784 Chr.19: 35143594-35143594 on Build GRCh38
rs955805815 Chr.19: 35143248-35143248 on Build GRCh38
rs928288059 Chr.19: 35143383-35143383 on Build GRCh38
rs143814512 Chr.19: 35143596-35143597 on Build GRCh38
rs943407628 Chr.19: 35143459-35143459 on Build GRCh38
rs570830909 Chr.19: 35143517-35143517 on Build GRCh38
rs1008119381 Chr.19: 35143554-35143554 on Build GRCh38
rs995441283 Chr.19: 35143149-35143149 on Build GRCh38
rs980252344 Chr.19: 35143556-35143556 on Build GRCh38
rs953936366 Chr.19: 35143197-35143197 on Build GRCh38
rs914246097 Chr.19: 35143251-35143251 on Build GRCh38
rs766333956 Chr.19: 35143405-35143405 on Build GRCh38
rs913093442 Chr.19: 35143392-35143392 on Build GRCh38
rs548491599 Chr.19: 35143139-35143139 on Build GRCh38
rs550240507 Chr.19: 35143541-35143541 on Build GRCh38
rs997749845 Chr.19: 35143492-35143492 on Build GRCh38
rs1042882801 Chr.19: 35143435-35143435 on Build GRCh38
rs899205396 Chr.19: 35143519-35143519 on Build GRCh38
rs1021325555 Chr.19: 35143598-35143598 on Build GRCh38
rs764410423 Chr.19: 35143367-35143367 on Build GRCh38
rs780677484 Chr.19: 35143394-35143394 on Build GRCh38
rs966320619 Chr.19: 35143246-35143247 on Build GRCh38
rs1019343925 Chr.19: 35143219-35143219 on Build GRCh38
rs71352908 Chr.19: 35143416-35143416 on Build GRCh38
rs1006354791 Chr.19: 35143523-35143523 on Build GRCh38
rs10424787 Chr.19: 35143294-35143294 on Build GRCh38
rs899133682 Chr.19: 35143147-35143147 on Build GRCh38
rs903881705 Chr.19: 35143174-35143174 on Build GRCh38
rs980158950 Chr.19: 35143603-35143603 on Build GRCh38
rs996574390 Chr.19: 35143521-35143521 on Build GRCh38
rs1022276946 Chr.19: 35143152-35143152 on Build GRCh38
rs926957065 Chr.19: 35143611-35143611 on Build GRCh38
rs779328534 Chr.19: 35143599-35143599 on Build GRCh38
rs756962530 Chr.19: 35143386-35143386 on Build GRCh38
rs1019417013 Chr.19: 35143557-35143557 on Build GRCh38
rs980418815 Chr.19: 35143286-35143286 on Build GRCh38
rs903912755 Chr.19: 35143192-35143192 on Build GRCh38
rs545450660 Chr.19: 35143513-35143514 on Build GRCh38
rs547926077 Chr.19: 35143465-35143465 on Build GRCh38
rs886578704 Chr.19: 35143441-35143441 on Build GRCh38
rs374098245 Chr.19: 35143525-35143525 on Build GRCh38
rs1687987 Chr.19: 35143494-35143494 on Build GRCh38
rs979796360 Chr.19: 35143326-35143326 on Build GRCh38
rs562309524 Chr.19: 35143253-35143253 on Build GRCh38
rs970180166 Chr.19: 35143322-35143322 on Build GRCh38
rs901831533 Chr.19: 35143468-35143468 on Build GRCh38
rs751191575 Chr.19: 35143379-35143379 on Build GRCh38
rs1043813930 Chr.19: 35143158-35143158 on Build GRCh38
rs569099505 Chr.19: 35143599-35143599 on Build GRCh38
rs903944298 Chr.19: 35143444-35143444 on Build GRCh38
rs938647635 Chr.19: 35143385-35143385 on Build GRCh38
rs987024267 Chr.19: 35143201-35143201 on Build GRCh38
rs891626777 Chr.19: 35143140-35143140 on Build GRCh38
rs753169995 Chr.19: 35143287-35143287 on Build GRCh38
rs997718886 Chr.19: 35143247-35143247 on Build GRCh38
rs1037753151 Chr.19: 35143488-35143488 on Build GRCh38
rs1028657862 Chr.19: 35143177-35143177 on Build GRCh38
rs988491551 Chr.19: 35143250-35143250 on Build GRCh38
rs947221204 Chr.19: 35143414-35143414 on Build GRCh38
rs867467191 Chr.19: 35143577-35143577 on Build GRCh38
rs867354562 Chr.19: 35143172-35143172 on Build GRCh38
rs375297127 Chr.19: 35143295-35143295 on Build GRCh38
rs1057339116 Chr.19: 35143439-35143439 on Build GRCh38
rs955838059 Chr.19: 35143564-35143564 on Build GRCh38
rs796941718 Chr.19: 35143419-35143420 on Build GRCh38
rs74376951 Chr.19: 35143514-35143514 on Build GRCh38
rs1009683993 Chr.19: 35143153-35143158 on Build GRCh38

View Full Product Details