Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35360384-35360932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FFAR3
Entrez Gene ID
2865
Gene Name
free fatty acid receptor 3
Gene Aliases
FFA3R, GPR41, GPR42
UniGene
Hs.248055
Gene Chromosome Location
Chr.19: 35358106-35360491 on Build GRCh38
Transcripts
NM_005304.3 XM_011526858.2

SNP Detail

SNP ID Chromosome Location
rs566861879 Chr.19: 35360660-35360660 on Build GRCh38
rs1001130759 Chr.19: 35360877-35360877 on Build GRCh38
rs908702016 Chr.19: 35360471-35360471 on Build GRCh38
rs75269464 Chr.19: 35360717-35360717 on Build GRCh38
rs10407548 Chr.19: 35360713-35360713 on Build GRCh38
rs554823291 Chr.19: 35360875-35360875 on Build GRCh38
rs578188621 Chr.19: 35360542-35360542 on Build GRCh38
rs546737308 Chr.19: 35360563-35360563 on Build GRCh38
rs538114454 Chr.19: 35360780-35360780 on Build GRCh38
rs938288556 Chr.19: 35360512-35360512 on Build GRCh38
rs996069148 Chr.19: 35360890-35360890 on Build GRCh38
rs530228294 Chr.19: 35360544-35360544 on Build GRCh38
rs920198590 Chr.19: 35360498-35360498 on Build GRCh38
rs184214974 Chr.19: 35360879-35360879 on Build GRCh38
rs375174972 Chr.19: 35360504-35360504 on Build GRCh38
rs902344682 Chr.19: 35360860-35360860 on Build GRCh38
rs983171523 Chr.19: 35360454-35360454 on Build GRCh38
rs1042532235 Chr.19: 35360795-35360795 on Build GRCh38
rs403862 Chr.19: 35360773-35360773 on Build GRCh38
rs1055104729 Chr.19: 35360555-35360555 on Build GRCh38
rs964958639 Chr.19: 35360881-35360881 on Build GRCh38
rs446226 Chr.19: 35360782-35360782 on Build GRCh38
rs868361413 Chr.19: 35360678-35360678 on Build GRCh38
rs940221523 Chr.19: 35360484-35360484 on Build GRCh38
rs896493228 Chr.19: 35360664-35360664 on Build GRCh38
rs10406885 Chr.19: 35360439-35360439 on Build GRCh38

View Full Product Details