Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35450908-35451433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FFAR2
Entrez Gene ID
2867
Gene Name
free fatty acid receptor 2
Gene Aliases
FFA2R, GPR43
UniGene
Hs.248056
Gene Chromosome Location
Chr.19: 35447965-35451767 on Build GRCh38
Transcripts
NM_005306.2 XM_017026709.1 XM_017026710.1 XM_017026711.1

SNP Detail

SNP ID Chromosome Location
rs62111703 Chr.19: 35451218-35451218 on Build GRCh38
rs416633 Chr.19: 35450977-35450977 on Build GRCh38
rs1033354499 Chr.19: 35451221-35451227 on Build GRCh38
rs956121889 Chr.19: 35451037-35451037 on Build GRCh38
rs894225897 Chr.19: 35451244-35451244 on Build GRCh38
rs113425207 Chr.19: 35451241-35451241 on Build GRCh38
rs780399437 Chr.19: 35451065-35451065 on Build GRCh38
rs1033155946 Chr.19: 35451317-35451317 on Build GRCh38
rs1028051993 Chr.19: 35451127-35451127 on Build GRCh38
rs896189438 Chr.19: 35451221-35451229 on Build GRCh38
rs1008353664 Chr.19: 35451404-35451404 on Build GRCh38
rs577031053 Chr.19: 35451089-35451089 on Build GRCh38
rs577416943 Chr.19: 35450957-35450957 on Build GRCh38
rs889551686 Chr.19: 35451395-35451395 on Build GRCh38
rs1013257006 Chr.19: 35451221-35451231 on Build GRCh38
rs898732808 Chr.19: 35451356-35451356 on Build GRCh38
rs926225082 Chr.19: 35451067-35451068 on Build GRCh38
rs938057888 Chr.19: 35451270-35451270 on Build GRCh38
rs55695970 Chr.19: 35451091-35451091 on Build GRCh38
rs562573504 Chr.19: 35451112-35451112 on Build GRCh38
rs374209813 Chr.19: 35451168-35451168 on Build GRCh38
rs1019832702 Chr.19: 35451172-35451172 on Build GRCh38
rs975600558 Chr.19: 35451386-35451386 on Build GRCh38
rs368664742 Chr.19: 35451347-35451347 on Build GRCh38
rs748425780 Chr.19: 35451158-35451158 on Build GRCh38
rs898199906 Chr.19: 35451093-35451093 on Build GRCh38
rs57187480 Chr.19: 35451116-35451116 on Build GRCh38
rs914486712 Chr.19: 35451201-35451201 on Build GRCh38
rs527936993 Chr.19: 35451291-35451291 on Build GRCh38
rs1049356161 Chr.19: 35451392-35451392 on Build GRCh38
rs773317719 Chr.19: 35451361-35451361 on Build GRCh38
rs374212678 Chr.19: 35451147-35451147 on Build GRCh38
rs781631500 Chr.19: 35451012-35451012 on Build GRCh38
rs912498671 Chr.19: 35451013-35451013 on Build GRCh38
rs995255200 Chr.19: 35450978-35450978 on Build GRCh38
rs957858283 Chr.19: 35451354-35451354 on Build GRCh38
rs1015639756 Chr.19: 35450976-35450976 on Build GRCh38
rs1056748825 Chr.19: 35450950-35450950 on Build GRCh38
rs978003783 Chr.19: 35451311-35451311 on Build GRCh38
rs185147923 Chr.19: 35451253-35451253 on Build GRCh38
rs113286222 Chr.19: 35451050-35451050 on Build GRCh38
rs926714882 Chr.19: 35451261-35451261 on Build GRCh38
rs772274832 Chr.19: 35451277-35451277 on Build GRCh38
rs370139892 Chr.19: 35451057-35451057 on Build GRCh38
rs867691513 Chr.19: 35451257-35451257 on Build GRCh38
rs564260757 Chr.19: 35451125-35451125 on Build GRCh38
rs989175432 Chr.19: 35451200-35451200 on Build GRCh38
rs60261537 Chr.19: 35451055-35451055 on Build GRCh38
rs1057003023 Chr.19: 35451072-35451072 on Build GRCh38
rs1028020667 Chr.19: 35450998-35450998 on Build GRCh38
rs1002088927 Chr.19: 35451218-35451218 on Build GRCh38
rs757669120 Chr.19: 35451011-35451011 on Build GRCh38
rs975401359 Chr.19: 35451080-35451080 on Build GRCh38
rs968070840 Chr.19: 35451212-35451212 on Build GRCh38
rs549506092 Chr.19: 35451148-35451148 on Build GRCh38
rs11332202 Chr.19: 35451243-35451244 on Build GRCh38
rs529885625 Chr.19: 35451153-35451153 on Build GRCh38
rs540233672 Chr.19: 35451041-35451041 on Build GRCh38
rs572269431 Chr.19: 35451068-35451068 on Build GRCh38
rs749131519 Chr.19: 35451078-35451078 on Build GRCh38
rs955986395 Chr.19: 35451184-35451184 on Build GRCh38
rs139874727 Chr.19: 35451051-35451051 on Build GRCh38
rs918117504 Chr.19: 35451278-35451278 on Build GRCh38
rs930997535 Chr.19: 35451387-35451387 on Build GRCh38
rs144374935 Chr.19: 35451066-35451066 on Build GRCh38
rs921222690 Chr.19: 35451084-35451084 on Build GRCh38
rs568666435 Chr.19: 35451280-35451280 on Build GRCh38
rs1056636000 Chr.19: 35451271-35451271 on Build GRCh38
rs1003894029 Chr.19: 35450966-35450966 on Build GRCh38
rs896740727 Chr.19: 35451075-35451075 on Build GRCh38

View Full Product Details