Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35500295-35500794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMKN
Entrez Gene ID
93099
Gene Name
dermokine
Gene Aliases
UNQ729, ZD52F10
UniGene
Hs.417795
Gene Chromosome Location
Chr.19: 35497217-35513678 on Build GRCh38
Transcripts
NM_001035516.3 NM_001126056.2 NM_001126059.2 NM_001190347.1 NM_001308380.1 NM_001308383.1 NM_033317.4 NR_033746.1 XM_006723477.1 XM_006723484.1 XM_006723489.1 XM_006723493.2 XM_006723494.2 XM_006723503.2 XM_011527494.2 XM_011527495.2 XM_011527496.2 XM_011527497.2 XM_011527498.2 XM_011527499.2 XM_011527500.2 XM_011527501.2 XM_011527502.2 XM_011527503.2 XM_011527504.2 XM_011527505.2 XM_011527506.2 XM_011527507.2 XM_011527508.2 XM_011527509.2 XM_011527510.2 XM_011527513.1 XM_011527514.1 XM_017027475.1 XM_017027476.1 XM_017027477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5735139 c.86C>T p.A29V Chr.19: 35500576-35500576 on Build GRCh38
3727555 c.379-2A>C p.? Chr.19: 35500582-35500582 on Build GRCh38
5735137 c.1121C>T p.A374V Chr.19: 35500576-35500576 on Build GRCh38
4843207 c.1164+7C>T p.? Chr.19: 35500526-35500526 on Build GRCh38
5735138 c.383C>T p.A128V Chr.19: 35500576-35500576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377520336 Chr.19: 35500590-35500590 on Build GRCh38
rs562332938 Chr.19: 35500363-35500363 on Build GRCh38
rs772286313 Chr.19: 35500345-35500345 on Build GRCh38
rs1001963534 Chr.19: 35500709-35500709 on Build GRCh38
rs61745872 Chr.19: 35500417-35500417 on Build GRCh38
rs1008540898 Chr.19: 35500418-35500418 on Build GRCh38
rs756815555 Chr.19: 35500464-35500464 on Build GRCh38
rs955342773 Chr.19: 35500520-35500520 on Build GRCh38
rs760080749 Chr.19: 35500548-35500548 on Build GRCh38
rs143476133 Chr.19: 35500543-35500543 on Build GRCh38
rs796187517 Chr.19: 35500539-35500540 on Build GRCh38
rs771123339 Chr.19: 35500355-35500355 on Build GRCh38
rs757512374 Chr.19: 35500544-35500544 on Build GRCh38
rs547929127 Chr.19: 35500441-35500441 on Build GRCh38
rs1030776242 Chr.19: 35500433-35500433 on Build GRCh38
rs982649378 Chr.19: 35500732-35500732 on Build GRCh38
rs774653766 Chr.19: 35500579-35500579 on Build GRCh38
rs781384998 Chr.19: 35500511-35500511 on Build GRCh38
rs1050629896 Chr.19: 35500398-35500398 on Build GRCh38
rs149828471 Chr.19: 35500576-35500576 on Build GRCh38
rs892119066 Chr.19: 35500635-35500635 on Build GRCh38
rs554728805 Chr.19: 35500596-35500596 on Build GRCh38
rs531375362 Chr.19: 35500758-35500758 on Build GRCh38
rs114875083 Chr.19: 35500722-35500722 on Build GRCh38
rs551998532 Chr.19: 35500350-35500350 on Build GRCh38
rs775832324 Chr.19: 35500532-35500532 on Build GRCh38
rs765749130 Chr.19: 35500607-35500607 on Build GRCh38
rs770385057 Chr.19: 35500521-35500521 on Build GRCh38
rs373665342 Chr.19: 35500751-35500751 on Build GRCh38
rs760298377 Chr.19: 35500603-35500603 on Build GRCh38
rs763208247 Chr.19: 35500432-35500432 on Build GRCh38
rs540294566 Chr.19: 35500620-35500620 on Build GRCh38
rs532251186 Chr.19: 35500346-35500346 on Build GRCh38
rs753819810 Chr.19: 35500609-35500609 on Build GRCh38
rs754696072 Chr.19: 35500490-35500490 on Build GRCh38
rs373319397 Chr.19: 35500552-35500552 on Build GRCh38
rs753707298 Chr.19: 35500553-35500553 on Build GRCh38
rs374771845 Chr.19: 35500575-35500575 on Build GRCh38
rs545651831 Chr.19: 35500679-35500679 on Build GRCh38
rs2293696 Chr.19: 35500577-35500577 on Build GRCh38
rs757510350 Chr.19: 35500571-35500571 on Build GRCh38
rs751861645 Chr.19: 35500570-35500570 on Build GRCh38
rs369555493 Chr.19: 35500519-35500519 on Build GRCh38
rs767849830 Chr.19: 35500624-35500624 on Build GRCh38
rs368869535 Chr.19: 35500492-35500492 on Build GRCh38
rs909072 Chr.19: 35500540-35500540 on Build GRCh38
rs1052130076 Chr.19: 35500633-35500633 on Build GRCh38
rs975246810 Chr.19: 35500509-35500509 on Build GRCh38
rs547354931 Chr.19: 35500538-35500538 on Build GRCh38
rs761476275 Chr.19: 35500584-35500584 on Build GRCh38
rs777291956 Chr.19: 35500356-35500356 on Build GRCh38
rs772707481 Chr.19: 35500597-35500597 on Build GRCh38
rs376517205 Chr.19: 35500505-35500505 on Build GRCh38
rs761383650 Chr.19: 35500542-35500542 on Build GRCh38
rs778784899 Chr.19: 35500569-35500569 on Build GRCh38
rs370675371 Chr.19: 35500615-35500615 on Build GRCh38
rs548192426 Chr.19: 35500500-35500500 on Build GRCh38
rs1038117799 Chr.19: 35500324-35500324 on Build GRCh38
rs758238877 Chr.19: 35500621-35500621 on Build GRCh38
rs79152633 Chr.19: 35500614-35500614 on Build GRCh38
rs899314065 Chr.19: 35500353-35500353 on Build GRCh38
rs749899125 Chr.19: 35500546-35500546 on Build GRCh38
rs576767456 Chr.19: 35500664-35500664 on Build GRCh38
rs997322519 Chr.19: 35500634-35500634 on Build GRCh38
rs1028187829 Chr.19: 35500664-35500664 on Build GRCh38
rs751796440 Chr.19: 35500449-35500450 on Build GRCh38
rs1022424517 Chr.19: 35500672-35500672 on Build GRCh38
rs2293695 Chr.19: 35500471-35500471 on Build GRCh38
rs563852315 Chr.19: 35500539-35500539 on Build GRCh38
rs754835367 Chr.19: 35500559-35500559 on Build GRCh38
rs962348752 Chr.19: 35500768-35500768 on Build GRCh38
rs771139816 Chr.19: 35500507-35500507 on Build GRCh38
rs767044882 Chr.19: 35500545-35500545 on Build GRCh38
rs754064311 Chr.19: 35500690-35500690 on Build GRCh38
rs746002914 Chr.19: 35500573-35500573 on Build GRCh38
rs374804708 Chr.19: 35500474-35500474 on Build GRCh38
rs751206014 Chr.19: 35500443-35500443 on Build GRCh38
rs111670139 Chr.19: 35500769-35500769 on Build GRCh38
rs1034917926 Chr.19: 35500716-35500716 on Build GRCh38
rs148011601 Chr.19: 35500625-35500625 on Build GRCh38
rs779735752 Chr.19: 35500473-35500473 on Build GRCh38
rs766952702 Chr.19: 35500465-35500465 on Build GRCh38
rs919572807 Chr.19: 35500752-35500752 on Build GRCh38
rs765556258 Chr.19: 35500376-35500376 on Build GRCh38
rs761780049 Chr.19: 35500606-35500606 on Build GRCh38
rs781252940 Chr.19: 35500572-35500572 on Build GRCh38
rs750328774 Chr.19: 35500467-35500467 on Build GRCh38
rs973710427 Chr.19: 35500750-35500750 on Build GRCh38

View Full Product Details