Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35641666-35642173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV2
Entrez Gene ID
2116
Gene Name
ETS variant 2
Gene Aliases
ER71, ETSRP71
UniGene
Hs.194061
Gene Chromosome Location
Chr.19: 35641175-35644871 on Build GRCh38
Transcripts
NM_001300974.1 NM_001304549.1 NM_014209.3 XM_005258652.2 XM_011526624.2 XM_017026472.1

SNP Detail

SNP ID Chromosome Location
rs906764757 Chr.19: 35641788-35641788 on Build GRCh38
rs960967569 Chr.19: 35641773-35641773 on Build GRCh38
rs1011360784 Chr.19: 35642033-35642033 on Build GRCh38
rs567094441 Chr.19: 35641704-35641704 on Build GRCh38
rs191624815 Chr.19: 35641985-35641985 on Build GRCh38
rs962242701 Chr.19: 35642010-35642010 on Build GRCh38
rs773027650 Chr.19: 35641953-35641953 on Build GRCh38
rs905261315 Chr.19: 35642082-35642082 on Build GRCh38
rs980775648 Chr.19: 35641754-35641754 on Build GRCh38
rs58470815 Chr.19: 35642020-35642020 on Build GRCh38
rs1050754626 Chr.19: 35641972-35641972 on Build GRCh38
rs552280261 Chr.19: 35641759-35641759 on Build GRCh38
rs568317731 Chr.19: 35642055-35642055 on Build GRCh38
rs765855059 Chr.19: 35642138-35642138 on Build GRCh38
rs532563278 Chr.19: 35641748-35641748 on Build GRCh38
rs187202985 Chr.19: 35641870-35641870 on Build GRCh38
rs1052973342 Chr.19: 35642004-35642004 on Build GRCh38
rs535345030 Chr.19: 35642139-35642139 on Build GRCh38
rs949858227 Chr.19: 35642090-35642090 on Build GRCh38
rs932421638 Chr.19: 35641969-35641969 on Build GRCh38
rs899621506 Chr.19: 35641688-35641688 on Build GRCh38
rs1051231368 Chr.19: 35641693-35641693 on Build GRCh38
rs1022774838 Chr.19: 35642046-35642046 on Build GRCh38
rs941503952 Chr.19: 35641910-35641910 on Build GRCh38
rs930373056 Chr.19: 35641746-35641746 on Build GRCh38
rs987861401 Chr.19: 35641965-35641966 on Build GRCh38
rs996517745 Chr.19: 35641689-35641689 on Build GRCh38
rs763420289 Chr.19: 35641736-35641736 on Build GRCh38
rs999196585 Chr.19: 35641699-35641699 on Build GRCh38
rs1022327410 Chr.19: 35641737-35641737 on Build GRCh38
rs908677868 Chr.19: 35641887-35641887 on Build GRCh38
rs953483353 Chr.19: 35641941-35641941 on Build GRCh38
rs920997243 Chr.19: 35641940-35641940 on Build GRCh38
rs1029072309 Chr.19: 35641892-35641892 on Build GRCh38
rs878860716 Chr.19: 35641863-35641863 on Build GRCh38
rs894094168 Chr.19: 35642150-35642150 on Build GRCh38
rs1031562540 Chr.19: 35641840-35641840 on Build GRCh38
rs902864465 Chr.19: 35642136-35642136 on Build GRCh38
rs1045780740 Chr.19: 35642101-35642101 on Build GRCh38
rs890804460 Chr.19: 35641974-35641974 on Build GRCh38
rs1038588254 Chr.19: 35641915-35641915 on Build GRCh38
rs983443716 Chr.19: 35641789-35641789 on Build GRCh38
rs957202940 Chr.19: 35641706-35641706 on Build GRCh38
rs1055591894 Chr.19: 35642062-35642062 on Build GRCh38
rs1010909960 Chr.19: 35641712-35641712 on Build GRCh38
rs534103733 Chr.19: 35642086-35642086 on Build GRCh38
rs891026966 Chr.19: 35641695-35641695 on Build GRCh38
rs899012214 Chr.19: 35641741-35641741 on Build GRCh38
rs1031475277 Chr.19: 35641700-35641700 on Build GRCh38
rs1002719757 Chr.19: 35642085-35642085 on Build GRCh38
rs982838664 Chr.19: 35642149-35642149 on Build GRCh38
rs911928030 Chr.19: 35642005-35642005 on Build GRCh38
rs114346731 Chr.19: 35641859-35641859 on Build GRCh38

View Full Product Details