Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35642060-35642561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV2
Entrez Gene ID
2116
Gene Name
ETS variant 2
Gene Aliases
ER71, ETSRP71
UniGene
Hs.194061
Gene Chromosome Location
Chr.19: 35641175-35644871 on Build GRCh38
Transcripts
NM_001300974.1 NM_001304549.1 NM_014209.3 XM_005258652.2 XM_011526624.2 XM_017026472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6465696 c.38A>G p.E13G Chr.19: 35642498-35642498 on Build GRCh38
5297640 c.25G>A p.A9T Chr.19: 35642485-35642485 on Build GRCh38
5725161 c.29C>T p.S10F Chr.19: 35642489-35642489 on Build GRCh38
7243844 c.52A>T p.N18Y Chr.19: 35642512-35642512 on Build GRCh38
6465694 c.37G>A p.E13K Chr.19: 35642497-35642497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112463460 Chr.19: 35642271-35642271 on Build GRCh38
rs184613720 Chr.19: 35642188-35642188 on Build GRCh38
rs775817404 Chr.19: 35642512-35642512 on Build GRCh38
rs539624523 Chr.19: 35642234-35642234 on Build GRCh38
rs1029191611 Chr.19: 35642229-35642229 on Build GRCh38
rs376202778 Chr.19: 35642285-35642285 on Build GRCh38
rs905261315 Chr.19: 35642082-35642082 on Build GRCh38
rs962979932 Chr.19: 35642175-35642175 on Build GRCh38
rs560390251 Chr.19: 35642386-35642386 on Build GRCh38
rs868182130 Chr.19: 35642407-35642407 on Build GRCh38
rs113982759 Chr.19: 35642322-35642322 on Build GRCh38
rs765855059 Chr.19: 35642138-35642138 on Build GRCh38
rs759903540 Chr.19: 35642429-35642429 on Build GRCh38
rs535345030 Chr.19: 35642139-35642139 on Build GRCh38
rs924236268 Chr.19: 35642462-35642462 on Build GRCh38
rs532601843 Chr.19: 35642540-35642540 on Build GRCh38
rs949858227 Chr.19: 35642090-35642090 on Build GRCh38
rs1045486699 Chr.19: 35642455-35642455 on Build GRCh38
rs1004198304 Chr.19: 35642494-35642494 on Build GRCh38
rs188932640 Chr.19: 35642238-35642238 on Build GRCh38
rs934312513 Chr.19: 35642252-35642252 on Build GRCh38
rs377390321 Chr.19: 35642430-35642430 on Build GRCh38
rs921445149 Chr.19: 35642235-35642235 on Build GRCh38
rs1008997904 Chr.19: 35642237-35642237 on Build GRCh38
rs192862284 Chr.19: 35642365-35642365 on Build GRCh38
rs1035508809 Chr.19: 35642395-35642395 on Build GRCh38
rs748082828 Chr.19: 35642433-35642433 on Build GRCh38
rs771992931 Chr.19: 35642435-35642435 on Build GRCh38
rs896609842 Chr.19: 35642415-35642415 on Build GRCh38
rs576225046 Chr.19: 35642242-35642242 on Build GRCh38
rs777489182 Chr.19: 35642445-35642445 on Build GRCh38
rs867491373 Chr.19: 35642507-35642507 on Build GRCh38
rs763222939 Chr.19: 35642531-35642531 on Build GRCh38
rs771462330 Chr.19: 35642343-35642343 on Build GRCh38
rs369643154 Chr.19: 35642538-35642538 on Build GRCh38
rs561730660 Chr.19: 35642348-35642348 on Build GRCh38
rs949815351 Chr.19: 35642397-35642397 on Build GRCh38
rs912195266 Chr.19: 35642250-35642250 on Build GRCh38
rs894094168 Chr.19: 35642150-35642150 on Build GRCh38
rs902864465 Chr.19: 35642136-35642136 on Build GRCh38
rs1045780740 Chr.19: 35642101-35642101 on Build GRCh38
rs534103733 Chr.19: 35642086-35642086 on Build GRCh38
rs78901955 Chr.19: 35642362-35642362 on Build GRCh38
rs754506693 Chr.19: 35642427-35642427 on Build GRCh38
rs1002719757 Chr.19: 35642085-35642085 on Build GRCh38
rs982838664 Chr.19: 35642149-35642149 on Build GRCh38
rs1015852968 Chr.19: 35642161-35642161 on Build GRCh38
rs1052756791 Chr.19: 35642259-35642259 on Build GRCh38
rs185880517 Chr.19: 35642373-35642373 on Build GRCh38
rs972357780 Chr.19: 35642262-35642262 on Build GRCh38
rs577157712 Chr.19: 35642194-35642194 on Build GRCh38
rs146305038 Chr.19: 35642513-35642513 on Build GRCh38
rs765004679 Chr.19: 35642504-35642504 on Build GRCh38
rs541511639 Chr.19: 35642282-35642282 on Build GRCh38
rs746800422 Chr.19: 35642481-35642481 on Build GRCh38
rs768856519 Chr.19: 35642532-35642532 on Build GRCh38
rs572419054 Chr.19: 35642500-35642500 on Build GRCh38

View Full Product Details