Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36142564-36143066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPNS1
Entrez Gene ID
826
Gene Name
calpain small subunit 1
Gene Aliases
CALPAIN4, CANP, CANPS, CAPN4, CDPS, CSS1
UniGene
Hs.515371
Gene Chromosome Location
Chr.19: 36139926-36150353 on Build GRCh38
Transcripts
NM_001003962.2 NM_001302632.1 NM_001302633.1 NM_001749.3 XM_005259295.1 XM_005259296.1 XM_011527359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2150428 c.292G>T p.V98F Chr.19: 36142700-36142700 on Build GRCh38
1194774 c.305G>C p.R102P Chr.19: 36142713-36142713 on Build GRCh38
995387 c.305G>A p.R102Q Chr.19: 36142713-36142713 on Build GRCh38
5768135 c.360C>T p.L120L Chr.19: 36142935-36142935 on Build GRCh38
5028552 c.282G>C p.E94D Chr.19: 36142690-36142690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754177544 Chr.19: 36142923-36142923 on Build GRCh38
rs779190677 Chr.19: 36143042-36143042 on Build GRCh38
rs374645485 Chr.19: 36142713-36142713 on Build GRCh38
rs755984538 Chr.19: 36142627-36142627 on Build GRCh38
rs747083464 Chr.19: 36142963-36142963 on Build GRCh38
rs780842861 Chr.19: 36142913-36142913 on Build GRCh38
rs11539824 Chr.19: 36142944-36142944 on Build GRCh38
rs767610570 Chr.19: 36142682-36142682 on Build GRCh38
rs972968845 Chr.19: 36142692-36142692 on Build GRCh38
rs186561390 Chr.19: 36142832-36142832 on Build GRCh38
rs878988441 Chr.19: 36142801-36142801 on Build GRCh38
rs761900601 Chr.19: 36142677-36142677 on Build GRCh38
rs759403474 Chr.19: 36142976-36142976 on Build GRCh38
rs200941006 Chr.19: 36142948-36142948 on Build GRCh38
rs966915244 Chr.19: 36142978-36142978 on Build GRCh38
rs778499068 Chr.19: 36142643-36142643 on Build GRCh38
rs763604929 Chr.19: 36142882-36142882 on Build GRCh38
rs1001134137 Chr.19: 36142603-36142603 on Build GRCh38
rs755375003 Chr.19: 36142927-36142927 on Build GRCh38
rs770182217 Chr.19: 36142601-36142601 on Build GRCh38
rs750134961 Chr.19: 36142624-36142624 on Build GRCh38
rs777534419 Chr.19: 36142651-36142651 on Build GRCh38
rs374159469 Chr.19: 36142596-36142596 on Build GRCh38
rs771258476 Chr.19: 36142966-36142966 on Build GRCh38
rs761276103 Chr.19: 36142990-36142990 on Build GRCh38
rs747547235 Chr.19: 36142986-36142986 on Build GRCh38
rs112854764 Chr.19: 36142744-36142744 on Build GRCh38
rs373217382 Chr.19: 36142645-36142645 on Build GRCh38
rs751028539 Chr.19: 36142984-36142984 on Build GRCh38
rs774886573 Chr.19: 36142770-36142770 on Build GRCh38
rs1036954331 Chr.19: 36142843-36142843 on Build GRCh38
rs756889786 Chr.19: 36142905-36142905 on Build GRCh38
rs751032918 Chr.19: 36142904-36142904 on Build GRCh38
rs549840423 Chr.19: 36142699-36142699 on Build GRCh38
rs760886049 Chr.19: 36142781-36142781 on Build GRCh38
rs769151310 Chr.19: 36142670-36142670 on Build GRCh38
rs985422780 Chr.19: 36142739-36142739 on Build GRCh38
rs748746397 Chr.19: 36142943-36142943 on Build GRCh38
rs753892238 Chr.19: 36142635-36142635 on Build GRCh38
rs756870116 Chr.19: 36142726-36142726 on Build GRCh38
rs777291107 Chr.19: 36142724-36142724 on Build GRCh38
rs961562994 Chr.19: 36142678-36142678 on Build GRCh38
rs748144226 Chr.19: 36142776-36142776 on Build GRCh38
rs767808433 Chr.19: 36142619-36142619 on Build GRCh38
rs754720342 Chr.19: 36142674-36142674 on Build GRCh38
rs113501827 Chr.19: 36142650-36142650 on Build GRCh38
rs757940775 Chr.19: 36142646-36142646 on Build GRCh38
rs199893278 Chr.19: 36143021-36143021 on Build GRCh38
rs1014206376 Chr.19: 36142900-36142900 on Build GRCh38
rs995526534 Chr.19: 36142859-36142859 on Build GRCh38
rs112429279 Chr.19: 36142743-36142743 on Build GRCh38
rs765239180 Chr.19: 36142875-36142875 on Build GRCh38
rs761953872 Chr.19: 36142618-36142618 on Build GRCh38
rs757838403 Chr.19: 36142880-36142880 on Build GRCh38
rs376072907 Chr.19: 36142852-36142852 on Build GRCh38
rs935699381 Chr.19: 36142718-36142718 on Build GRCh38
rs780325714 Chr.19: 36142737-36142737 on Build GRCh38
rs369323031 Chr.19: 36142780-36142780 on Build GRCh38
rs190077569 Chr.19: 36142758-36142758 on Build GRCh38
rs773292355 Chr.19: 36142780-36142780 on Build GRCh38
rs759454444 Chr.19: 36142869-36142869 on Build GRCh38
rs140322191 Chr.19: 36142942-36142942 on Build GRCh38
rs751647878 Chr.19: 36142617-36142617 on Build GRCh38
rs773523619 Chr.19: 36142683-36142683 on Build GRCh38
rs750141458 Chr.19: 36143012-36143012 on Build GRCh38
rs747694630 Chr.19: 36142644-36142644 on Build GRCh38
rs182191517 Chr.19: 36142777-36142777 on Build GRCh38
rs777036361 Chr.19: 36142967-36142967 on Build GRCh38
rs977126502 Chr.19: 36142706-36142706 on Build GRCh38
rs17878500 Chr.19: 36142962-36142962 on Build GRCh38
rs754928347 Chr.19: 36142707-36142707 on Build GRCh38
rs560846443 Chr.19: 36142668-36142668 on Build GRCh38
rs779823964 Chr.19: 36142628-36142628 on Build GRCh38
rs113039946 Chr.19: 36142586-36142586 on Build GRCh38
rs202081010 Chr.19: 36142671-36142671 on Build GRCh38
rs959554716 Chr.19: 36142891-36142891 on Build GRCh38
rs371312155 Chr.19: 36142658-36142658 on Build GRCh38
rs866083785 Chr.19: 36142659-36142659 on Build GRCh38
rs759073283 Chr.19: 36142865-36142866 on Build GRCh38
rs978344509 Chr.19: 36142982-36142982 on Build GRCh38
rs758520101 Chr.19: 36142959-36142959 on Build GRCh38
rs116114216 Chr.19: 36142657-36142657 on Build GRCh38
rs752750806 Chr.19: 36142877-36142877 on Build GRCh38
rs766542228 Chr.19: 36142867-36142867 on Build GRCh38
rs765290273 Chr.19: 36142712-36142712 on Build GRCh38
rs776109817 Chr.19: 36142605-36142605 on Build GRCh38
rs757969728 Chr.19: 36142717-36142717 on Build GRCh38
rs766140029 Chr.19: 36142687-36142687 on Build GRCh38
rs767124270 Chr.19: 36143011-36143011 on Build GRCh38
rs763020514 Chr.19: 36142607-36142607 on Build GRCh38
rs367819061 Chr.19: 36142612-36142612 on Build GRCh38
rs760381510 Chr.19: 36142684-36142684 on Build GRCh38
rs371615875 Chr.19: 36142851-36142851 on Build GRCh38
rs769099865 Chr.19: 36142767-36142767 on Build GRCh38
rs775165678 Chr.19: 36142979-36142979 on Build GRCh38
rs775851016 Chr.19: 36142662-36142662 on Build GRCh38
rs10424687 Chr.19: 36142933-36142933 on Build GRCh38
rs749681659 Chr.19: 36142753-36142753 on Build GRCh38

View Full Product Details