Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38217404-38217910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPF1
Entrez Gene ID
8193
Gene Name
double PHD fingers 1
Gene Aliases
BAF45b, NEUD4, neuro-d4
UniGene
Hs.631576
Gene Chromosome Location
Chr.19: 38211006-38229714 on Build GRCh38
Transcripts
NM_001135155.2 NM_001135156.2 NM_001289978.1 NM_004647.3 XM_005259289.2 XM_005259290.3 XM_005259292.4 XM_006723407.2 XM_006723408.3 XM_006723409.2 XM_006723410.2 XM_011527356.1 XM_011527357.2 XM_011527358.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408564 c.571C>T p.R191* Chr.19: 38217822-38217822 on Build GRCh38
5855634 c.662C>T p.P221L Chr.19: 38217812-38217812 on Build GRCh38
6422647 c.578A>T p.K193M Chr.19: 38217815-38217815 on Build GRCh38
4679235 c.544C>T p.R182C Chr.19: 38217849-38217849 on Build GRCh38
6345628 c.580C>A p.P194T Chr.19: 38217813-38217813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925476645 Chr.19: 38217496-38217496 on Build GRCh38
rs779313294 Chr.19: 38217777-38217777 on Build GRCh38
rs775382744 Chr.19: 38217884-38217887 on Build GRCh38
rs747431265 Chr.19: 38217819-38217819 on Build GRCh38
rs747124096 Chr.19: 38217601-38217601 on Build GRCh38
rs371151271 Chr.19: 38217874-38217874 on Build GRCh38
rs753044378 Chr.19: 38217780-38217780 on Build GRCh38
rs774380444 Chr.19: 38217854-38217854 on Build GRCh38
rs576425865 Chr.19: 38217442-38217442 on Build GRCh38
rs971143147 Chr.19: 38217697-38217697 on Build GRCh38
rs778346952 Chr.19: 38217817-38217817 on Build GRCh38
rs938603849 Chr.19: 38217840-38217840 on Build GRCh38
rs376720192 Chr.19: 38217791-38217791 on Build GRCh38
rs758821009 Chr.19: 38217881-38217881 on Build GRCh38
rs995135326 Chr.19: 38217566-38217566 on Build GRCh38
rs541108299 Chr.19: 38217710-38217710 on Build GRCh38
rs200830027 Chr.19: 38217756-38217756 on Build GRCh38
rs755728378 Chr.19: 38217772-38217772 on Build GRCh38
rs141451158 Chr.19: 38217811-38217811 on Build GRCh38
rs932478281 Chr.19: 38217426-38217426 on Build GRCh38
rs896367228 Chr.19: 38217745-38217745 on Build GRCh38
rs565385226 Chr.19: 38217676-38217676 on Build GRCh38
rs988228973 Chr.19: 38217855-38217855 on Build GRCh38
rs139810469 Chr.19: 38217643-38217643 on Build GRCh38
rs756919311 Chr.19: 38217490-38217490 on Build GRCh38
rs993995900 Chr.19: 38217737-38217737 on Build GRCh38
rs780913902 Chr.19: 38217517-38217517 on Build GRCh38
rs753364216 Chr.19: 38217879-38217879 on Build GRCh38
rs145138452 Chr.19: 38217837-38217837 on Build GRCh38
rs138969701 Chr.19: 38217838-38217838 on Build GRCh38
rs781399025 Chr.19: 38217822-38217822 on Build GRCh38
rs933338468 Chr.19: 38217631-38217631 on Build GRCh38
rs755371004 Chr.19: 38217532-38217532 on Build GRCh38
rs773220186 Chr.19: 38217600-38217600 on Build GRCh38
rs759782498 Chr.19: 38217738-38217738 on Build GRCh38
rs969513094 Chr.19: 38217478-38217478 on Build GRCh38
rs772626735 Chr.19: 38217599-38217599 on Build GRCh38
rs923522257 Chr.19: 38217630-38217630 on Build GRCh38
rs977053052 Chr.19: 38217706-38217706 on Build GRCh38
rs777024292 Chr.19: 38217610-38217610 on Build GRCh38
rs146689969 Chr.19: 38217721-38217721 on Build GRCh38
rs779203063 Chr.19: 38217563-38217563 on Build GRCh38
rs761611223 Chr.19: 38217861-38217861 on Build GRCh38
rs762821032 Chr.19: 38217849-38217849 on Build GRCh38
rs769894743 Chr.19: 38217608-38217608 on Build GRCh38
rs890559547 Chr.19: 38217505-38217505 on Build GRCh38
rs750102485 Chr.19: 38217768-38217768 on Build GRCh38
rs956854755 Chr.19: 38217545-38217545 on Build GRCh38
rs766901326 Chr.19: 38217764-38217764 on Build GRCh38
rs2304174 Chr.19: 38217689-38217689 on Build GRCh38
rs1008890988 Chr.19: 38217513-38217513 on Build GRCh38
rs751398827 Chr.19: 38217759-38217759 on Build GRCh38
rs527478375 Chr.19: 38217743-38217743 on Build GRCh38
rs770848427 Chr.19: 38217605-38217605 on Build GRCh38
rs1013051374 Chr.19: 38217677-38217677 on Build GRCh38
rs1037773732 Chr.19: 38217692-38217692 on Build GRCh38
rs767067156 Chr.19: 38217867-38217867 on Build GRCh38
rs774560536 Chr.19: 38217757-38217757 on Build GRCh38
rs1028330730 Chr.19: 38217579-38217579 on Build GRCh38
rs745459511 Chr.19: 38217526-38217526 on Build GRCh38
rs771065602 Chr.19: 38217821-38217821 on Build GRCh38
rs911961796 Chr.19: 38217803-38217803 on Build GRCh38
rs748689849 Chr.19: 38217564-38217564 on Build GRCh38
rs2304175 Chr.19: 38217758-38217758 on Build GRCh38
rs768271396 Chr.19: 38217850-38217850 on Build GRCh38

View Full Product Details