Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38352170-38352678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERG
Entrez Gene ID
57828
Gene Name
cation channel sperm associated auxiliary subunit gamma
Gene Aliases
C19orf15
UniGene
Hs.324335
Gene Chromosome Location
Chr.19: 38335563-38370949 on Build GRCh38
Transcripts
NM_021185.4 XM_005259114.4 XM_005259115.4 XM_005259116.2 XM_006723308.3 XM_006723310.2 XM_006723311.2 XM_011527174.2 XM_011527175.2 XM_011527176.2 XM_011527177.2 XM_011527178.2 XM_011527179.1 XM_011527182.2 XM_017027053.1 XM_017027054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6017472 c.1015C>T p.L339L Chr.19: 38352474-38352474 on Build GRCh38
5351908 c.989G>A p.R330H Chr.19: 38352424-38352424 on Build GRCh38
5106138 c.859G>A p.A287T Chr.19: 38352318-38352318 on Build GRCh38
1612171 c.1036+9G>T p.? Chr.19: 38352504-38352504 on Build GRCh38
5351909 c.965G>A p.R322H Chr.19: 38352424-38352424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778480657 Chr.19: 38352597-38352598 on Build GRCh38
rs910052120 Chr.19: 38352364-38352364 on Build GRCh38
rs1031808021 Chr.19: 38352457-38352457 on Build GRCh38
rs186593851 Chr.19: 38352262-38352262 on Build GRCh38
rs368632875 Chr.19: 38352477-38352477 on Build GRCh38
rs764197189 Chr.19: 38352396-38352396 on Build GRCh38
rs765527935 Chr.19: 38352322-38352322 on Build GRCh38
rs759710702 Chr.19: 38352288-38352288 on Build GRCh38
rs985659834 Chr.19: 38352579-38352579 on Build GRCh38
rs754826970 Chr.19: 38352540-38352540 on Build GRCh38
rs972210659 Chr.19: 38352596-38352600 on Build GRCh38
rs944397745 Chr.19: 38352345-38352345 on Build GRCh38
rs1022566121 Chr.19: 38352492-38352493 on Build GRCh38
rs897253172 Chr.19: 38352408-38352408 on Build GRCh38
rs1024053557 Chr.19: 38352434-38352434 on Build GRCh38
rs370881014 Chr.19: 38352336-38352336 on Build GRCh38
rs941563318 Chr.19: 38352382-38352382 on Build GRCh38
rs1029277105 Chr.19: 38352547-38352547 on Build GRCh38
rs568606872 Chr.19: 38352290-38352292 on Build GRCh38
rs755811756 Chr.19: 38352429-38352429 on Build GRCh38
rs117832024 Chr.19: 38352194-38352194 on Build GRCh38
rs986298762 Chr.19: 38352531-38352531 on Build GRCh38
rs529790200 Chr.19: 38352448-38352448 on Build GRCh38
rs748577403 Chr.19: 38352484-38352484 on Build GRCh38
rs989474451 Chr.19: 38352491-38352491 on Build GRCh38
rs548975126 Chr.19: 38352564-38352564 on Build GRCh38
rs571505750 Chr.19: 38352265-38352265 on Build GRCh38
rs915437784 Chr.19: 38352202-38352202 on Build GRCh38
rs1037513424 Chr.19: 38352383-38352383 on Build GRCh38
rs1004030873 Chr.19: 38352451-38352451 on Build GRCh38
rs1045575393 Chr.19: 38352648-38352648 on Build GRCh38
rs377138163 Chr.19: 38352593-38352593 on Build GRCh38
rs565645654 Chr.19: 38352425-38352425 on Build GRCh38
rs61179863 Chr.19: 38352575-38352575 on Build GRCh38
rs894400497 Chr.19: 38352644-38352644 on Build GRCh38
rs747494216 Chr.19: 38352517-38352517 on Build GRCh38
rs757166002 Chr.19: 38352423-38352423 on Build GRCh38
rs763331703 Chr.19: 38352195-38352195 on Build GRCh38
rs955884518 Chr.19: 38352253-38352253 on Build GRCh38
rs779666820 Chr.19: 38352433-38352433 on Build GRCh38
rs530867698 Chr.19: 38352374-38352374 on Build GRCh38
rs372812402 Chr.19: 38352424-38352424 on Build GRCh38
rs1050553480 Chr.19: 38352264-38352264 on Build GRCh38
rs948637070 Chr.19: 38352645-38352645 on Build GRCh38
rs1036066382 Chr.19: 38352355-38352355 on Build GRCh38
rs773568577 Chr.19: 38352501-38352501 on Build GRCh38
rs369830498 Chr.19: 38352440-38352441 on Build GRCh38
rs747766955 Chr.19: 38352596-38352597 on Build GRCh38
rs985710593 Chr.19: 38352347-38352347 on Build GRCh38
rs575694671 Chr.19: 38352437-38352438 on Build GRCh38
rs758257271 Chr.19: 38352344-38352344 on Build GRCh38
rs745349659 Chr.19: 38352427-38352427 on Build GRCh38
rs932157813 Chr.19: 38352257-38352257 on Build GRCh38
rs576701757 Chr.19: 38352596-38352596 on Build GRCh38
rs772429826 Chr.19: 38352489-38352489 on Build GRCh38
rs879137466 Chr.19: 38352571-38352571 on Build GRCh38
rs1000060846 Chr.19: 38352453-38352453 on Build GRCh38
rs879209440 Chr.19: 38352561-38352561 on Build GRCh38
rs930322846 Chr.19: 38352596-38352605 on Build GRCh38
rs764395187 Chr.19: 38352426-38352426 on Build GRCh38
rs200982088 Chr.19: 38352363-38352363 on Build GRCh38
rs796292378 Chr.19: 38352385-38352385 on Build GRCh38
rs955833879 Chr.19: 38352474-38352474 on Build GRCh38
rs548423597 Chr.19: 38352454-38352454 on Build GRCh38
rs1048691794 Chr.19: 38352596-38352606 on Build GRCh38
rs569895559 Chr.19: 38352522-38352522 on Build GRCh38
rs751460339 Chr.19: 38352414-38352414 on Build GRCh38
rs918952323 Chr.19: 38352596-38352604 on Build GRCh38
rs776725564 Chr.19: 38352255-38352255 on Build GRCh38

View Full Product Details