Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38408759-38409254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98C
Entrez Gene ID
147965
Gene Name
family with sequence similarity 98 member C
Gene Aliases
-
UniGene
Hs.355162
Gene Chromosome Location
Chr.19: 38403124-38409088 on Build GRCh38
Transcripts
NM_174905.3 NM_001146202.1 NM_001146203.1 NM_001146204.1 NM_001146205.1 NM_001146206.1 NM_001146207.1 NM_170604.2

Gene Symbol
RASGRP4
Entrez Gene ID
115727
Gene Name
RAS guanyl releasing protein 4
Gene Aliases
-
UniGene
Hs.130434
Gene Chromosome Location
Chr.19: 38409055-38426305 on Build GRCh38
Transcripts
NM_174905.3 NM_001146202.1 NM_001146203.1 NM_001146204.1 NM_001146205.1 NM_001146206.1 NM_001146207.1 NM_170604.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3533352 c.940G>A p.D314N Chr.19: 38408772-38408772 on Build GRCh38
6481841 c.953G>T p.R318L Chr.19: 38408785-38408785 on Build GRCh38
3533353 c.979C>T p.P327S Chr.19: 38408811-38408811 on Build GRCh38
6660361 c.953G>A p.R318H Chr.19: 38408785-38408785 on Build GRCh38
3746487 c.1048T>A p.*350K Chr.19: 38408880-38408880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188318328 Chr.19: 38409195-38409195 on Build GRCh38
rs1042557956 Chr.19: 38408950-38408950 on Build GRCh38
rs948286928 Chr.19: 38409144-38409144 on Build GRCh38
rs781334100 Chr.19: 38408879-38408879 on Build GRCh38
rs981862547 Chr.19: 38409078-38409078 on Build GRCh38
rs976876731 Chr.19: 38409116-38409116 on Build GRCh38
rs527286799 Chr.19: 38409127-38409127 on Build GRCh38
rs944851309 Chr.19: 38409176-38409176 on Build GRCh38
rs748712642 Chr.19: 38408825-38408825 on Build GRCh38
rs961514760 Chr.19: 38409045-38409045 on Build GRCh38
rs536487743 Chr.19: 38408954-38408954 on Build GRCh38
rs754022580 Chr.19: 38408781-38408781 on Build GRCh38
rs777190647 Chr.19: 38408909-38408909 on Build GRCh38
rs773547682 Chr.19: 38408827-38408827 on Build GRCh38
rs145249718 Chr.19: 38409005-38409005 on Build GRCh38
rs553897186 Chr.19: 38408936-38408936 on Build GRCh38
rs371810386 Chr.19: 38408808-38408808 on Build GRCh38
rs964812738 Chr.19: 38408980-38408980 on Build GRCh38
rs902653492 Chr.19: 38409161-38409161 on Build GRCh38
rs111788975 Chr.19: 38409142-38409142 on Build GRCh38
rs887783886 Chr.19: 38409207-38409207 on Build GRCh38
rs770346857 Chr.19: 38409037-38409037 on Build GRCh38
rs531387005 Chr.19: 38409149-38409149 on Build GRCh38
rs778071249 Chr.19: 38408922-38408927 on Build GRCh38
rs777540452 Chr.19: 38408794-38408794 on Build GRCh38
rs767622128 Chr.19: 38409229-38409229 on Build GRCh38
rs752762275 Chr.19: 38408783-38408783 on Build GRCh38
rs3745964 Chr.19: 38408908-38408908 on Build GRCh38
rs894050364 Chr.19: 38408925-38408925 on Build GRCh38
rs1005437662 Chr.19: 38409211-38409211 on Build GRCh38
rs779826981 Chr.19: 38408898-38408898 on Build GRCh38
rs189945269 Chr.19: 38408904-38408904 on Build GRCh38
rs142790326 Chr.19: 38409230-38409230 on Build GRCh38
rs1018737836 Chr.19: 38409092-38409092 on Build GRCh38
rs764264994 Chr.19: 38408860-38408860 on Build GRCh38
rs775998597 Chr.19: 38409085-38409085 on Build GRCh38
rs761649590 Chr.19: 38408864-38408865 on Build GRCh38
rs373686254 Chr.19: 38408818-38408818 on Build GRCh38
rs984445729 Chr.19: 38409035-38409035 on Build GRCh38
rs892205778 Chr.19: 38409177-38409177 on Build GRCh38
rs138806414 Chr.19: 38409187-38409187 on Build GRCh38
rs559743819 Chr.19: 38409059-38409059 on Build GRCh38
rs781137669 Chr.19: 38408803-38408803 on Build GRCh38
rs778628339 Chr.19: 38408862-38408867 on Build GRCh38
rs562657477 Chr.19: 38409017-38409017 on Build GRCh38
rs779846569 Chr.19: 38408788-38408789 on Build GRCh38
rs200397704 Chr.19: 38408905-38408905 on Build GRCh38
rs375320861 Chr.19: 38408776-38408776 on Build GRCh38
rs116403677 Chr.19: 38409227-38409227 on Build GRCh38
rs142977446 Chr.19: 38408861-38408862 on Build GRCh38
rs768258248 Chr.19: 38408826-38408826 on Build GRCh38
rs765442289 Chr.19: 38408844-38408844 on Build GRCh38
rs953235439 Chr.19: 38408877-38408878 on Build GRCh38
rs990441022 Chr.19: 38409138-38409138 on Build GRCh38
rs756010256 Chr.19: 38408888-38408888 on Build GRCh38
rs759954394 Chr.19: 38408843-38408843 on Build GRCh38
rs567564690 Chr.19: 38408901-38408901 on Build GRCh38
rs758571821 Chr.19: 38408784-38408784 on Build GRCh38
rs755704162 Chr.19: 38408776-38408776 on Build GRCh38
rs746989100 Chr.19: 38408796-38408796 on Build GRCh38
rs751384843 Chr.19: 38408868-38408868 on Build GRCh38
rs766666619 Chr.19: 38408839-38408839 on Build GRCh38
rs201398869 Chr.19: 38409147-38409148 on Build GRCh38
rs765136129 Chr.19: 38409095-38409096 on Build GRCh38
rs763364452 Chr.19: 38409098-38409098 on Build GRCh38
rs1026136891 Chr.19: 38409128-38409128 on Build GRCh38
rs370595662 Chr.19: 38409142-38409142 on Build GRCh38
rs202085765 Chr.19: 38408899-38408899 on Build GRCh38
rs185123414 Chr.19: 38409143-38409143 on Build GRCh38
rs367871769 Chr.19: 38408802-38408802 on Build GRCh38
rs116620263 Chr.19: 38409196-38409196 on Build GRCh38
rs1019460506 Chr.19: 38408967-38408967 on Build GRCh38
rs180692206 Chr.19: 38408955-38408955 on Build GRCh38
rs761279628 Chr.19: 38408835-38408835 on Build GRCh38
rs571779060 Chr.19: 38409164-38409164 on Build GRCh38
rs539293874 Chr.19: 38408935-38408935 on Build GRCh38
rs760921841 Chr.19: 38408777-38408777 on Build GRCh38
rs1010969080 Chr.19: 38409053-38409053 on Build GRCh38
rs11451323 Chr.19: 38409142-38409143 on Build GRCh38
rs200637370 Chr.19: 38408859-38408859 on Build GRCh38
rs34348646 Chr.19: 38409143-38409144 on Build GRCh38
rs750236035 Chr.19: 38408885-38408885 on Build GRCh38
rs149162166 Chr.19: 38409016-38409016 on Build GRCh38
rs1022206505 Chr.19: 38409067-38409072 on Build GRCh38
rs577874404 Chr.19: 38409036-38409036 on Build GRCh38
rs555097755 Chr.19: 38408988-38408988 on Build GRCh38
rs1043383438 Chr.19: 38409194-38409194 on Build GRCh38
rs759864573 Chr.19: 38408917-38408917 on Build GRCh38
rs908408242 Chr.19: 38408870-38408870 on Build GRCh38
rs766792478 Chr.19: 38408780-38408780 on Build GRCh38
rs201683996 Chr.19: 38408877-38408877 on Build GRCh38
rs776996387 Chr.19: 38408842-38408842 on Build GRCh38
rs772121849 Chr.19: 38408904-38408905 on Build GRCh38
rs59917662 Chr.19: 38408879-38408880 on Build GRCh38
rs550126874 Chr.19: 38409150-38409150 on Build GRCh38
rs200551058 Chr.19: 38408852-38408852 on Build GRCh38
rs770193936 Chr.19: 38408922-38408922 on Build GRCh38
rs114144035 Chr.19: 38409201-38409201 on Build GRCh38
rs146021822 Chr.19: 38409057-38409057 on Build GRCh38
rs184620941 Chr.19: 38408819-38408819 on Build GRCh38
rs748459114 Chr.19: 38408862-38408864 on Build GRCh38
rs1034852633 Chr.19: 38409231-38409231 on Build GRCh38

View Full Product Details