Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38636800-38637279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3K
Entrez Gene ID
27335
Gene Name
eukaryotic translation initiation factor 3 subunit K
Gene Aliases
ARG134, EIF3-p28, EIF3S12, HSPC029, M9, MSTP001, PLAC-24, PLAC24, PRO1474, PTD001
UniGene
Hs.314359
Gene Chromosome Location
Chr.19: 38619072-38636959 on Build GRCh38
Transcripts
NM_001300992.1 NM_001308393.1 NM_013234.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7116109 c.639C>T p.I213I Chr.19: 38636902-38636902 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs567256537 Chr.19: 38636968-38636968 on Build GRCh38
rs930127899 Chr.19: 38637172-38637172 on Build GRCh38
rs970979489 Chr.19: 38636886-38636886 on Build GRCh38
rs747761830 Chr.19: 38636907-38636907 on Build GRCh38
rs564920371 Chr.19: 38637116-38637116 on Build GRCh38
rs113951699 Chr.19: 38636954-38636954 on Build GRCh38
rs1041180905 Chr.19: 38637030-38637030 on Build GRCh38
rs991287993 Chr.19: 38636958-38636958 on Build GRCh38
rs762887222 Chr.19: 38636986-38636987 on Build GRCh38
rs199747253 Chr.19: 38636883-38636883 on Build GRCh38
rs756698719 Chr.19: 38636979-38636979 on Build GRCh38
rs1022820462 Chr.19: 38637207-38637207 on Build GRCh38
rs994413897 Chr.19: 38637039-38637039 on Build GRCh38
rs972217156 Chr.19: 38637139-38637139 on Build GRCh38
rs543924492 Chr.19: 38637112-38637112 on Build GRCh38
rs1031855846 Chr.19: 38637239-38637239 on Build GRCh38
rs911636504 Chr.19: 38637137-38637137 on Build GRCh38
rs758322113 Chr.19: 38637116-38637118 on Build GRCh38
rs532409463 Chr.19: 38637119-38637119 on Build GRCh38
rs976450539 Chr.19: 38637252-38637252 on Build GRCh38
rs911618824 Chr.19: 38637019-38637019 on Build GRCh38
rs888663486 Chr.19: 38637214-38637214 on Build GRCh38
rs560017476 Chr.19: 38637162-38637162 on Build GRCh38
rs571294670 Chr.19: 38637046-38637046 on Build GRCh38
rs200802866 Chr.19: 38636914-38636914 on Build GRCh38
rs2080989 Chr.19: 38637042-38637042 on Build GRCh38
rs770505920 Chr.19: 38636850-38636850 on Build GRCh38
rs1003472175 Chr.19: 38636833-38636833 on Build GRCh38
rs953039446 Chr.19: 38637117-38637117 on Build GRCh38
rs772584889 Chr.19: 38637041-38637041 on Build GRCh38
rs941485012 Chr.19: 38637105-38637105 on Build GRCh38
rs758275280 Chr.19: 38636963-38636963 on Build GRCh38
rs759587776 Chr.19: 38636921-38636921 on Build GRCh38
rs555006986 Chr.19: 38637095-38637095 on Build GRCh38
rs572675467 Chr.19: 38637078-38637078 on Build GRCh38
rs544898278 Chr.19: 38637138-38637138 on Build GRCh38
rs751247438 Chr.19: 38636978-38636978 on Build GRCh38
rs753692671 Chr.19: 38636916-38636916 on Build GRCh38
rs548918557 Chr.19: 38637236-38637236 on Build GRCh38
rs1000003524 Chr.19: 38637224-38637224 on Build GRCh38
rs1045111869 Chr.19: 38637227-38637227 on Build GRCh38
rs985761477 Chr.19: 38637130-38637130 on Build GRCh38
rs567323920 Chr.19: 38637243-38637243 on Build GRCh38
rs924328964 Chr.19: 38637033-38637033 on Build GRCh38
rs765114656 Chr.19: 38636923-38636923 on Build GRCh38
rs1030865514 Chr.19: 38637100-38637100 on Build GRCh38
rs780795117 Chr.19: 38636988-38636988 on Build GRCh38
rs749848817 Chr.19: 38636868-38636868 on Build GRCh38
rs542857152 Chr.19: 38637079-38637079 on Build GRCh38
rs764377050 Chr.19: 38636994-38636994 on Build GRCh38
rs752484688 Chr.19: 38636930-38636930 on Build GRCh38
rs1049961049 Chr.19: 38637082-38637082 on Build GRCh38
rs780375651 Chr.19: 38636866-38636866 on Build GRCh38
rs895770012 Chr.19: 38637165-38637165 on Build GRCh38
rs554548680 Chr.19: 38637066-38637066 on Build GRCh38
rs113024296 Chr.19: 38636955-38636955 on Build GRCh38
rs923857012 Chr.19: 38636934-38636934 on Build GRCh38
rs11667512 Chr.19: 38636825-38636825 on Build GRCh38
rs766532978 Chr.19: 38636913-38636913 on Build GRCh38
rs539325381 Chr.19: 38637047-38637047 on Build GRCh38
rs774762055 Chr.19: 38636875-38636875 on Build GRCh38
rs200576883 Chr.19: 38636917-38636917 on Build GRCh38
rs768869837 Chr.19: 38636874-38636874 on Build GRCh38
rs771552520 Chr.19: 38637021-38637021 on Build GRCh38
rs1012780690 Chr.19: 38637204-38637204 on Build GRCh38
rs768525811 Chr.19: 38637147-38637147 on Build GRCh38
rs114289876 Chr.19: 38637108-38637108 on Build GRCh38
rs892029164 Chr.19: 38637086-38637086 on Build GRCh38
rs144992865 Chr.19: 38637036-38637037 on Build GRCh38
rs1048638803 Chr.19: 38637197-38637197 on Build GRCh38
rs537811114 Chr.19: 38636997-38636997 on Build GRCh38
rs2233003 Chr.19: 38636849-38636849 on Build GRCh38
rs527439170 Chr.19: 38637190-38637190 on Build GRCh38
rs772695653 Chr.19: 38637059-38637059 on Build GRCh38
rs201760106 Chr.19: 38636896-38636896 on Build GRCh38
rs748440372 Chr.19: 38636994-38636994 on Build GRCh38
rs769262177 Chr.19: 38636991-38636991 on Build GRCh38
rs968943816 Chr.19: 38637215-38637215 on Build GRCh38
rs968536282 Chr.19: 38637003-38637003 on Build GRCh38

View Full Product Details