Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38815721-38816215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECH1
Entrez Gene ID
1891
Gene Name
enoyl-CoA hydratase 1
Gene Aliases
HPXEL
UniGene
Hs.196176
Gene Chromosome Location
Chr.19: 38815422-38831857 on Build GRCh38
Transcripts
NM_001398.2 XM_017026448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6437038 c.732C>A p.S244R Chr.19: 38816007-38816007 on Build GRCh38
6376623 c.751G>T p.E251* Chr.19: 38815988-38815988 on Build GRCh38
5056614 c.777G>A p.A259A Chr.19: 38815962-38815962 on Build GRCh38
5427076 c.883-6T>C p.? Chr.19: 38815723-38815723 on Build GRCh38
4508717 c.785C>T p.A262V Chr.19: 38815954-38815954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144551354 Chr.19: 38815954-38815954 on Build GRCh38
rs780730678 Chr.19: 38815981-38815981 on Build GRCh38
rs552130567 Chr.19: 38815845-38815845 on Build GRCh38
rs752338357 Chr.19: 38815962-38815962 on Build GRCh38
rs933038881 Chr.19: 38815837-38815837 on Build GRCh38
rs770277115 Chr.19: 38815917-38815917 on Build GRCh38
rs770365506 Chr.19: 38815978-38815978 on Build GRCh38
rs139491123 Chr.19: 38815953-38815953 on Build GRCh38
rs771518931 Chr.19: 38815906-38815906 on Build GRCh38
rs768943807 Chr.19: 38815990-38815990 on Build GRCh38
rs777581948 Chr.19: 38816047-38816047 on Build GRCh38
rs920094266 Chr.19: 38816068-38816068 on Build GRCh38
rs375332852 Chr.19: 38815892-38815892 on Build GRCh38
rs552290076 Chr.19: 38816085-38816085 on Build GRCh38
rs374648175 Chr.19: 38815875-38815875 on Build GRCh38
rs767633667 Chr.19: 38815945-38815945 on Build GRCh38
rs746522011 Chr.19: 38815969-38815969 on Build GRCh38
rs750349960 Chr.19: 38815946-38815946 on Build GRCh38
rs773213223 Chr.19: 38816010-38816010 on Build GRCh38
rs777560195 Chr.19: 38815839-38815840 on Build GRCh38
rs192745108 Chr.19: 38815835-38815835 on Build GRCh38
rs113851915 Chr.19: 38816067-38816067 on Build GRCh38
rs748890648 Chr.19: 38815890-38815890 on Build GRCh38
rs773981477 Chr.19: 38815831-38815831 on Build GRCh38
rs528510594 Chr.19: 38816188-38816188 on Build GRCh38
rs552518184 Chr.19: 38815807-38815807 on Build GRCh38
rs370357377 Chr.19: 38815840-38815840 on Build GRCh38
rs762001871 Chr.19: 38816005-38816005 on Build GRCh38
rs954511326 Chr.19: 38816017-38816017 on Build GRCh38
rs58750431 Chr.19: 38815889-38815889 on Build GRCh38
rs921717271 Chr.19: 38816013-38816013 on Build GRCh38
rs763240178 Chr.19: 38815926-38815926 on Build GRCh38
rs373602260 Chr.19: 38815933-38815933 on Build GRCh38
rs748993959 Chr.19: 38815820-38815820 on Build GRCh38
rs149531549 Chr.19: 38815897-38815897 on Build GRCh38
rs755934996 Chr.19: 38815814-38815814 on Build GRCh38
rs150833592 Chr.19: 38815931-38815931 on Build GRCh38
rs755884220 Chr.19: 38815876-38815876 on Build GRCh38
rs538528426 Chr.19: 38815860-38815860 on Build GRCh38
rs779650677 Chr.19: 38815817-38815817 on Build GRCh38
rs751711165 Chr.19: 38815988-38815989 on Build GRCh38
rs764291702 Chr.19: 38815748-38815748 on Build GRCh38
rs199700435 Chr.19: 38815898-38815898 on Build GRCh38
rs2075587 Chr.19: 38816163-38816163 on Build GRCh38
rs753754369 Chr.19: 38816028-38816028 on Build GRCh38
rs200226678 Chr.19: 38815957-38815957 on Build GRCh38
rs2075588 Chr.19: 38815785-38815785 on Build GRCh38
rs765745607 Chr.19: 38815847-38815847 on Build GRCh38
rs374422461 Chr.19: 38815841-38815841 on Build GRCh38
rs569568697 Chr.19: 38815745-38815745 on Build GRCh38
rs767402572 Chr.19: 38815874-38815874 on Build GRCh38
rs565533232 Chr.19: 38815872-38815872 on Build GRCh38
rs907796158 Chr.19: 38816015-38816015 on Build GRCh38
rs775841218 Chr.19: 38815850-38815850 on Build GRCh38
rs199702625 Chr.19: 38816025-38816025 on Build GRCh38
rs530366183 Chr.19: 38815783-38815783 on Build GRCh38
rs530927065 Chr.19: 38815791-38815791 on Build GRCh38
rs781007249 Chr.19: 38815766-38815766 on Build GRCh38
rs974345677 Chr.19: 38815797-38815797 on Build GRCh38
rs1064972 Chr.19: 38815927-38815927 on Build GRCh38
rs768040303 Chr.19: 38815822-38815822 on Build GRCh38
rs758054951 Chr.19: 38816046-38816046 on Build GRCh38
rs146638554 Chr.19: 38816003-38816003 on Build GRCh38
rs775929230 Chr.19: 38815925-38815925 on Build GRCh38
rs758384942 Chr.19: 38815747-38815747 on Build GRCh38
rs764996966 Chr.19: 38816037-38816037 on Build GRCh38
rs751410665 Chr.19: 38816055-38816055 on Build GRCh38
rs1021592001 Chr.19: 38816168-38816168 on Build GRCh38
rs751445868 Chr.19: 38815751-38815751 on Build GRCh38
rs939980631 Chr.19: 38816021-38816021 on Build GRCh38
rs200148431 Chr.19: 38816034-38816034 on Build GRCh38
rs374473358 Chr.19: 38815744-38815744 on Build GRCh38
rs752387975 Chr.19: 38816039-38816039 on Build GRCh38
rs1033886239 Chr.19: 38815846-38815846 on Build GRCh38
rs777490260 Chr.19: 38815964-38815964 on Build GRCh38
rs751064177 Chr.19: 38816048-38816048 on Build GRCh38
rs778594450 Chr.19: 38815960-38815960 on Build GRCh38
rs749625289 Chr.19: 38815986-38815986 on Build GRCh38
rs772137811 Chr.19: 38816006-38816006 on Build GRCh38
rs931509721 Chr.19: 38816108-38816108 on Build GRCh38
rs891404950 Chr.19: 38815858-38815858 on Build GRCh38
rs369378100 Chr.19: 38815911-38815911 on Build GRCh38
rs763300655 Chr.19: 38815851-38815851 on Build GRCh38
rs558563893 Chr.19: 38815916-38815916 on Build GRCh38
rs760760219 Chr.19: 38816018-38816018 on Build GRCh38
rs887132626 Chr.19: 38815784-38815784 on Build GRCh38
rs559979174 Chr.19: 38815773-38815773 on Build GRCh38
rs148749158 Chr.19: 38815869-38815869 on Build GRCh38
rs756859563 Chr.19: 38816051-38816051 on Build GRCh38
rs761923532 Chr.19: 38815944-38815944 on Build GRCh38
rs966989702 Chr.19: 38816180-38816180 on Build GRCh38

View Full Product Details