Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38831621-38832159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECH1
Entrez Gene ID
1891
Gene Name
enoyl-CoA hydratase 1
Gene Aliases
HPXEL
UniGene
Hs.196176
Gene Chromosome Location
Chr.19: 38815422-38831857 on Build GRCh38
Transcripts
NM_001398.2 XM_017026448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4636653 c.29G>T p.R10I Chr.19: 38831744-38831744 on Build GRCh38
4140613 c.40C>G p.L14V Chr.19: 38831733-38831733 on Build GRCh38
5183153 c.51G>A p.R17R Chr.19: 38831722-38831722 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs965581369 Chr.19: 38831689-38831689 on Build GRCh38
rs367950248 Chr.19: 38831692-38831692 on Build GRCh38
rs774274035 Chr.19: 38831807-38831807 on Build GRCh38
rs952891463 Chr.19: 38832098-38832098 on Build GRCh38
rs903175354 Chr.19: 38832023-38832023 on Build GRCh38
rs908184930 Chr.19: 38831936-38831936 on Build GRCh38
rs1015838662 Chr.19: 38832116-38832116 on Build GRCh38
rs75206555 Chr.19: 38832121-38832121 on Build GRCh38
rs948999679 Chr.19: 38832129-38832133 on Build GRCh38
rs1032291339 Chr.19: 38831788-38831788 on Build GRCh38
rs190290587 Chr.19: 38831799-38831799 on Build GRCh38
rs957474099 Chr.19: 38831798-38831798 on Build GRCh38
rs984329494 Chr.19: 38832107-38832107 on Build GRCh38
rs78254559 Chr.19: 38831885-38831885 on Build GRCh38
rs772752969 Chr.19: 38831738-38831738 on Build GRCh38
rs576691278 Chr.19: 38831927-38831927 on Build GRCh38
rs934947828 Chr.19: 38831945-38831945 on Build GRCh38
rs867145902 Chr.19: 38831793-38831793 on Build GRCh38
rs748563864 Chr.19: 38831762-38831763 on Build GRCh38
rs199739067 Chr.19: 38832124-38832125 on Build GRCh38
rs1053273521 Chr.19: 38831950-38831951 on Build GRCh38
rs773792171 Chr.19: 38831718-38831719 on Build GRCh38
rs766029567 Chr.19: 38831771-38831771 on Build GRCh38
rs753489805 Chr.19: 38831822-38831822 on Build GRCh38
rs893661194 Chr.19: 38831960-38831960 on Build GRCh38
rs775483003 Chr.19: 38831751-38831751 on Build GRCh38
rs1020106994 Chr.19: 38832049-38832049 on Build GRCh38
rs949049871 Chr.19: 38831905-38831905 on Build GRCh38
rs759679528 Chr.19: 38831656-38831656 on Build GRCh38
rs778441499 Chr.19: 38831781-38831781 on Build GRCh38
rs199707116 Chr.19: 38831819-38831819 on Build GRCh38
rs769717958 Chr.19: 38832120-38832120 on Build GRCh38
rs778390273 Chr.19: 38831744-38831744 on Build GRCh38
rs1028846350 Chr.19: 38831979-38831979 on Build GRCh38
rs372144332 Chr.19: 38831765-38831765 on Build GRCh38
rs997360071 Chr.19: 38832067-38832067 on Build GRCh38
rs1042333466 Chr.19: 38832017-38832017 on Build GRCh38
rs1028409952 Chr.19: 38832079-38832079 on Build GRCh38
rs559422929 Chr.19: 38832057-38832057 on Build GRCh38
rs764050776 Chr.19: 38831753-38831753 on Build GRCh38
rs746222157 Chr.19: 38831789-38831789 on Build GRCh38
rs762991921 Chr.19: 38831752-38831752 on Build GRCh38
rs1037151799 Chr.19: 38831685-38831685 on Build GRCh38
rs965913221 Chr.19: 38831878-38831878 on Build GRCh38
rs1019738016 Chr.19: 38831870-38831870 on Build GRCh38
rs373332671 Chr.19: 38831818-38831818 on Build GRCh38
rs893756106 Chr.19: 38831826-38831826 on Build GRCh38
rs374275115 Chr.19: 38831797-38831797 on Build GRCh38
rs3760656 Chr.19: 38832126-38832126 on Build GRCh38
rs35295397 Chr.19: 38832122-38832122 on Build GRCh38
rs749393482 Chr.19: 38831802-38831802 on Build GRCh38
rs375702558 Chr.19: 38831831-38831831 on Build GRCh38
rs981770729 Chr.19: 38832130-38832133 on Build GRCh38
rs879153826 Chr.19: 38831771-38831772 on Build GRCh38
rs984340905 Chr.19: 38831900-38831900 on Build GRCh38
rs531370913 Chr.19: 38831757-38831757 on Build GRCh38
rs1019604730 Chr.19: 38832106-38832108 on Build GRCh38
rs775743829 Chr.19: 38831795-38831795 on Build GRCh38
rs537221165 Chr.19: 38831940-38831940 on Build GRCh38
rs111265972 Chr.19: 38831718-38831718 on Build GRCh38
rs541836756 Chr.19: 38832122-38832122 on Build GRCh38
rs767148522 Chr.19: 38831763-38831763 on Build GRCh38
rs761804703 Chr.19: 38831808-38831808 on Build GRCh38
rs866435092 Chr.19: 38832118-38832118 on Build GRCh38
rs768664398 Chr.19: 38831804-38831804 on Build GRCh38
rs398034564 Chr.19: 38832132-38832132 on Build GRCh38
rs762087352 Chr.19: 38831955-38831955 on Build GRCh38
rs73930207 Chr.19: 38831816-38831816 on Build GRCh38
rs747455186 Chr.19: 38831785-38831785 on Build GRCh38
rs771368717 Chr.19: 38831747-38831747 on Build GRCh38
rs866858145 Chr.19: 38831695-38831695 on Build GRCh38
rs927758592 Chr.19: 38831952-38831952 on Build GRCh38
rs577251797 Chr.19: 38831980-38831980 on Build GRCh38
rs376800152 Chr.19: 38831786-38831786 on Build GRCh38
rs977520191 Chr.19: 38831648-38831648 on Build GRCh38
rs1054937102 Chr.19: 38831958-38831958 on Build GRCh38
rs761610089 Chr.19: 38831758-38831758 on Build GRCh38
rs866522793 Chr.19: 38832132-38832132 on Build GRCh38
rs967372607 Chr.19: 38831858-38831858 on Build GRCh38
rs999309298 Chr.19: 38831869-38831869 on Build GRCh38
rs35074952 Chr.19: 38831750-38831750 on Build GRCh38
rs939724157 Chr.19: 38831938-38831938 on Build GRCh38
rs748815662 Chr.19: 38831736-38831736 on Build GRCh38
rs11549276 Chr.19: 38831729-38831729 on Build GRCh38
rs541169332 Chr.19: 38831981-38831981 on Build GRCh38
rs776920494 Chr.19: 38831748-38831748 on Build GRCh38
rs767475221 Chr.19: 38831812-38831812 on Build GRCh38
rs765166644 Chr.19: 38831660-38831660 on Build GRCh38
rs763999495 Chr.19: 38831671-38831671 on Build GRCh38
rs1021610746 Chr.19: 38831842-38831842 on Build GRCh38
rs916219916 Chr.19: 38831902-38831903 on Build GRCh38
rs754494335 Chr.19: 38831777-38831777 on Build GRCh38
rs111656875 Chr.19: 38831719-38831719 on Build GRCh38
rs527572284 Chr.19: 38831662-38831662 on Build GRCh38
rs182370972 Chr.19: 38831895-38831895 on Build GRCh38
rs769950628 Chr.19: 38831792-38831792 on Build GRCh38
rs755776416 Chr.19: 38831767-38831767 on Build GRCh38
rs368453801 Chr.19: 38831724-38831724 on Build GRCh38
rs921447341 Chr.19: 38831882-38831882 on Build GRCh38
rs78637797 Chr.19: 38831712-38831712 on Build GRCh38
rs1011634432 Chr.19: 38831969-38831969 on Build GRCh38
rs560996634 Chr.19: 38832125-38832125 on Build GRCh38
rs755652897 Chr.19: 38831723-38831723 on Build GRCh38
rs117820732 Chr.19: 38831774-38831774 on Build GRCh38
rs747405167 Chr.19: 38831745-38831745 on Build GRCh38
rs1045975859 Chr.19: 38831934-38831934 on Build GRCh38
rs574449902 Chr.19: 38832108-38832108 on Build GRCh38
rs998892958 Chr.19: 38832034-38832034 on Build GRCh38
rs975638321 Chr.19: 38831880-38831880 on Build GRCh38
rs931087608 Chr.19: 38831702-38831702 on Build GRCh38
rs74427873 Chr.19: 38831953-38831953 on Build GRCh38
rs745807837 Chr.19: 38832118-38832122 on Build GRCh38
rs548817124 Chr.19: 38831708-38831708 on Build GRCh38
rs369103554 Chr.19: 38832125-38832126 on Build GRCh38
rs375573622 Chr.19: 38831683-38831683 on Build GRCh38
rs767140449 Chr.19: 38832110-38832113 on Build GRCh38
rs967424960 Chr.19: 38831639-38831639 on Build GRCh38

View Full Product Details