Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39328103-39328599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMFG
Entrez Gene ID
9535
Gene Name
glia maturation factor gamma
Gene Aliases
GMF-GAMMA
UniGene
Hs.5210
Gene Chromosome Location
Chr.19: 39328357-39336086 on Build GRCh38
Transcripts
NM_001301008.1 NM_004877.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4581012 c.393G>A p.E131E Chr.19: 39328513-39328513 on Build GRCh38
7184167 c.424C>T p.R142C Chr.19: 39328482-39328482 on Build GRCh38
6008472 c.371G>A p.R124H Chr.19: 39328535-39328535 on Build GRCh38
4405612 c.408A>T p.E136D Chr.19: 39328498-39328498 on Build GRCh38
1612222 c.397T>A p.W133R Chr.19: 39328509-39328509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369865986 Chr.19: 39328558-39328558 on Build GRCh38
rs752561944 Chr.19: 39328477-39328477 on Build GRCh38
rs145410023 Chr.19: 39328510-39328510 on Build GRCh38
rs988008236 Chr.19: 39328181-39328181 on Build GRCh38
rs761458133 Chr.19: 39328568-39328568 on Build GRCh38
rs557036197 Chr.19: 39328365-39328365 on Build GRCh38
rs538449741 Chr.19: 39328351-39328351 on Build GRCh38
rs112724573 Chr.19: 39328513-39328513 on Build GRCh38
rs751261073 Chr.19: 39328484-39328484 on Build GRCh38
rs377738809 Chr.19: 39328454-39328454 on Build GRCh38
rs896674986 Chr.19: 39328277-39328277 on Build GRCh38
rs992901323 Chr.19: 39328377-39328377 on Build GRCh38
rs941627302 Chr.19: 39328161-39328161 on Build GRCh38
rs917375581 Chr.19: 39328411-39328411 on Build GRCh38
rs919925319 Chr.19: 39328546-39328546 on Build GRCh38
rs1038818295 Chr.19: 39328255-39328255 on Build GRCh38
rs765230794 Chr.19: 39328472-39328472 on Build GRCh38
rs751027176 Chr.19: 39328565-39328565 on Build GRCh38
rs759481928 Chr.19: 39328466-39328466 on Build GRCh38
rs768387432 Chr.19: 39328405-39328405 on Build GRCh38
rs780659869 Chr.19: 39328508-39328508 on Build GRCh38
rs374823721 Chr.19: 39328180-39328180 on Build GRCh38
rs969567364 Chr.19: 39328352-39328352 on Build GRCh38
rs34035414 Chr.19: 39328500-39328500 on Build GRCh38
rs143817028 Chr.19: 39328528-39328528 on Build GRCh38
rs994429847 Chr.19: 39328273-39328273 on Build GRCh38
rs536796444 Chr.19: 39328415-39328415 on Build GRCh38
rs933114556 Chr.19: 39328236-39328236 on Build GRCh38
rs775461658 Chr.19: 39328556-39328556 on Build GRCh38
rs145903044 Chr.19: 39328166-39328166 on Build GRCh38
rs956873209 Chr.19: 39328177-39328177 on Build GRCh38
rs776799285 Chr.19: 39328465-39328465 on Build GRCh38
rs1047306400 Chr.19: 39328274-39328274 on Build GRCh38
rs1051139605 Chr.19: 39328240-39328240 on Build GRCh38
rs779632252 Chr.19: 39328516-39328516 on Build GRCh38
rs564550103 Chr.19: 39328433-39328433 on Build GRCh38
rs772616377 Chr.19: 39328518-39328518 on Build GRCh38
rs776655324 Chr.19: 39328543-39328543 on Build GRCh38
rs769857010 Chr.19: 39328552-39328552 on Build GRCh38
rs149972285 Chr.19: 39328481-39328481 on Build GRCh38
rs898489129 Chr.19: 39328257-39328257 on Build GRCh38
rs1032455151 Chr.19: 39328387-39328387 on Build GRCh38
rs547964647 Chr.19: 39328325-39328325 on Build GRCh38
rs570929462 Chr.19: 39328150-39328150 on Build GRCh38
rs575485840 Chr.19: 39328434-39328434 on Build GRCh38
rs1000131960 Chr.19: 39328320-39328320 on Build GRCh38
rs890279388 Chr.19: 39328233-39328233 on Build GRCh38
rs551971728 Chr.19: 39328453-39328453 on Build GRCh38
rs759663223 Chr.19: 39328550-39328550 on Build GRCh38
rs748653985 Chr.19: 39328517-39328517 on Build GRCh38
rs942783860 Chr.19: 39328254-39328254 on Build GRCh38
rs993634537 Chr.19: 39328184-39328184 on Build GRCh38
rs756902069 Chr.19: 39328503-39328503 on Build GRCh38
rs746393225 Chr.19: 39328555-39328556 on Build GRCh38
rs781779937 Chr.19: 39328456-39328457 on Build GRCh38
rs36110047 Chr.19: 39328542-39328542 on Build GRCh38
rs758041464 Chr.19: 39328199-39328199 on Build GRCh38
rs911325804 Chr.19: 39328242-39328242 on Build GRCh38
rs763915912 Chr.19: 39328563-39328563 on Build GRCh38
rs751377742 Chr.19: 39328567-39328567 on Build GRCh38
rs1013732560 Chr.19: 39328309-39328309 on Build GRCh38
rs942815841 Chr.19: 39328537-39328537 on Build GRCh38
rs371601820 Chr.19: 39328536-39328536 on Build GRCh38
rs569740740 Chr.19: 39328373-39328373 on Build GRCh38

View Full Product Details