Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39502682-39503175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLL3
Entrez Gene ID
10683
Gene Name
delta like canonical Notch ligand 3
Gene Aliases
SCDO1
UniGene
Hs.127792
Gene Chromosome Location
Chr.19: 39498917-39508481 on Build GRCh38
Transcripts
NM_016941.3 NM_203486.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
398742 c.580C>A p.R194S Chr.19: 39502985-39502985 on Build GRCh38
6391494 c.448C>G p.R150G Chr.19: 39502853-39502853 on Build GRCh38
6539175 c.593C>G p.S198W Chr.19: 39502998-39502998 on Build GRCh38
4987115 c.425T>A p.L142Q Chr.19: 39502830-39502830 on Build GRCh38
5657107 c.467G>T p.G156V Chr.19: 39502872-39502872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541279295 Chr.19: 39502776-39502776 on Build GRCh38
rs962122621 Chr.19: 39502922-39502922 on Build GRCh38
rs772960131 Chr.19: 39503028-39503028 on Build GRCh38
rs1055435030 Chr.19: 39502748-39502748 on Build GRCh38
rs768310008 Chr.19: 39502857-39502857 on Build GRCh38
rs925186060 Chr.19: 39502858-39502858 on Build GRCh38
rs964140589 Chr.19: 39503081-39503081 on Build GRCh38
rs774107458 Chr.19: 39502861-39502861 on Build GRCh38
rs1045208902 Chr.19: 39503036-39503036 on Build GRCh38
rs762121920 Chr.19: 39503097-39503097 on Build GRCh38
rs771875570 Chr.19: 39503026-39503026 on Build GRCh38
rs746558629 Chr.19: 39503029-39503029 on Build GRCh38
rs764416429 Chr.19: 39502783-39502783 on Build GRCh38
rs376984786 Chr.19: 39503142-39503142 on Build GRCh38
rs765614169 Chr.19: 39502718-39502718 on Build GRCh38
rs758654270 Chr.19: 39502781-39502781 on Build GRCh38
rs1007844764 Chr.19: 39503073-39503073 on Build GRCh38
rs1030526921 Chr.19: 39503065-39503065 on Build GRCh38
rs2278440 Chr.19: 39502775-39502775 on Build GRCh38
rs764597918 Chr.19: 39503092-39503092 on Build GRCh38
rs786200899 Chr.19: 39503008-39503009 on Build GRCh38
rs8106337 Chr.19: 39502951-39502951 on Build GRCh38
rs8107127 Chr.19: 39502920-39502920 on Build GRCh38
rs774717100 Chr.19: 39503095-39503095 on Build GRCh38
rs756066182 Chr.19: 39502828-39502828 on Build GRCh38
rs953389900 Chr.19: 39502914-39502914 on Build GRCh38
rs1052592318 Chr.19: 39503022-39503022 on Build GRCh38
rs775830225 Chr.19: 39502931-39502931 on Build GRCh38
rs1042783044 Chr.19: 39503006-39503006 on Build GRCh38
rs942513166 Chr.19: 39502853-39502853 on Build GRCh38
rs753223980 Chr.19: 39502722-39502722 on Build GRCh38
rs765809448 Chr.19: 39502927-39502927 on Build GRCh38
rs760217365 Chr.19: 39502923-39502923 on Build GRCh38
rs756151698 Chr.19: 39502999-39502999 on Build GRCh38
rs1036646915 Chr.19: 39502720-39502720 on Build GRCh38
rs749148035 Chr.19: 39502847-39502847 on Build GRCh38
rs1017969999 Chr.19: 39503015-39503015 on Build GRCh38
rs192624990 Chr.19: 39503023-39503023 on Build GRCh38
rs1017898599 Chr.19: 39503076-39503076 on Build GRCh38
rs894164280 Chr.19: 39502749-39502749 on Build GRCh38
rs949405375 Chr.19: 39503147-39503147 on Build GRCh38
rs376905666 Chr.19: 39503021-39503021 on Build GRCh38
rs886054432 Chr.19: 39503025-39503025 on Build GRCh38
rs780088167 Chr.19: 39503020-39503020 on Build GRCh38
rs903880731 Chr.19: 39502716-39502716 on Build GRCh38
rs770329421 Chr.19: 39503042-39503042 on Build GRCh38
rs751710148 Chr.19: 39502791-39502791 on Build GRCh38
rs188178519 Chr.19: 39502730-39502730 on Build GRCh38
rs960151733 Chr.19: 39503093-39503093 on Build GRCh38
rs896576849 Chr.19: 39502729-39502729 on Build GRCh38
rs962071242 Chr.19: 39503058-39503058 on Build GRCh38
rs750529359 Chr.19: 39502970-39502970 on Build GRCh38
rs1011290528 Chr.19: 39502753-39502753 on Build GRCh38
rs984792348 Chr.19: 39502918-39502918 on Build GRCh38
rs935221144 Chr.19: 39502876-39502876 on Build GRCh38
rs1031616535 Chr.19: 39502849-39502849 on Build GRCh38
rs1028956837 Chr.19: 39502875-39502875 on Build GRCh38
rs557730459 Chr.19: 39502984-39502984 on Build GRCh38
rs956549712 Chr.19: 39503099-39503099 on Build GRCh38
rs771785908 Chr.19: 39502898-39502898 on Build GRCh38
rs767676342 Chr.19: 39502969-39502969 on Build GRCh38
rs757481580 Chr.19: 39502957-39502957 on Build GRCh38
rs115194365 Chr.19: 39502948-39502948 on Build GRCh38
rs942088440 Chr.19: 39503044-39503045 on Build GRCh38
rs986488775 Chr.19: 39502843-39502843 on Build GRCh38
rs1023823678 Chr.19: 39502747-39502747 on Build GRCh38
rs1038288359 Chr.19: 39503045-39503045 on Build GRCh38
rs763424873 Chr.19: 39502947-39502947 on Build GRCh38
rs1013696397 Chr.19: 39502936-39502936 on Build GRCh38
rs111827161 Chr.19: 39503090-39503090 on Build GRCh38
rs867838103 Chr.19: 39502896-39502896 on Build GRCh38
rs776126232 Chr.19: 39503054-39503054 on Build GRCh38
rs528327447 Chr.19: 39503072-39503072 on Build GRCh38
rs997746180 Chr.19: 39503025-39503025 on Build GRCh38
rs55741253 Chr.19: 39502830-39502830 on Build GRCh38
rs786200902 Chr.19: 39503023-39503023 on Build GRCh38
rs947027933 Chr.19: 39502986-39502986 on Build GRCh38
rs991371734 Chr.19: 39503117-39503117 on Build GRCh38
rs911077015 Chr.19: 39502852-39502852 on Build GRCh38
rs962616128 Chr.19: 39502960-39502960 on Build GRCh38
rs1045157695 Chr.19: 39502943-39502943 on Build GRCh38
rs867974498 Chr.19: 39502956-39502956 on Build GRCh38
rs141944861 Chr.19: 39503128-39503128 on Build GRCh38
rs938625000 Chr.19: 39502745-39502745 on Build GRCh38
rs572869684 Chr.19: 39503039-39503039 on Build GRCh38
rs376907733 Chr.19: 39502883-39502883 on Build GRCh38
rs757387396 Chr.19: 39502794-39502794 on Build GRCh38
rs781246209 Chr.19: 39502820-39502820 on Build GRCh38

View Full Product Details