Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46513410-46513938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMBX1
Entrez Gene ID
127343
Gene Name
diencephalon/mesencephalon homeobox 1
Gene Aliases
MBX, OTX3, PAXB
UniGene
Hs.375623
Gene Chromosome Location
Chr.1: 46489837-46514226 on Build GRCh38
Transcripts
NM_147192.2 NM_172225.1 XM_011540668.2 XM_017000289.1

SNP Detail

SNP ID Chromosome Location
rs1030256301 Chr.1: 46513711-46513711 on Build GRCh38
rs576849322 Chr.1: 46513771-46513771 on Build GRCh38
rs1013161623 Chr.1: 46513724-46513724 on Build GRCh38
rs776128804 Chr.1: 46513514-46513514 on Build GRCh38
rs59809955 Chr.1: 46513882-46513882 on Build GRCh38
rs752275100 Chr.1: 46513633-46513633 on Build GRCh38
rs914880901 Chr.1: 46513875-46513875 on Build GRCh38
rs544601694 Chr.1: 46513601-46513601 on Build GRCh38
rs927813345 Chr.1: 46513892-46513892 on Build GRCh38
rs754494826 Chr.1: 46513694-46513694 on Build GRCh38
rs955543929 Chr.1: 46513721-46513721 on Build GRCh38
rs1051434530 Chr.1: 46513548-46513548 on Build GRCh38
rs764590910 Chr.1: 46513695-46513695 on Build GRCh38
rs987208601 Chr.1: 46513791-46513791 on Build GRCh38
rs1035765053 Chr.1: 46513706-46513706 on Build GRCh38
rs759277007 Chr.1: 46513519-46513519 on Build GRCh38
rs140564592 Chr.1: 46513792-46513792 on Build GRCh38
rs375714215 Chr.1: 46513691-46513691 on Build GRCh38
rs912742648 Chr.1: 46513549-46513549 on Build GRCh38
rs977847751 Chr.1: 46513888-46513888 on Build GRCh38
rs182212130 Chr.1: 46513699-46513699 on Build GRCh38
rs1016215799 Chr.1: 46513725-46513725 on Build GRCh38
rs1040286354 Chr.1: 46513846-46513846 on Build GRCh38
rs551463197 Chr.1: 46513512-46513512 on Build GRCh38
rs927792773 Chr.1: 46513526-46513526 on Build GRCh38
rs375433020 Chr.1: 46513817-46513817 on Build GRCh38
rs963632034 Chr.1: 46513770-46513770 on Build GRCh38
rs117377891 Chr.1: 46513582-46513582 on Build GRCh38
rs770036689 Chr.1: 46513812-46513812 on Build GRCh38
rs7515750 Chr.1: 46513671-46513671 on Build GRCh38
rs945108525 Chr.1: 46513481-46513481 on Build GRCh38
rs989150293 Chr.1: 46513855-46513855 on Build GRCh38
rs1038454834 Chr.1: 46513678-46513678 on Build GRCh38
rs964414998 Chr.1: 46513841-46513841 on Build GRCh38
rs1011799796 Chr.1: 46513618-46513618 on Build GRCh38
rs4660355 Chr.1: 46513550-46513550 on Build GRCh38
rs1039996733 Chr.1: 46513559-46513559 on Build GRCh38

View Full Product Details