Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46652339-46652855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATPAF1
Entrez Gene ID
64756
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Gene Aliases
ATP11, ATP11p
UniGene
Hs.100874
Gene Chromosome Location
Chr.1: 46635039-46668427 on Build GRCh38
Transcripts
NM_001042546.2 NM_001243728.1 NM_001256418.1 NM_022745.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3977706 c.571G>A p.A191T Chr.1: 46652598-46652598 on Build GRCh38
910350 c.544G>A p.E182K Chr.1: 46652625-46652625 on Build GRCh38
5132453 c.568C>T p.R190W Chr.1: 46652601-46652601 on Build GRCh38
6322008 c.568C>A p.R190R Chr.1: 46652601-46652601 on Build GRCh38
1660703 c.574C>A p.Q192K Chr.1: 46652595-46652595 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs577170555 Chr.1: 46652436-46652436 on Build GRCh38
rs560359383 Chr.1: 46652687-46652687 on Build GRCh38
rs925255113 Chr.1: 46652601-46652601 on Build GRCh38
rs375071560 Chr.1: 46652616-46652616 on Build GRCh38
rs772328893 Chr.1: 46652663-46652663 on Build GRCh38
rs751215386 Chr.1: 46652568-46652568 on Build GRCh38
rs371007611 Chr.1: 46652614-46652614 on Build GRCh38
rs367715157 Chr.1: 46652669-46652669 on Build GRCh38
rs773607383 Chr.1: 46652552-46652552 on Build GRCh38
rs762043362 Chr.1: 46652533-46652533 on Build GRCh38
rs778934835 Chr.1: 46652639-46652639 on Build GRCh38
rs961894321 Chr.1: 46652541-46652541 on Build GRCh38
rs1045478273 Chr.1: 46652431-46652431 on Build GRCh38
rs553431085 Chr.1: 46652438-46652438 on Build GRCh38
rs540419604 Chr.1: 46652414-46652414 on Build GRCh38
rs775981236 Chr.1: 46652598-46652598 on Build GRCh38
rs762496708 Chr.1: 46652562-46652562 on Build GRCh38
rs1001302187 Chr.1: 46652457-46652457 on Build GRCh38
rs1052646959 Chr.1: 46652379-46652379 on Build GRCh38
rs906976024 Chr.1: 46652456-46652456 on Build GRCh38
rs771214515 Chr.1: 46652674-46652674 on Build GRCh38
rs2289446 Chr.1: 46652424-46652424 on Build GRCh38
rs1006792678 Chr.1: 46652768-46652768 on Build GRCh38
rs766908930 Chr.1: 46652558-46652558 on Build GRCh38
rs373775435 Chr.1: 46652569-46652569 on Build GRCh38
rs374585795 Chr.1: 46652664-46652664 on Build GRCh38
rs748278917 Chr.1: 46652644-46652644 on Build GRCh38
rs2289447 Chr.1: 46652496-46652496 on Build GRCh38
rs937039454 Chr.1: 46652789-46652789 on Build GRCh38
rs905372719 Chr.1: 46652772-46652772 on Build GRCh38
rs945735501 Chr.1: 46652375-46652375 on Build GRCh38
rs1033986031 Chr.1: 46652491-46652491 on Build GRCh38
rs185666554 Chr.1: 46652561-46652561 on Build GRCh38
rs957242367 Chr.1: 46652835-46652835 on Build GRCh38
rs1006103067 Chr.1: 46652501-46652501 on Build GRCh38
rs953442302 Chr.1: 46652567-46652567 on Build GRCh38
rs374179672 Chr.1: 46652770-46652770 on Build GRCh38
rs1005635258 Chr.1: 46652517-46652517 on Build GRCh38
rs620913 Chr.1: 46652395-46652395 on Build GRCh38
rs777482999 Chr.1: 46652631-46652633 on Build GRCh38
rs976348821 Chr.1: 46652557-46652557 on Build GRCh38
rs370174546 Chr.1: 46652544-46652544 on Build GRCh38
rs955301811 Chr.1: 46652751-46652751 on Build GRCh38
rs620431 Chr.1: 46652517-46652517 on Build GRCh38
rs112067834 Chr.1: 46652570-46652570 on Build GRCh38
rs780755963 Chr.1: 46652589-46652589 on Build GRCh38
rs528162036 Chr.1: 46652692-46652692 on Build GRCh38
rs914173184 Chr.1: 46652631-46652631 on Build GRCh38
rs546389417 Chr.1: 46652728-46652728 on Build GRCh38
rs761006367 Chr.1: 46652553-46652553 on Build GRCh38
rs763722095 Chr.1: 46652563-46652563 on Build GRCh38
rs755925763 Chr.1: 46652600-46652600 on Build GRCh38
rs768670861 Chr.1: 46652635-46652635 on Build GRCh38
rs201092265 Chr.1: 46652607-46652607 on Build GRCh38
rs368299807 Chr.1: 46652628-46652628 on Build GRCh38
rs906802875 Chr.1: 46652409-46652409 on Build GRCh38

View Full Product Details