Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46653736-46654227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATPAF1
Entrez Gene ID
64756
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Gene Aliases
ATP11, ATP11p
UniGene
Hs.100874
Gene Chromosome Location
Chr.1: 46635039-46668427 on Build GRCh38
Transcripts
NM_001042546.2 NM_001243728.1 NM_001256418.1 NM_022745.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5381141 c.538C>T p.P180S Chr.1: 46653819-46653819 on Build GRCh38
1748484 c.498G>A p.Q166Q Chr.1: 46653859-46653859 on Build GRCh38
1474045 c.525C>T p.V175V Chr.1: 46653832-46653832 on Build GRCh38
4664595 c.511G>A p.A171T Chr.1: 46653846-46653846 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768816016 Chr.1: 46654082-46654082 on Build GRCh38
rs893171051 Chr.1: 46654134-46654134 on Build GRCh38
rs1003130151 Chr.1: 46654154-46654156 on Build GRCh38
rs1017080987 Chr.1: 46654145-46654145 on Build GRCh38
rs910237001 Chr.1: 46654165-46654165 on Build GRCh38
rs761286529 Chr.1: 46653809-46653809 on Build GRCh38
rs918880949 Chr.1: 46653782-46653782 on Build GRCh38
rs1011935909 Chr.1: 46653768-46653768 on Build GRCh38
rs191638043 Chr.1: 46654091-46654091 on Build GRCh38
rs538380770 Chr.1: 46654083-46654083 on Build GRCh38
rs912661005 Chr.1: 46654062-46654062 on Build GRCh38
rs566642766 Chr.1: 46653997-46653997 on Build GRCh38
rs533762913 Chr.1: 46654001-46654001 on Build GRCh38
rs991169492 Chr.1: 46653945-46653945 on Build GRCh38
rs930139531 Chr.1: 46654040-46654040 on Build GRCh38
rs76452199 Chr.1: 46654154-46654154 on Build GRCh38
rs750045160 Chr.1: 46653825-46653825 on Build GRCh38
rs767194914 Chr.1: 46653812-46653812 on Build GRCh38
rs758417409 Chr.1: 46653899-46653899 on Build GRCh38
rs761588704 Chr.1: 46654139-46654139 on Build GRCh38
rs141637108 Chr.1: 46653878-46653878 on Build GRCh38
rs148411950 Chr.1: 46653859-46653859 on Build GRCh38
rs1033910822 Chr.1: 46654030-46654030 on Build GRCh38
rs771029568 Chr.1: 46653880-46653880 on Build GRCh38
rs753733066 Chr.1: 46653856-46653856 on Build GRCh38
rs893647159 Chr.1: 46653999-46653999 on Build GRCh38
rs532268293 Chr.1: 46653780-46653780 on Build GRCh38
rs773974690 Chr.1: 46653800-46653800 on Build GRCh38
rs766181455 Chr.1: 46653829-46653829 on Build GRCh38
rs943262688 Chr.1: 46654193-46654193 on Build GRCh38
rs1002586088 Chr.1: 46653868-46653868 on Build GRCh38
rs72679900 Chr.1: 46653909-46653909 on Build GRCh38
rs919050024 Chr.1: 46654151-46654151 on Build GRCh38
rs371196515 Chr.1: 46653789-46653789 on Build GRCh38
rs749066885 Chr.1: 46653873-46653873 on Build GRCh38
rs570833251 Chr.1: 46654051-46654051 on Build GRCh38
rs961572445 Chr.1: 46654146-46654146 on Build GRCh38
rs748259655 Chr.1: 46653883-46653883 on Build GRCh38
rs942681015 Chr.1: 46654087-46654087 on Build GRCh38
rs1003573412 Chr.1: 46654028-46654028 on Build GRCh38
rs889061966 Chr.1: 46653874-46653874 on Build GRCh38
rs116658198 Chr.1: 46654161-46654161 on Build GRCh38
rs367951336 Chr.1: 46653828-46653828 on Build GRCh38
rs1039665732 Chr.1: 46654110-46654110 on Build GRCh38
rs558751192 Chr.1: 46654011-46654011 on Build GRCh38
rs984384418 Chr.1: 46654137-46654137 on Build GRCh38
rs1053632306 Chr.1: 46654199-46654199 on Build GRCh38
rs574500655 Chr.1: 46654138-46654138 on Build GRCh38
rs201480006 Chr.1: 46653857-46653857 on Build GRCh38
rs972223712 Chr.1: 46654147-46654147 on Build GRCh38
rs774385315 Chr.1: 46653902-46653902 on Build GRCh38
rs150528437 Chr.1: 46653931-46653931 on Build GRCh38
rs562025604 Chr.1: 46654008-46654008 on Build GRCh38
rs906519553 Chr.1: 46654009-46654009 on Build GRCh38
rs780218473 Chr.1: 46653987-46653987 on Build GRCh38
rs1015962609 Chr.1: 46653954-46653954 on Build GRCh38
rs760153942 Chr.1: 46653783-46653783 on Build GRCh38

View Full Product Details