Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47038579-47039090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4X1
Entrez Gene ID
260293
Gene Name
cytochrome P450 family 4 subfamily X member 1
Gene Aliases
CYPIVX1
UniGene
Hs.439760
Gene Chromosome Location
Chr.1: 46961364-47055432 on Build GRCh38
Transcripts
NM_001320289.1 NM_001320290.1 NM_178033.1 XM_017000973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4791947 c.780A>G p.T260T Chr.1: 47038664-47038664 on Build GRCh38
6453189 c.817G>A p.V273I Chr.1: 47038701-47038701 on Build GRCh38
681880 c.826G>A p.D276N Chr.1: 47038710-47038710 on Build GRCh38
221909 c.803C>T p.S268F Chr.1: 47038687-47038687 on Build GRCh38
681879 c.862G>T p.D288Y Chr.1: 47038746-47038746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760827883 Chr.1: 47038851-47038851 on Build GRCh38
rs922063895 Chr.1: 47038962-47038962 on Build GRCh38
rs554464008 Chr.1: 47038708-47038708 on Build GRCh38
rs370040143 Chr.1: 47038968-47038968 on Build GRCh38
rs967810367 Chr.1: 47038961-47038961 on Build GRCh38
rs750643338 Chr.1: 47039042-47039042 on Build GRCh38
rs757594947 Chr.1: 47038624-47038624 on Build GRCh38
rs572767042 Chr.1: 47038724-47038724 on Build GRCh38
rs369584412 Chr.1: 47038802-47038802 on Build GRCh38
rs990845842 Chr.1: 47038938-47038938 on Build GRCh38
rs749763180 Chr.1: 47038729-47038729 on Build GRCh38
rs867339305 Chr.1: 47038674-47038674 on Build GRCh38
rs745594509 Chr.1: 47038805-47038805 on Build GRCh38
rs367694773 Chr.1: 47038678-47038678 on Build GRCh38
rs371712304 Chr.1: 47038721-47038721 on Build GRCh38
rs376000916 Chr.1: 47038736-47038736 on Build GRCh38
rs957646762 Chr.1: 47038909-47038909 on Build GRCh38
rs75779875 Chr.1: 47039054-47039054 on Build GRCh38
rs912126425 Chr.1: 47039023-47039023 on Build GRCh38
rs764332839 Chr.1: 47038852-47038852 on Build GRCh38
rs772745954 Chr.1: 47038659-47038659 on Build GRCh38
rs896851013 Chr.1: 47038745-47038745 on Build GRCh38
rs963402663 Chr.1: 47038948-47038948 on Build GRCh38
rs869173919 Chr.1: 47038773-47038781 on Build GRCh38
rs905676171 Chr.1: 47038907-47038907 on Build GRCh38
rs981503952 Chr.1: 47038972-47038972 on Build GRCh38
rs746170511 Chr.1: 47038645-47038645 on Build GRCh38
rs141762705 Chr.1: 47038726-47038726 on Build GRCh38
rs752001897 Chr.1: 47038862-47038862 on Build GRCh38
rs769456719 Chr.1: 47038814-47038814 on Build GRCh38
rs1014886506 Chr.1: 47038944-47038944 on Build GRCh38
rs759191077 Chr.1: 47038687-47038687 on Build GRCh38
rs757637376 Chr.1: 47038973-47038973 on Build GRCh38
rs781347381 Chr.1: 47038976-47038976 on Build GRCh38
rs372717174 Chr.1: 47038655-47038655 on Build GRCh38
rs749862358 Chr.1: 47038653-47038653 on Build GRCh38
rs1029391005 Chr.1: 47038833-47038833 on Build GRCh38
rs2292059 Chr.1: 47039045-47039045 on Build GRCh38
rs1052807789 Chr.1: 47038848-47038848 on Build GRCh38
rs763036133 Chr.1: 47038701-47038701 on Build GRCh38
rs1004801881 Chr.1: 47038934-47038934 on Build GRCh38
rs774240879 Chr.1: 47038854-47038854 on Build GRCh38
rs181567268 Chr.1: 47038816-47038816 on Build GRCh38
rs148362520 Chr.1: 47038920-47038921 on Build GRCh38
rs987691091 Chr.1: 47038989-47038989 on Build GRCh38
rs780241413 Chr.1: 47039055-47039055 on Build GRCh38
rs375053239 Chr.1: 47038672-47038672 on Build GRCh38
rs756504466 Chr.1: 47038646-47038646 on Build GRCh38
rs780515105 Chr.1: 47038727-47038727 on Build GRCh38
rs776483581 Chr.1: 47038711-47038711 on Build GRCh38
rs751651489 Chr.1: 47038720-47038720 on Build GRCh38
rs763582362 Chr.1: 47038747-47038747 on Build GRCh38
rs764994612 Chr.1: 47038966-47038966 on Build GRCh38
rs913223585 Chr.1: 47038945-47038945 on Build GRCh38
rs114314734 Chr.1: 47038716-47038716 on Build GRCh38
rs116368479 Chr.1: 47038664-47038664 on Build GRCh38
rs770483152 Chr.1: 47038754-47038754 on Build GRCh38
rs943676367 Chr.1: 47038826-47038826 on Build GRCh38
rs779411320 Chr.1: 47038737-47038737 on Build GRCh38
rs558356674 Chr.1: 47038863-47038863 on Build GRCh38
rs537407619 Chr.1: 47038977-47038977 on Build GRCh38
rs960024696 Chr.1: 47038920-47038923 on Build GRCh38
rs765151005 Chr.1: 47038695-47038695 on Build GRCh38
rs781508168 Chr.1: 47038644-47038644 on Build GRCh38
rs780420610 Chr.1: 47038651-47038651 on Build GRCh38
rs1018449133 Chr.1: 47038865-47038865 on Build GRCh38

View Full Product Details