Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47046373-47046915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4X1
Entrez Gene ID
260293
Gene Name
cytochrome P450 family 4 subfamily X member 1
Gene Aliases
CYPIVX1
UniGene
Hs.439760
Gene Chromosome Location
Chr.1: 46961364-47055432 on Build GRCh38
Transcripts
NM_001320289.1 NM_001320290.1 NM_178033.1 XM_017000973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6507277 c.1092G>A p.M364I Chr.1: 47046485-47046485 on Build GRCh38
4676036 c.1099A>G p.T367A Chr.1: 47046492-47046492 on Build GRCh38
3490445 c.1159G>A p.D387N Chr.1: 47046552-47046552 on Build GRCh38
5892563 c.1183C>T p.P395S Chr.1: 47046576-47046576 on Build GRCh38
3490444 c.1117G>A p.E373K Chr.1: 47046510-47046510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748096160 Chr.1: 47046433-47046434 on Build GRCh38
rs746979570 Chr.1: 47046579-47046579 on Build GRCh38
rs1020393104 Chr.1: 47046697-47046697 on Build GRCh38
rs371034972 Chr.1: 47046603-47046603 on Build GRCh38
rs899246907 Chr.1: 47046648-47046648 on Build GRCh38
rs763312421 Chr.1: 47046612-47046612 on Build GRCh38
rs768547554 Chr.1: 47046424-47046424 on Build GRCh38
rs769828914 Chr.1: 47046436-47046437 on Build GRCh38
rs912948393 Chr.1: 47046808-47046808 on Build GRCh38
rs953616745 Chr.1: 47046405-47046405 on Build GRCh38
rs937278070 Chr.1: 47046571-47046571 on Build GRCh38
rs61742240 Chr.1: 47046539-47046539 on Build GRCh38
rs773032325 Chr.1: 47046454-47046454 on Build GRCh38
rs771062649 Chr.1: 47046483-47046483 on Build GRCh38
rs754545262 Chr.1: 47046646-47046646 on Build GRCh38
rs764434913 Chr.1: 47046617-47046617 on Build GRCh38
rs978331076 Chr.1: 47046731-47046731 on Build GRCh38
rs770978903 Chr.1: 47046589-47046589 on Build GRCh38
rs188593334 Chr.1: 47046791-47046791 on Build GRCh38
rs111378209 Chr.1: 47046740-47046740 on Build GRCh38
rs550336520 Chr.1: 47046880-47046880 on Build GRCh38
rs571222973 Chr.1: 47046858-47046858 on Build GRCh38
rs183819145 Chr.1: 47046779-47046779 on Build GRCh38
rs762447868 Chr.1: 47046630-47046630 on Build GRCh38
rs957379898 Chr.1: 47046665-47046665 on Build GRCh38
rs966498798 Chr.1: 47046723-47046723 on Build GRCh38
rs1038052934 Chr.1: 47046855-47046855 on Build GRCh38
rs373629505 Chr.1: 47046474-47046474 on Build GRCh38
rs372889638 Chr.1: 47046499-47046499 on Build GRCh38
rs375390499 Chr.1: 47046624-47046624 on Build GRCh38
rs759805454 Chr.1: 47046495-47046495 on Build GRCh38
rs267598628 Chr.1: 47046583-47046583 on Build GRCh38
rs755646900 Chr.1: 47046536-47046536 on Build GRCh38
rs776736370 Chr.1: 47046597-47046597 on Build GRCh38
rs530443675 Chr.1: 47046702-47046702 on Build GRCh38
rs778891378 Chr.1: 47046432-47046432 on Build GRCh38
rs941044753 Chr.1: 47046807-47046807 on Build GRCh38
rs747915842 Chr.1: 47046560-47046560 on Build GRCh38
rs1019235945 Chr.1: 47046757-47046757 on Build GRCh38
rs769956225 Chr.1: 47046601-47046601 on Build GRCh38
rs760728168 Chr.1: 47046716-47046716 on Build GRCh38
rs994979401 Chr.1: 47046680-47046681 on Build GRCh38
rs965440714 Chr.1: 47046514-47046514 on Build GRCh38
rs79113880 Chr.1: 47046871-47046871 on Build GRCh38
rs147541736 Chr.1: 47046550-47046550 on Build GRCh38
rs1011641938 Chr.1: 47046684-47046684 on Build GRCh38
rs146623813 Chr.1: 47046741-47046741 on Build GRCh38
rs368977020 Chr.1: 47046488-47046488 on Build GRCh38
rs112171351 Chr.1: 47046742-47046742 on Build GRCh38
rs916282190 Chr.1: 47046498-47046498 on Build GRCh38
rs749574923 Chr.1: 47046772-47046772 on Build GRCh38
rs372294459 Chr.1: 47046638-47046638 on Build GRCh38
rs758128782 Chr.1: 47046573-47046573 on Build GRCh38
rs775794824 Chr.1: 47046510-47046510 on Build GRCh38
rs994304525 Chr.1: 47046569-47046569 on Build GRCh38
rs562884722 Chr.1: 47046660-47046660 on Build GRCh38
rs145695080 Chr.1: 47046576-47046576 on Build GRCh38
rs778729788 Chr.1: 47046541-47046541 on Build GRCh38
rs766034515 Chr.1: 47046538-47046538 on Build GRCh38
rs376322382 Chr.1: 47046444-47046444 on Build GRCh38
rs377497903 Chr.1: 47046534-47046534 on Build GRCh38
rs747925371 Chr.1: 47046434-47046434 on Build GRCh38
rs768169853 Chr.1: 47046636-47046636 on Build GRCh38
rs867689698 Chr.1: 47046582-47046582 on Build GRCh38
rs192884867 Chr.1: 47046826-47046826 on Build GRCh38
rs560678413 Chr.1: 47046770-47046770 on Build GRCh38
rs990781094 Chr.1: 47046781-47046781 on Build GRCh38
rs1029208059 Chr.1: 47046402-47046402 on Build GRCh38
rs775503726 Chr.1: 47046519-47046519 on Build GRCh38

View Full Product Details