Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47067246-47067743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4Z1
Entrez Gene ID
199974
Gene Name
cytochrome P450 family 4 subfamily Z member 1
Gene Aliases
CYP4A20
UniGene
Hs.176588
Gene Chromosome Location
Chr.1: 47067488-47118320 on Build GRCh38
Transcripts
NM_178134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7307732 c.45G>T p.L15F Chr.1: 47067535-47067535 on Build GRCh38
3490463 c.169C>T p.H57Y Chr.1: 47067659-47067659 on Build GRCh38
3490462 c.162C>T p.F54F Chr.1: 47067652-47067652 on Build GRCh38
3966630 c.21G>C p.Q7H Chr.1: 47067511-47067511 on Build GRCh38
3490461 c.139C>T p.P47S Chr.1: 47067629-47067629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041015539 Chr.1: 47067274-47067274 on Build GRCh38
rs116390521 Chr.1: 47067313-47067313 on Build GRCh38
rs751710132 Chr.1: 47067503-47067503 on Build GRCh38
rs145478155 Chr.1: 47067638-47067638 on Build GRCh38
rs751025533 Chr.1: 47067682-47067682 on Build GRCh38
rs368911622 Chr.1: 47067552-47067552 on Build GRCh38
rs564652252 Chr.1: 47067607-47067607 on Build GRCh38
rs746529579 Chr.1: 47067554-47067554 on Build GRCh38
rs958937822 Chr.1: 47067375-47067375 on Build GRCh38
rs368380182 Chr.1: 47067522-47067522 on Build GRCh38
rs768825844 Chr.1: 47067615-47067615 on Build GRCh38
rs199709621 Chr.1: 47067679-47067679 on Build GRCh38
rs548967488 Chr.1: 47067668-47067668 on Build GRCh38
rs763125700 Chr.1: 47067501-47067501 on Build GRCh38
rs918260183 Chr.1: 47067478-47067478 on Build GRCh38
rs763033972 Chr.1: 47067613-47067613 on Build GRCh38
rs569596729 Chr.1: 47067455-47067455 on Build GRCh38
rs901159907 Chr.1: 47067276-47067276 on Build GRCh38
rs555609744 Chr.1: 47067617-47067617 on Build GRCh38
rs952529626 Chr.1: 47067479-47067479 on Build GRCh38
rs372321679 Chr.1: 47067525-47067525 on Build GRCh38
rs767665833 Chr.1: 47067511-47067511 on Build GRCh38
rs750307082 Chr.1: 47067669-47067669 on Build GRCh38
rs780893510 Chr.1: 47067694-47067694 on Build GRCh38
rs769799961 Chr.1: 47067490-47067490 on Build GRCh38
rs866352423 Chr.1: 47067493-47067493 on Build GRCh38
rs769783483 Chr.1: 47067604-47067604 on Build GRCh38
rs375915739 Chr.1: 47067540-47067540 on Build GRCh38
rs761809766 Chr.1: 47067625-47067625 on Build GRCh38
rs754308581 Chr.1: 47067663-47067663 on Build GRCh38
rs267598629 Chr.1: 47067512-47067512 on Build GRCh38
rs761013597 Chr.1: 47067645-47067645 on Build GRCh38
rs770566853 Chr.1: 47067569-47067569 on Build GRCh38
rs767710259 Chr.1: 47067635-47067635 on Build GRCh38
rs754400619 Chr.1: 47067528-47067528 on Build GRCh38
rs922487285 Chr.1: 47067642-47067642 on Build GRCh38
rs780725041 Chr.1: 47067589-47067589 on Build GRCh38
rs1020313144 Chr.1: 47067301-47067301 on Build GRCh38
rs528660840 Chr.1: 47067294-47067294 on Build GRCh38
rs753086586 Chr.1: 47067525-47067526 on Build GRCh38
rs780534056 Chr.1: 47067526-47067526 on Build GRCh38
rs142469011 Chr.1: 47067649-47067649 on Build GRCh38
rs372632854 Chr.1: 47067418-47067418 on Build GRCh38
rs750739802 Chr.1: 47067523-47067523 on Build GRCh38
rs764327783 Chr.1: 47067502-47067502 on Build GRCh38
rs533054607 Chr.1: 47067317-47067317 on Build GRCh38
rs551128692 Chr.1: 47067361-47067361 on Build GRCh38
rs1034930324 Chr.1: 47067334-47067334 on Build GRCh38
rs60000772 Chr.1: 47067687-47067687 on Build GRCh38
rs759273712 Chr.1: 47067483-47067483 on Build GRCh38
rs868399298 Chr.1: 47067629-47067629 on Build GRCh38
rs767835636 Chr.1: 47067517-47067518 on Build GRCh38
rs547197836 Chr.1: 47067307-47067307 on Build GRCh38
rs749129206 Chr.1: 47067707-47067707 on Build GRCh38
rs756654835 Chr.1: 47067533-47067534 on Build GRCh38
rs775555015 Chr.1: 47067606-47067606 on Build GRCh38
rs762137487 Chr.1: 47067505-47067505 on Build GRCh38
rs764545950 Chr.1: 47067591-47067592 on Build GRCh38
rs755955156 Chr.1: 47067703-47067703 on Build GRCh38
rs768591658 Chr.1: 47067709-47067709 on Build GRCh38
rs745595031 Chr.1: 47067595-47067595 on Build GRCh38
rs779093090 Chr.1: 47067690-47067690 on Build GRCh38
rs553610945 Chr.1: 47067706-47067706 on Build GRCh38
rs774157098 Chr.1: 47067717-47067717 on Build GRCh38
rs201327598 Chr.1: 47067464-47067464 on Build GRCh38
rs184537310 Chr.1: 47067701-47067701 on Build GRCh38

View Full Product Details