Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47435871-47436369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXD2
Entrez Gene ID
2306
Gene Name
forkhead box D2
Gene Aliases
FKHL17, FREAC-9, FREAC9
UniGene
Hs.166188
Gene Chromosome Location
Chr.1: 47436017-47440691 on Build GRCh38
Transcripts
NM_004474.3 NR_126355.1

Gene Symbol
LINC01389
Entrez Gene ID
102724077
Gene Name
long intergenic non-protein coding RNA 1389
Gene Aliases
-
UniGene
Hs.159718
Gene Chromosome Location
Chr.1: 47380796-47437317 on Build GRCh38
Transcripts
NM_004474.3 NR_126355.1

SNP Detail

SNP ID Chromosome Location
rs559133510 Chr.1: 47436285-47436285 on Build GRCh38
rs41290150 Chr.1: 47435914-47435914 on Build GRCh38
rs967664388 Chr.1: 47436201-47436201 on Build GRCh38
rs937365859 Chr.1: 47436227-47436227 on Build GRCh38
rs141097619 Chr.1: 47436049-47436049 on Build GRCh38
rs1006543771 Chr.1: 47436064-47436064 on Build GRCh38
rs928882920 Chr.1: 47436209-47436209 on Build GRCh38
rs978382370 Chr.1: 47436150-47436150 on Build GRCh38
rs909341062 Chr.1: 47436279-47436279 on Build GRCh38
rs1018065966 Chr.1: 47436110-47436110 on Build GRCh38
rs529465934 Chr.1: 47436050-47436050 on Build GRCh38
rs1009702767 Chr.1: 47436305-47436305 on Build GRCh38
rs968325456 Chr.1: 47436137-47436137 on Build GRCh38
rs545775458 Chr.1: 47435903-47435903 on Build GRCh38
rs896951199 Chr.1: 47436312-47436312 on Build GRCh38
rs940539691 Chr.1: 47436180-47436180 on Build GRCh38
rs146529604 Chr.1: 47436015-47436015 on Build GRCh38
rs926983159 Chr.1: 47436151-47436151 on Build GRCh38
rs1057347576 Chr.1: 47435919-47435919 on Build GRCh38
rs562183264 Chr.1: 47436303-47436303 on Build GRCh38
rs773931649 Chr.1: 47436187-47436187 on Build GRCh38
rs145652402 Chr.1: 47436297-47436297 on Build GRCh38
rs953598527 Chr.1: 47436045-47436045 on Build GRCh38
rs1022446538 Chr.1: 47436309-47436309 on Build GRCh38
rs368460738 Chr.1: 47436063-47436063 on Build GRCh38
rs887847405 Chr.1: 47435921-47435921 on Build GRCh38
rs537973306 Chr.1: 47436002-47436002 on Build GRCh38
rs912999846 Chr.1: 47436111-47436111 on Build GRCh38
rs982075879 Chr.1: 47436207-47436207 on Build GRCh38
rs560001456 Chr.1: 47436157-47436157 on Build GRCh38
rs6691723 Chr.1: 47435916-47435916 on Build GRCh38
rs765146206 Chr.1: 47436037-47436038 on Build GRCh38
rs895692717 Chr.1: 47435923-47435923 on Build GRCh38
rs761405891 Chr.1: 47436273-47436273 on Build GRCh38
rs1005376238 Chr.1: 47435970-47435970 on Build GRCh38
rs1036425195 Chr.1: 47436192-47436192 on Build GRCh38
rs937254809 Chr.1: 47436152-47436152 on Build GRCh38
rs78418970 Chr.1: 47436164-47436164 on Build GRCh38
rs185308544 Chr.1: 47436163-47436163 on Build GRCh38
rs1038174432 Chr.1: 47436302-47436302 on Build GRCh38
rs1036458362 Chr.1: 47435983-47435983 on Build GRCh38
rs1038164124 Chr.1: 47435999-47435999 on Build GRCh38

View Full Product Details