Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47436661-47437167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXD2
Entrez Gene ID
2306
Gene Name
forkhead box D2
Gene Aliases
FKHL17, FREAC-9, FREAC9
UniGene
Hs.166188
Gene Chromosome Location
Chr.1: 47436017-47440691 on Build GRCh38
Transcripts
NM_004474.3 NR_126355.1

Gene Symbol
LINC01389
Entrez Gene ID
102724077
Gene Name
long intergenic non-protein coding RNA 1389
Gene Aliases
-
UniGene
Hs.159718
Gene Chromosome Location
Chr.1: 47380796-47437317 on Build GRCh38
Transcripts
NM_004474.3 NR_126355.1

SNP Detail

SNP ID Chromosome Location
rs77279256 Chr.1: 47437000-47437000 on Build GRCh38
rs944813832 Chr.1: 47436786-47436786 on Build GRCh38
rs1043965387 Chr.1: 47436803-47436803 on Build GRCh38
rs527634936 Chr.1: 47437040-47437040 on Build GRCh38
rs986190486 Chr.1: 47436772-47436772 on Build GRCh38
rs1055281601 Chr.1: 47437002-47437002 on Build GRCh38
rs1039397113 Chr.1: 47436806-47436806 on Build GRCh38
rs896687467 Chr.1: 47437028-47437028 on Build GRCh38
rs910398724 Chr.1: 47436916-47436916 on Build GRCh38
rs894212738 Chr.1: 47436852-47436853 on Build GRCh38
rs1035822897 Chr.1: 47436893-47436893 on Build GRCh38
rs1024421158 Chr.1: 47436695-47436696 on Build GRCh38
rs925553557 Chr.1: 47436998-47436998 on Build GRCh38
rs963372046 Chr.1: 47436940-47436940 on Build GRCh38
rs373856551 Chr.1: 47436790-47436790 on Build GRCh38
rs183582883 Chr.1: 47436704-47436704 on Build GRCh38
rs951829468 Chr.1: 47436902-47436902 on Build GRCh38
rs549459855 Chr.1: 47436932-47436932 on Build GRCh38
rs902933820 Chr.1: 47437092-47437092 on Build GRCh38
rs1014446227 Chr.1: 47436863-47436863 on Build GRCh38
rs868811309 Chr.1: 47436811-47436811 on Build GRCh38
rs1027351631 Chr.1: 47437104-47437104 on Build GRCh38
rs1020480487 Chr.1: 47436840-47436840 on Build GRCh38
rs576732533 Chr.1: 47436722-47436722 on Build GRCh38
rs533655445 Chr.1: 47436796-47436796 on Build GRCh38
rs1023972070 Chr.1: 47436875-47436875 on Build GRCh38
rs910649643 Chr.1: 47436778-47436779 on Build GRCh38
rs945622980 Chr.1: 47436801-47436801 on Build GRCh38
rs961969376 Chr.1: 47436696-47436696 on Build GRCh38
rs1036280531 Chr.1: 47437130-47437130 on Build GRCh38
rs913871347 Chr.1: 47436931-47436931 on Build GRCh38
rs993373671 Chr.1: 47436899-47436899 on Build GRCh38
rs746688663 Chr.1: 47436758-47436758 on Build GRCh38
rs991672293 Chr.1: 47436910-47436910 on Build GRCh38
rs3738316 Chr.1: 47437145-47437145 on Build GRCh38
rs1003489479 Chr.1: 47436887-47436887 on Build GRCh38
rs547550825 Chr.1: 47437045-47437045 on Build GRCh38
rs1041378335 Chr.1: 47437069-47437069 on Build GRCh38
rs769998406 Chr.1: 47437064-47437064 on Build GRCh38
rs1045892111 Chr.1: 47437042-47437042 on Build GRCh38
rs192572116 Chr.1: 47436683-47436683 on Build GRCh38
rs942557595 Chr.1: 47436804-47436804 on Build GRCh38
rs987043631 Chr.1: 47436996-47436996 on Build GRCh38
rs966150476 Chr.1: 47436921-47436921 on Build GRCh38
rs1002347704 Chr.1: 47436809-47436809 on Build GRCh38
rs972077877 Chr.1: 47436947-47436947 on Build GRCh38
rs545410276 Chr.1: 47436828-47436828 on Build GRCh38
rs972535420 Chr.1: 47436708-47436708 on Build GRCh38
rs1002592811 Chr.1: 47437099-47437099 on Build GRCh38
rs143263465 Chr.1: 47436895-47436895 on Build GRCh38
rs181182968 Chr.1: 47437098-47437098 on Build GRCh38
rs902921724 Chr.1: 47436872-47436872 on Build GRCh38
rs929828644 Chr.1: 47436729-47436729 on Build GRCh38
rs1011226842 Chr.1: 47436824-47436824 on Build GRCh38
rs892779923 Chr.1: 47436807-47436807 on Build GRCh38
rs919370998 Chr.1: 47436955-47436955 on Build GRCh38
rs933418480 Chr.1: 47436992-47436992 on Build GRCh38
rs976606090 Chr.1: 47436931-47436931 on Build GRCh38
rs567752860 Chr.1: 47436999-47436999 on Build GRCh38
rs148352222 Chr.1: 47436873-47436873 on Build GRCh38
rs188255170 Chr.1: 47436756-47436756 on Build GRCh38
rs947067511 Chr.1: 47437052-47437052 on Build GRCh38
rs573852930 Chr.1: 47436710-47436710 on Build GRCh38

View Full Product Details