Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47440218-47440715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXD2
Entrez Gene ID
2306
Gene Name
forkhead box D2
Gene Aliases
FKHL17, FREAC-9, FREAC9
UniGene
Hs.166188
Gene Chromosome Location
Chr.1: 47436017-47440691 on Build GRCh38
Transcripts
NM_004474.3

SNP Detail

SNP ID Chromosome Location
rs1031599897 Chr.1: 47440330-47440330 on Build GRCh38
rs948212188 Chr.1: 47440496-47440496 on Build GRCh38
rs928859581 Chr.1: 47440445-47440445 on Build GRCh38
rs777116704 Chr.1: 47440395-47440395 on Build GRCh38
rs908603304 Chr.1: 47440477-47440477 on Build GRCh38
rs892200821 Chr.1: 47440553-47440553 on Build GRCh38
rs181478049 Chr.1: 47440354-47440354 on Build GRCh38
rs908183158 Chr.1: 47440466-47440466 on Build GRCh38
rs982864494 Chr.1: 47440465-47440465 on Build GRCh38
rs923224448 Chr.1: 47440526-47440526 on Build GRCh38
rs550448073 Chr.1: 47440690-47440690 on Build GRCh38
rs931849900 Chr.1: 47440529-47440529 on Build GRCh38
rs72686276 Chr.1: 47440442-47440442 on Build GRCh38
rs975907172 Chr.1: 47440489-47440489 on Build GRCh38
rs570422403 Chr.1: 47440367-47440368 on Build GRCh38
rs961902185 Chr.1: 47440482-47440482 on Build GRCh38
rs547418913 Chr.1: 47440302-47440302 on Build GRCh38
rs956043891 Chr.1: 47440349-47440349 on Build GRCh38
rs560778798 Chr.1: 47440381-47440381 on Build GRCh38
rs775655686 Chr.1: 47440483-47440483 on Build GRCh38
rs905459871 Chr.1: 47440646-47440646 on Build GRCh38
rs1050524431 Chr.1: 47440537-47440537 on Build GRCh38
rs1038068485 Chr.1: 47440479-47440479 on Build GRCh38
rs1013746025 Chr.1: 47440555-47440555 on Build GRCh38
rs1046508377 Chr.1: 47440630-47440630 on Build GRCh38
rs1026952828 Chr.1: 47440452-47440452 on Build GRCh38
rs564699951 Chr.1: 47440470-47440470 on Build GRCh38
rs528360618 Chr.1: 47440409-47440409 on Build GRCh38
rs574633737 Chr.1: 47440328-47440328 on Build GRCh38
rs2820966 Chr.1: 47440666-47440666 on Build GRCh38
rs759802347 Chr.1: 47440420-47440420 on Build GRCh38
rs956013158 Chr.1: 47440610-47440610 on Build GRCh38
rs1053050514 Chr.1: 47440281-47440281 on Build GRCh38
rs1031735555 Chr.1: 47440595-47440595 on Build GRCh38
rs765051088 Chr.1: 47440633-47440633 on Build GRCh38
rs952920515 Chr.1: 47440461-47440461 on Build GRCh38
rs936313709 Chr.1: 47440540-47440540 on Build GRCh38
rs1025213660 Chr.1: 47440317-47440317 on Build GRCh38
rs1008227888 Chr.1: 47440579-47440579 on Build GRCh38
rs568961280 Chr.1: 47440548-47440548 on Build GRCh38
rs997070809 Chr.1: 47440249-47440249 on Build GRCh38
rs939680529 Chr.1: 47440472-47440472 on Build GRCh38
rs368097251 Chr.1: 47440247-47440247 on Build GRCh38
rs552806503 Chr.1: 47440541-47440541 on Build GRCh38
rs898387403 Chr.1: 47440492-47440492 on Build GRCh38
rs924946940 Chr.1: 47440538-47440538 on Build GRCh38
rs747505746 Chr.1: 47440282-47440282 on Build GRCh38
rs532140366 Chr.1: 47440528-47440528 on Build GRCh38

View Full Product Details