Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51265280-51265780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIGLECL1
Entrez Gene ID
284369
Gene Name
SIGLEC family like 1
Gene Aliases
C19orf75, SIGLEC23P, SIGLECP7
UniGene
Hs.381087
Gene Chromosome Location
Chr.19: 51251064-51269330 on Build GRCh38
Transcripts
NM_001308437.1 NM_173635.2 NR_102324.1 XM_005258799.4 XM_011526821.2 XM_017026647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1129739 c.104C>T p.P35L Chr.19: 51265449-51265449 on Build GRCh38
5718064 c.125G>A p.G42E Chr.19: 51265470-51265470 on Build GRCh38
3727468 c.118_119insG p.M41fs*27 Chr.19: 51265463-51265464 on Build GRCh38
5715733 c.107C>T p.S36F Chr.19: 51265452-51265452 on Build GRCh38
3835503 c.242G>A p.R81K Chr.19: 51265587-51265587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765065891 Chr.19: 51265678-51265678 on Build GRCh38
rs758744287 Chr.19: 51265444-51265444 on Build GRCh38
rs1007207362 Chr.19: 51265700-51265700 on Build GRCh38
rs199591888 Chr.19: 51265692-51265692 on Build GRCh38
rs748129434 Chr.19: 51265338-51265338 on Build GRCh38
rs376539635 Chr.19: 51265465-51265465 on Build GRCh38
rs770701275 Chr.19: 51265492-51265492 on Build GRCh38
rs775760620 Chr.19: 51265753-51265753 on Build GRCh38
rs889731471 Chr.19: 51265675-51265675 on Build GRCh38
rs754458202 Chr.19: 51265698-51265698 on Build GRCh38
rs551566574 Chr.19: 51265504-51265504 on Build GRCh38
rs757146042 Chr.19: 51265674-51265674 on Build GRCh38
rs770319733 Chr.19: 51265501-51265501 on Build GRCh38
rs201813184 Chr.19: 51265353-51265353 on Build GRCh38
rs146040864 Chr.19: 51265506-51265506 on Build GRCh38
rs762439800 Chr.19: 51265403-51265403 on Build GRCh38
rs1054656298 Chr.19: 51265617-51265617 on Build GRCh38
rs749097089 Chr.19: 51265486-51265486 on Build GRCh38
rs751815517 Chr.19: 51265453-51265453 on Build GRCh38
rs746687080 Chr.19: 51265697-51265697 on Build GRCh38
rs767523056 Chr.19: 51265546-51265546 on Build GRCh38
rs962532393 Chr.19: 51265714-51265714 on Build GRCh38
rs138878557 Chr.19: 51265645-51265645 on Build GRCh38
rs781071937 Chr.19: 51265330-51265330 on Build GRCh38
rs781276468 Chr.19: 51265475-51265475 on Build GRCh38
rs780594586 Chr.19: 51265730-51265730 on Build GRCh38
rs773742543 Chr.19: 51265622-51265622 on Build GRCh38
rs1049108846 Chr.19: 51265304-51265304 on Build GRCh38
rs909323421 Chr.19: 51265666-51265666 on Build GRCh38
rs947885674 Chr.19: 51265662-51265662 on Build GRCh38
rs751727740 Chr.19: 51265530-51265530 on Build GRCh38
rs759789104 Chr.19: 51265534-51265534 on Build GRCh38
rs1029617725 Chr.19: 51265437-51265437 on Build GRCh38
rs181307292 Chr.19: 51265558-51265558 on Build GRCh38
rs757054094 Chr.19: 51265578-51265578 on Build GRCh38
rs768089178 Chr.19: 51265616-51265616 on Build GRCh38
rs771863729 Chr.19: 51265589-51265589 on Build GRCh38
rs775143727 Chr.19: 51265500-51265500 on Build GRCh38
rs140988748 Chr.19: 51265366-51265366 on Build GRCh38
rs755115343 Chr.19: 51265328-51265328 on Build GRCh38
rs755290333 Chr.19: 51265626-51265636 on Build GRCh38
rs189787158 Chr.19: 51265399-51265399 on Build GRCh38
rs746230636 Chr.19: 51265745-51265745 on Build GRCh38
rs992049805 Chr.19: 51265623-51265623 on Build GRCh38
rs539316539 Chr.19: 51265634-51265634 on Build GRCh38
rs759699154 Chr.19: 51265655-51265655 on Build GRCh38
rs777763483 Chr.19: 51265565-51265565 on Build GRCh38
rs372679715 Chr.19: 51265519-51265519 on Build GRCh38
rs114737752 Chr.19: 51265669-51265669 on Build GRCh38
rs369594050 Chr.19: 51265649-51265649 on Build GRCh38
rs772468283 Chr.19: 51265752-51265752 on Build GRCh38
rs200624136 Chr.19: 51265691-51265691 on Build GRCh38
rs768096686 Chr.19: 51265658-51265658 on Build GRCh38
rs766616657 Chr.19: 51265447-51265447 on Build GRCh38
rs529230865 Chr.19: 51265362-51265362 on Build GRCh38
rs763388576 Chr.19: 51265642-51265642 on Build GRCh38
rs771445197 Chr.19: 51265742-51265742 on Build GRCh38
rs368913658 Chr.19: 51265470-51265470 on Build GRCh38
rs749740307 Chr.19: 51265731-51265731 on Build GRCh38
rs765488467 Chr.19: 51265661-51265661 on Build GRCh38
rs753662946 Chr.19: 51265522-51265548 on Build GRCh38
rs200975543 Chr.19: 51265481-51265481 on Build GRCh38
rs779818809 Chr.19: 51265694-51265694 on Build GRCh38
rs972802579 Chr.19: 51265417-51265417 on Build GRCh38
rs752594887 Chr.19: 51265480-51265480 on Build GRCh38
rs752985085 Chr.19: 51265661-51265661 on Build GRCh38
rs1875425 Chr.19: 51265713-51265713 on Build GRCh38
rs776429563 Chr.19: 51265409-51265409 on Build GRCh38
rs766297308 Chr.19: 51265495-51265497 on Build GRCh38
rs953534898 Chr.19: 51265449-51265449 on Build GRCh38
rs778596682 Chr.19: 51265495-51265495 on Build GRCh38
rs201947873 Chr.19: 51265522-51265522 on Build GRCh38
rs887788962 Chr.19: 51265307-51265307 on Build GRCh38
rs201513690 Chr.19: 51265668-51265668 on Build GRCh38
rs778874793 Chr.19: 51265581-51265581 on Build GRCh38
rs766882539 Chr.19: 51265381-51265382 on Build GRCh38
rs773934178 Chr.19: 51265371-51265371 on Build GRCh38
rs747074504 Chr.19: 51265322-51265322 on Build GRCh38
rs771667314 Chr.19: 51265497-51265497 on Build GRCh38
rs752790795 Chr.19: 51265554-51265554 on Build GRCh38
rs539276316 Chr.19: 51265400-51265400 on Build GRCh38
rs376395750 Chr.19: 51265615-51265615 on Build GRCh38
rs760648589 Chr.19: 51265483-51265483 on Build GRCh38
rs201504998 Chr.19: 51265743-51265743 on Build GRCh38
rs369666963 Chr.19: 51265321-51265321 on Build GRCh38
rs750811347 Chr.19: 51265412-51265412 on Build GRCh38
rs766699638 Chr.19: 51265524-51265524 on Build GRCh38
rs758283083 Chr.19: 51265466-51265466 on Build GRCh38
rs866540825 Chr.19: 51265484-51265484 on Build GRCh38
rs555449153 Chr.19: 51265610-51265610 on Build GRCh38
rs745566229 Chr.19: 51265496-51265496 on Build GRCh38
rs768355009 Chr.19: 51265378-51265378 on Build GRCh38
rs777377064 Chr.19: 51265485-51265485 on Build GRCh38
rs761660442 Chr.19: 51265410-51265410 on Build GRCh38
rs773208210 Chr.19: 51265463-51265464 on Build GRCh38
rs765214386 Chr.19: 51265664-51265664 on Build GRCh38
rs753762107 Chr.19: 51265577-51265577 on Build GRCh38
rs201133234 Chr.19: 51265687-51265687 on Build GRCh38

View Full Product Details