Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51366179-51366677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETFB
Entrez Gene ID
2109
Gene Name
electron transfer flavoprotein beta subunit
Gene Aliases
FP585, MADD
UniGene
Hs.348531
Gene Chromosome Location
Chr.19: 51345155-51366418 on Build GRCh38
Transcripts
NM_001985.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1172651 c.48C>G p.Y16* Chr.19: 51366279-51366279 on Build GRCh38
3938238 c.35G>A p.R12K Chr.19: 51366292-51366292 on Build GRCh38
6848798 c.57+1G>A p.? Chr.19: 51366269-51366269 on Build GRCh38
3537594 c.41T>C p.I14T Chr.19: 51366286-51366286 on Build GRCh38
6302508 c.32A>G p.K11R Chr.19: 51366295-51366295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976881929 Chr.19: 51366563-51366563 on Build GRCh38
rs531136177 Chr.19: 51366305-51366305 on Build GRCh38
rs749447765 Chr.19: 51366219-51366219 on Build GRCh38
rs977622819 Chr.19: 51366390-51366390 on Build GRCh38
rs199908626 Chr.19: 51366563-51366566 on Build GRCh38
rs890741300 Chr.19: 51366631-51366631 on Build GRCh38
rs761117970 Chr.19: 51366357-51366357 on Build GRCh38
rs768140061 Chr.19: 51366348-51366348 on Build GRCh38
rs376323196 Chr.19: 51366442-51366442 on Build GRCh38
rs370961463 Chr.19: 51366320-51366320 on Build GRCh38
rs572600030 Chr.19: 51366309-51366309 on Build GRCh38
rs200019312 Chr.19: 51366306-51366306 on Build GRCh38
rs745810923 Chr.19: 51366312-51366312 on Build GRCh38
rs750556328 Chr.19: 51366367-51366367 on Build GRCh38
rs79014357 Chr.19: 51366568-51366568 on Build GRCh38
rs982058479 Chr.19: 51366405-51366405 on Build GRCh38
rs767574879 Chr.19: 51366499-51366499 on Build GRCh38
rs370928045 Chr.19: 51366233-51366233 on Build GRCh38
rs949269058 Chr.19: 51366313-51366313 on Build GRCh38
rs184345178 Chr.19: 51366613-51366613 on Build GRCh38
rs532871800 Chr.19: 51366582-51366582 on Build GRCh38
rs550270514 Chr.19: 51366524-51366524 on Build GRCh38
rs986738204 Chr.19: 51366439-51366439 on Build GRCh38
rs757359427 Chr.19: 51366360-51366360 on Build GRCh38
rs191102821 Chr.19: 51366573-51366573 on Build GRCh38
rs993486919 Chr.19: 51366591-51366591 on Build GRCh38
rs1017767353 Chr.19: 51366447-51366447 on Build GRCh38
rs757249379 Chr.19: 51366222-51366222 on Build GRCh38
rs761369109 Chr.19: 51366247-51366247 on Build GRCh38
rs971906805 Chr.19: 51366389-51366389 on Build GRCh38
rs753150123 Chr.19: 51366276-51366276 on Build GRCh38
rs769316868 Chr.19: 51366339-51366339 on Build GRCh38
rs374376098 Chr.19: 51366342-51366342 on Build GRCh38
rs923144754 Chr.19: 51366398-51366399 on Build GRCh38
rs764471064 Chr.19: 51366301-51366301 on Build GRCh38
rs183857804 Chr.19: 51366224-51366224 on Build GRCh38
rs958319657 Chr.19: 51366540-51366540 on Build GRCh38
rs749836128 Chr.19: 51366347-51366347 on Build GRCh38
rs775707781 Chr.19: 51366355-51366355 on Build GRCh38
rs747618707 Chr.19: 51366335-51366335 on Build GRCh38
rs987789074 Chr.19: 51366536-51366536 on Build GRCh38
rs754140700 Chr.19: 51366359-51366359 on Build GRCh38
rs754286628 Chr.19: 51366340-51366341 on Build GRCh38
rs759897705 Chr.19: 51366263-51366263 on Build GRCh38
rs970618581 Chr.19: 51366398-51366398 on Build GRCh38
rs186691166 Chr.19: 51366234-51366234 on Build GRCh38
rs954969761 Chr.19: 51366437-51366437 on Build GRCh38
rs985998825 Chr.19: 51366592-51366592 on Build GRCh38
rs775552959 Chr.19: 51366504-51366504 on Build GRCh38
rs202118581 Chr.19: 51366362-51366362 on Build GRCh38
rs766807520 Chr.19: 51366256-51366256 on Build GRCh38
rs1018237484 Chr.19: 51366650-51366650 on Build GRCh38
rs148261223 Chr.19: 51366287-51366287 on Build GRCh38
rs1010821340 Chr.19: 51366551-51366551 on Build GRCh38
rs1051763290 Chr.19: 51366619-51366619 on Build GRCh38
rs111454736 Chr.19: 51366384-51366384 on Build GRCh38
rs764342231 Chr.19: 51366358-51366358 on Build GRCh38
rs1037411459 Chr.19: 51366461-51366461 on Build GRCh38
rs1026174014 Chr.19: 51366392-51366392 on Build GRCh38
rs754801507 Chr.19: 51366371-51366371 on Build GRCh38
rs370777932 Chr.19: 51366202-51366202 on Build GRCh38
rs768283703 Chr.19: 51366327-51366327 on Build GRCh38
rs949325814 Chr.19: 51366550-51366550 on Build GRCh38
rs981006954 Chr.19: 51366283-51366283 on Build GRCh38
rs536036940 Chr.19: 51366558-51366558 on Build GRCh38
rs751253707 Chr.19: 51366370-51366370 on Build GRCh38
rs760595402 Chr.19: 51366609-51366609 on Build GRCh38
rs941836051 Chr.19: 51366562-51366562 on Build GRCh38
rs746888203 Chr.19: 51366325-51366325 on Build GRCh38
rs760298365 Chr.19: 51366560-51366560 on Build GRCh38
rs377667900 Chr.19: 51366227-51366227 on Build GRCh38
rs761283628 Chr.19: 51366237-51366237 on Build GRCh38
rs771793144 Chr.19: 51366228-51366228 on Build GRCh38
rs563834980 Chr.19: 51366299-51366299 on Build GRCh38
rs774872090 Chr.19: 51366260-51366260 on Build GRCh38
rs7249325 Chr.19: 51366496-51366496 on Build GRCh38
rs114105512 Chr.19: 51366433-51366433 on Build GRCh38
rs546258653 Chr.19: 51366427-51366427 on Build GRCh38
rs529887209 Chr.19: 51366254-51366254 on Build GRCh38
rs371772513 Chr.19: 51366369-51366369 on Build GRCh38
rs759933741 Chr.19: 51366343-51366343 on Build GRCh38
rs969249875 Chr.19: 51366462-51366462 on Build GRCh38
rs139296016 Chr.19: 51366537-51366537 on Build GRCh38
rs933203320 Chr.19: 51366402-51366402 on Build GRCh38
rs190805513 Chr.19: 51366638-51366638 on Build GRCh38
rs368994872 Chr.19: 51366625-51366625 on Build GRCh38
rs767751467 Chr.19: 51366271-51366271 on Build GRCh38
rs776277833 Chr.19: 51366329-51366329 on Build GRCh38
rs111340257 Chr.19: 51366460-51366460 on Build GRCh38
rs956380221 Chr.19: 51366520-51366520 on Build GRCh38

View Full Product Details