Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53290790-53291282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC8
Entrez Gene ID
112401
Gene Name
baculoviral IAP repeat containing 8
Gene Aliases
ILP-2, ILP2, hILP2
UniGene
Hs.348263
Gene Chromosome Location
Chr.19: 53289601-53291622 on Build GRCh38
Transcripts
NM_033341.4

SNP Detail

SNP ID Chromosome Location
rs1050054156 Chr.19: 53291008-53291008 on Build GRCh38
rs903825707 Chr.19: 53291044-53291044 on Build GRCh38
rs182281884 Chr.19: 53291199-53291199 on Build GRCh38
rs527287494 Chr.19: 53290956-53290956 on Build GRCh38
rs923228093 Chr.19: 53290994-53290994 on Build GRCh38
rs969763714 Chr.19: 53291168-53291168 on Build GRCh38
rs1026621315 Chr.19: 53291251-53291251 on Build GRCh38
rs574583908 Chr.19: 53291244-53291244 on Build GRCh38
rs962148396 Chr.19: 53291212-53291212 on Build GRCh38
rs78399115 Chr.19: 53290930-53290930 on Build GRCh38
rs1031283260 Chr.19: 53290922-53290922 on Build GRCh38
rs902914806 Chr.19: 53291096-53291096 on Build GRCh38
rs77065858 Chr.19: 53290914-53290914 on Build GRCh38
rs377307617 Chr.19: 53290995-53290995 on Build GRCh38
rs995194645 Chr.19: 53291243-53291243 on Build GRCh38
rs919554396 Chr.19: 53290905-53290905 on Build GRCh38
rs985256756 Chr.19: 53290962-53290962 on Build GRCh38
rs117480526 Chr.19: 53290945-53290945 on Build GRCh38
rs960954324 Chr.19: 53291150-53291150 on Build GRCh38
rs909057093 Chr.19: 53290981-53290981 on Build GRCh38
rs10426632 Chr.19: 53291196-53291196 on Build GRCh38
rs963821239 Chr.19: 53290898-53290898 on Build GRCh38
rs552174853 Chr.19: 53290993-53290993 on Build GRCh38
rs923070762 Chr.19: 53291225-53291225 on Build GRCh38
rs531470023 Chr.19: 53291087-53291087 on Build GRCh38
rs999511572 Chr.19: 53291049-53291049 on Build GRCh38
rs969502551 Chr.19: 53290953-53290953 on Build GRCh38
rs369085357 Chr.19: 53290929-53290930 on Build GRCh38
rs73590784 Chr.19: 53291214-53291214 on Build GRCh38
rs888758890 Chr.19: 53291079-53291079 on Build GRCh38
rs549592039 Chr.19: 53291095-53291095 on Build GRCh38
rs933260277 Chr.19: 53291235-53291235 on Build GRCh38
rs150466843 Chr.19: 53291186-53291186 on Build GRCh38
rs75354845 Chr.19: 53290929-53290929 on Build GRCh38
rs11554684 Chr.19: 53290902-53290902 on Build GRCh38
rs910457092 Chr.19: 53291247-53291247 on Build GRCh38
rs999735478 Chr.19: 53291100-53291100 on Build GRCh38
rs529920806 Chr.19: 53291215-53291215 on Build GRCh38
rs11554685 Chr.19: 53290901-53290901 on Build GRCh38
rs888881216 Chr.19: 53290862-53290862 on Build GRCh38
rs1013806209 Chr.19: 53291162-53291162 on Build GRCh38
rs1052373736 Chr.19: 53291144-53291144 on Build GRCh38
rs749453080 Chr.19: 53290900-53290900 on Build GRCh38
rs940545584 Chr.19: 53290992-53290992 on Build GRCh38
rs544709958 Chr.19: 53290919-53290919 on Build GRCh38
rs536440576 Chr.19: 53291155-53291155 on Build GRCh38
rs548875246 Chr.19: 53291177-53291177 on Build GRCh38
rs533944037 Chr.19: 53291188-53291188 on Build GRCh38
rs569483338 Chr.19: 53291137-53291137 on Build GRCh38
rs868169873 Chr.19: 53291167-53291167 on Build GRCh38
rs951080897 Chr.19: 53291253-53291253 on Build GRCh38
rs113135162 Chr.19: 53290837-53290838 on Build GRCh38
rs1011705311 Chr.19: 53290892-53290892 on Build GRCh38
rs138318721 Chr.19: 53291239-53291239 on Build GRCh38
rs559759304 Chr.19: 53290879-53290879 on Build GRCh38
rs947065224 Chr.19: 53291262-53291262 on Build GRCh38
rs1031167710 Chr.19: 53291136-53291136 on Build GRCh38
rs1050043918 Chr.19: 53290850-53290850 on Build GRCh38

View Full Product Details