Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54011882-54012388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG6
Entrez Gene ID
59285
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 6
Gene Aliases
-
UniGene
Hs.631560
Gene Chromosome Location
Chr.19: 53991149-54012666 on Build GRCh38
Transcripts
NM_031897.2 NM_145814.1 NM_145815.1 NR_102308.1 XM_006723316.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7186868 c.728T>A p.L243Q Chr.19: 54012134-54012134 on Build GRCh38
4081155 c.615G>A p.P205P Chr.19: 54012021-54012021 on Build GRCh38
1396196 c.622delC p.P209fs*21 Chr.19: 54012028-54012028 on Build GRCh38
713819 c.608T>C p.V203A Chr.19: 54012014-54012014 on Build GRCh38
1396195 c.585C>T p.S195S Chr.19: 54011991-54011991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749675655 Chr.19: 54011918-54011918 on Build GRCh38
rs776788761 Chr.19: 54012179-54012179 on Build GRCh38
rs747892500 Chr.19: 54012167-54012167 on Build GRCh38
rs760644366 Chr.19: 54012172-54012172 on Build GRCh38
rs780420395 Chr.19: 54012036-54012036 on Build GRCh38
rs1008774385 Chr.19: 54012211-54012211 on Build GRCh38
rs369729281 Chr.19: 54012032-54012032 on Build GRCh38
rs929186326 Chr.19: 54012351-54012351 on Build GRCh38
rs892255076 Chr.19: 54011937-54011937 on Build GRCh38
rs186979452 Chr.19: 54012257-54012257 on Build GRCh38
rs750548219 Chr.19: 54011958-54011958 on Build GRCh38
rs773675067 Chr.19: 54012304-54012304 on Build GRCh38
rs777674290 Chr.19: 54012235-54012235 on Build GRCh38
rs766454497 Chr.19: 54012174-54012174 on Build GRCh38
rs112544975 Chr.19: 54011928-54011928 on Build GRCh38
rs926398071 Chr.19: 54012321-54012321 on Build GRCh38
rs374782267 Chr.19: 54011998-54011998 on Build GRCh38
rs767495494 Chr.19: 54012029-54012029 on Build GRCh38
rs200480751 Chr.19: 54012080-54012080 on Build GRCh38
rs745367034 Chr.19: 54012072-54012072 on Build GRCh38
rs370862252 Chr.19: 54012185-54012185 on Build GRCh38
rs768136238 Chr.19: 54012155-54012155 on Build GRCh38
rs753274467 Chr.19: 54012221-54012221 on Build GRCh38
rs760231071 Chr.19: 54011920-54011920 on Build GRCh38
rs773976957 Chr.19: 54012082-54012082 on Build GRCh38
rs752418848 Chr.19: 54011933-54011933 on Build GRCh38
rs758762265 Chr.19: 54012052-54012052 on Build GRCh38
rs763833590 Chr.19: 54012017-54012017 on Build GRCh38
rs949111352 Chr.19: 54012079-54012079 on Build GRCh38
rs148905506 Chr.19: 54012170-54012170 on Build GRCh38
rs74403906 Chr.19: 54012298-54012298 on Build GRCh38
rs913639979 Chr.19: 54012058-54012058 on Build GRCh38
rs534072928 Chr.19: 54012303-54012303 on Build GRCh38
rs61738518 Chr.19: 54012145-54012145 on Build GRCh38
rs1033246081 Chr.19: 54012245-54012245 on Build GRCh38
rs780898797 Chr.19: 54012008-54012009 on Build GRCh38
rs981849349 Chr.19: 54012308-54012308 on Build GRCh38
rs79135916 Chr.19: 54012326-54012326 on Build GRCh38
rs749657710 Chr.19: 54011972-54011972 on Build GRCh38
rs754156793 Chr.19: 54012039-54012039 on Build GRCh38
rs751492357 Chr.19: 54012020-54012020 on Build GRCh38
rs754584633 Chr.19: 54012222-54012222 on Build GRCh38
rs201854302 Chr.19: 54011940-54011940 on Build GRCh38
rs761682241 Chr.19: 54012085-54012085 on Build GRCh38
rs775961001 Chr.19: 54011996-54011996 on Build GRCh38
rs988571477 Chr.19: 54012059-54012059 on Build GRCh38
rs773351063 Chr.19: 54012088-54012088 on Build GRCh38
rs762748167 Chr.19: 54011934-54011934 on Build GRCh38
rs113483278 Chr.19: 54011919-54011919 on Build GRCh38
rs1047681185 Chr.19: 54012363-54012363 on Build GRCh38
rs759143333 Chr.19: 54011929-54011929 on Build GRCh38
rs770065137 Chr.19: 54012080-54012103 on Build GRCh38
rs778401588 Chr.19: 54012225-54012225 on Build GRCh38
rs756338330 Chr.19: 54011961-54011961 on Build GRCh38
rs775178620 Chr.19: 54012012-54012012 on Build GRCh38
rs752943229 Chr.19: 54012128-54012128 on Build GRCh38
rs2291068 Chr.19: 54012307-54012307 on Build GRCh38
rs376472409 Chr.19: 54012109-54012109 on Build GRCh38
rs989035958 Chr.19: 54012282-54012282 on Build GRCh38
rs779446783 Chr.19: 54011982-54011982 on Build GRCh38
rs764003596 Chr.19: 54011939-54011939 on Build GRCh38
rs765496615 Chr.19: 54012125-54012125 on Build GRCh38
rs780438713 Chr.19: 54011966-54011966 on Build GRCh38
rs1002101805 Chr.19: 54011947-54011947 on Build GRCh38
rs764897530 Chr.19: 54011932-54011932 on Build GRCh38
rs376108604 Chr.19: 54012042-54012042 on Build GRCh38
rs751942145 Chr.19: 54012203-54012203 on Build GRCh38
rs139947484 Chr.19: 54011984-54011984 on Build GRCh38
rs145464848 Chr.19: 54012055-54012055 on Build GRCh38
rs761605328 Chr.19: 54012025-54012025 on Build GRCh38
rs375248786 Chr.19: 54011924-54011924 on Build GRCh38
rs769376031 Chr.19: 54011999-54011999 on Build GRCh38
rs201389291 Chr.19: 54012040-54012040 on Build GRCh38
rs766617187 Chr.19: 54012098-54012098 on Build GRCh38
rs201421758 Chr.19: 54011944-54011944 on Build GRCh38
rs747553099 Chr.19: 54012138-54012138 on Build GRCh38
rs755120014 Chr.19: 54012106-54012106 on Build GRCh38
rs770869382 Chr.19: 54011979-54011980 on Build GRCh38
rs778136049 Chr.19: 54012134-54012134 on Build GRCh38
rs763193820 Chr.19: 54012196-54012196 on Build GRCh38
rs769213773 Chr.19: 54011974-54011974 on Build GRCh38
rs936337599 Chr.19: 54012051-54012051 on Build GRCh38
rs769876729 Chr.19: 54012286-54012286 on Build GRCh38
rs199614002 Chr.19: 54012075-54012075 on Build GRCh38
rs771636505 Chr.19: 54012323-54012323 on Build GRCh38
rs12980121 Chr.19: 54012161-54012161 on Build GRCh38
rs940536570 Chr.19: 54012333-54012333 on Build GRCh38
rs200238789 Chr.19: 54012181-54012181 on Build GRCh38
rs369577894 Chr.19: 54012050-54012050 on Build GRCh38
rs920441250 Chr.19: 54012337-54012337 on Build GRCh38
rs576293186 Chr.19: 54012034-54012034 on Build GRCh38
rs182399347 Chr.19: 54012226-54012226 on Build GRCh38
rs775091115 Chr.19: 54012076-54012076 on Build GRCh38
rs757720939 Chr.19: 54012205-54012205 on Build GRCh38
rs753997457 Chr.19: 54012103-54012103 on Build GRCh38
rs768377025 Chr.19: 54012078-54012078 on Build GRCh38
rs773263020 Chr.19: 54012171-54012171 on Build GRCh38
rs899129997 Chr.19: 54011969-54011969 on Build GRCh38
rs745648688 Chr.19: 54012028-54012028 on Build GRCh38
rs199694557 Chr.19: 54012206-54012206 on Build GRCh38
rs771648024 Chr.19: 54012227-54012227 on Build GRCh38
rs764488922 Chr.19: 54012197-54012197 on Build GRCh38
rs368389668 Chr.19: 54011915-54011915 on Build GRCh38
rs149744809 Chr.19: 54012147-54012147 on Build GRCh38
rs762730085 Chr.19: 54012015-54012015 on Build GRCh38
rs753267707 Chr.19: 54012250-54012250 on Build GRCh38
rs758852047 Chr.19: 54012129-54012129 on Build GRCh38
rs755597457 Chr.19: 54012140-54012140 on Build GRCh38
rs866833754 Chr.19: 54012137-54012137 on Build GRCh38
rs147610426 Chr.19: 54012087-54012087 on Build GRCh38
rs772659738 Chr.19: 54011985-54011985 on Build GRCh38
rs373753169 Chr.19: 54012096-54012096 on Build GRCh38

View Full Product Details