Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55075961-55076453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS8L1
Entrez Gene ID
54869
Gene Name
EPS8 like 1
Gene Aliases
DRC3, EPS8R1, PP10566
UniGene
Hs.438862
Gene Chromosome Location
Chr.19: 55075853-55087923 on Build GRCh38
Transcripts
NM_133180.2 XM_005259020.1 XM_011527051.2 XM_011527052.2

SNP Detail

SNP ID Chromosome Location
rs565103533 Chr.19: 55076374-55076374 on Build GRCh38
rs945515437 Chr.19: 55076343-55076343 on Build GRCh38
rs59229408 Chr.19: 55076322-55076322 on Build GRCh38
rs573625092 Chr.19: 55076294-55076294 on Build GRCh38
rs575792974 Chr.19: 55076383-55076383 on Build GRCh38
rs1042448647 Chr.19: 55076348-55076348 on Build GRCh38
rs372107788 Chr.19: 55076269-55076271 on Build GRCh38
rs1056862234 Chr.19: 55076358-55076358 on Build GRCh38
rs149375299 Chr.19: 55076093-55076093 on Build GRCh38
rs145504856 Chr.19: 55076219-55076219 on Build GRCh38
rs758419941 Chr.19: 55076415-55076415 on Build GRCh38
rs58358874 Chr.19: 55076315-55076316 on Build GRCh38
rs61448344 Chr.19: 55076300-55076300 on Build GRCh38
rs767081873 Chr.19: 55076285-55076285 on Build GRCh38
rs573795150 Chr.19: 55076206-55076207 on Build GRCh38
rs62124197 Chr.19: 55076290-55076290 on Build GRCh38
rs111331426 Chr.19: 55076331-55076331 on Build GRCh38
rs553797970 Chr.19: 55076266-55076266 on Build GRCh38
rs57079191 Chr.19: 55076286-55076286 on Build GRCh38
rs750607598 Chr.19: 55076167-55076167 on Build GRCh38
rs952476119 Chr.19: 55076402-55076402 on Build GRCh38
rs761462935 Chr.19: 55076097-55076097 on Build GRCh38
rs767621636 Chr.19: 55076004-55076004 on Build GRCh38
rs561533023 Chr.19: 55076373-55076373 on Build GRCh38
rs1000579466 Chr.19: 55076027-55076027 on Build GRCh38
rs56845287 Chr.19: 55076171-55076171 on Build GRCh38
rs185230963 Chr.19: 55076002-55076002 on Build GRCh38
rs952497072 Chr.19: 55076043-55076043 on Build GRCh38
rs952662192 Chr.19: 55076313-55076313 on Build GRCh38
rs57894149 Chr.19: 55076163-55076163 on Build GRCh38
rs59132518 Chr.19: 55076215-55076216 on Build GRCh38
rs903016015 Chr.19: 55075983-55075983 on Build GRCh38
rs991182063 Chr.19: 55076022-55076022 on Build GRCh38
rs796363302 Chr.19: 55076103-55076103 on Build GRCh38
rs570984092 Chr.19: 55076272-55076272 on Build GRCh38
rs893763625 Chr.19: 55076336-55076336 on Build GRCh38
rs777408540 Chr.19: 55076418-55076418 on Build GRCh38
rs573185509 Chr.19: 55076354-55076354 on Build GRCh38
rs57362846 Chr.19: 55076177-55076177 on Build GRCh38
rs60051147 Chr.19: 55076257-55076257 on Build GRCh38
rs59407886 Chr.19: 55076278-55076278 on Build GRCh38
rs59964243 Chr.19: 55076275-55076276 on Build GRCh38
rs201342041 Chr.19: 55076352-55076352 on Build GRCh38
rs61013635 Chr.19: 55076230-55076231 on Build GRCh38
rs111231617 Chr.19: 55076204-55076204 on Build GRCh38
rs374680581 Chr.19: 55076403-55076403 on Build GRCh38
rs114567136 Chr.19: 55076156-55076156 on Build GRCh38
rs113280777 Chr.19: 55076021-55076021 on Build GRCh38
rs58832161 Chr.19: 55076236-55076236 on Build GRCh38
rs372742476 Chr.19: 55076281-55076281 on Build GRCh38
rs796271670 Chr.19: 55076129-55076135 on Build GRCh38
rs982906421 Chr.19: 55076057-55076057 on Build GRCh38
rs559273322 Chr.19: 55075985-55075985 on Build GRCh38
rs62124194 Chr.19: 55076100-55076100 on Build GRCh38
rs62124195 Chr.19: 55076132-55076132 on Build GRCh38
rs60762307 Chr.19: 55076285-55076285 on Build GRCh38
rs1007831795 Chr.19: 55076088-55076088 on Build GRCh38
rs112632410 Chr.19: 55076128-55076129 on Build GRCh38
rs371722650 Chr.19: 55076401-55076401 on Build GRCh38
rs975320958 Chr.19: 55076284-55076284 on Build GRCh38
rs1015336152 Chr.19: 55076429-55076429 on Build GRCh38
rs62124196 Chr.19: 55076134-55076134 on Build GRCh38
rs1019270628 Chr.19: 55076179-55076179 on Build GRCh38
rs758596786 Chr.19: 55076200-55076200 on Build GRCh38
rs542735815 Chr.19: 55076031-55076031 on Build GRCh38
rs778489991 Chr.19: 55076409-55076409 on Build GRCh38
rs948259158 Chr.19: 55076350-55076350 on Build GRCh38
rs573122303 Chr.19: 55075997-55075997 on Build GRCh38
rs12460727 Chr.19: 55076019-55076019 on Build GRCh38
rs908746427 Chr.19: 55076194-55076194 on Build GRCh38
rs982671900 Chr.19: 55076321-55076321 on Build GRCh38
rs10853876 Chr.19: 55076040-55076040 on Build GRCh38
rs756624545 Chr.19: 55076090-55076090 on Build GRCh38
rs12460747 Chr.19: 55076110-55076110 on Build GRCh38
rs796943788 Chr.19: 55076166-55076167 on Build GRCh38
rs193177771 Chr.19: 55076159-55076159 on Build GRCh38
rs12460728 Chr.19: 55076078-55076078 on Build GRCh38
rs748759989 Chr.19: 55076096-55076096 on Build GRCh38
rs185380492 Chr.19: 55076263-55076263 on Build GRCh38
rs561368980 Chr.19: 55076119-55076119 on Build GRCh38
rs903048593 Chr.19: 55076353-55076353 on Build GRCh38
rs538805896 Chr.19: 55076175-55076175 on Build GRCh38
rs1020122687 Chr.19: 55075980-55075980 on Build GRCh38
rs963902957 Chr.19: 55076211-55076211 on Build GRCh38
rs200448020 Chr.19: 55076230-55076235 on Build GRCh38
rs56894949 Chr.19: 55076152-55076152 on Build GRCh38
rs796116141 Chr.19: 55076171-55076172 on Build GRCh38
rs61712477 Chr.19: 55076157-55076157 on Build GRCh38
rs61001933 Chr.19: 55076145-55076145 on Build GRCh38
rs748030180 Chr.19: 55076410-55076410 on Build GRCh38
rs769320052 Chr.19: 55076093-55076093 on Build GRCh38
rs867776429 Chr.19: 55076140-55076140 on Build GRCh38

View Full Product Details