Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55159145-55159650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAAF3
Entrez Gene ID
352909
Gene Name
dynein axonemal assembly factor 3
Gene Aliases
C19orf51, CILD2, DAB1, PCD, PF22
UniGene
Hs.351582
Gene Chromosome Location
Chr.19: 55158661-55166722 on Build GRCh38
Transcripts
NM_001256714.1 NM_001256715.1 NM_001256716.1 NM_178837.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960529 c.1163A>T p.Q388L Chr.19: 55159363-55159363 on Build GRCh38
6152259 c.1267G>C p.D423H Chr.19: 55159259-55159259 on Build GRCh38
3539360 c.1443C>T p.Y481Y Chr.19: 55159446-55159446 on Build GRCh38
7189771 c.1365-7T>C p.? Chr.19: 55159614-55159614 on Build GRCh38
5484338 c.1192G>A p.A398T Chr.19: 55159334-55159334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576940831 Chr.19: 55159330-55159330 on Build GRCh38
rs1050524902 Chr.19: 55159215-55159215 on Build GRCh38
rs918071971 Chr.19: 55159351-55159351 on Build GRCh38
rs377325487 Chr.19: 55159275-55159275 on Build GRCh38
rs985181220 Chr.19: 55159277-55159277 on Build GRCh38
rs767754398 Chr.19: 55159295-55159295 on Build GRCh38
rs765899538 Chr.19: 55159586-55159586 on Build GRCh38
rs374560751 Chr.19: 55159299-55159299 on Build GRCh38
rs754336193 Chr.19: 55159401-55159401 on Build GRCh38
rs758777169 Chr.19: 55159491-55159491 on Build GRCh38
rs114601492 Chr.19: 55159283-55159283 on Build GRCh38
rs776506898 Chr.19: 55159256-55159256 on Build GRCh38
rs201919395 Chr.19: 55159274-55159274 on Build GRCh38
rs756412762 Chr.19: 55159217-55159217 on Build GRCh38
rs760765736 Chr.19: 55159465-55159465 on Build GRCh38
rs764356342 Chr.19: 55159276-55159276 on Build GRCh38
rs760023977 Chr.19: 55159548-55159548 on Build GRCh38
rs373798404 Chr.19: 55159562-55159562 on Build GRCh38
rs201887376 Chr.19: 55159335-55159335 on Build GRCh38
rs562819833 Chr.19: 55159358-55159358 on Build GRCh38
rs761934655 Chr.19: 55159290-55159290 on Build GRCh38
rs71367155 Chr.19: 55159613-55159613 on Build GRCh38
rs760679876 Chr.19: 55159360-55159360 on Build GRCh38
rs770461794 Chr.19: 55159255-55159255 on Build GRCh38
rs886054638 Chr.19: 55159294-55159294 on Build GRCh38
rs750058869 Chr.19: 55159567-55159567 on Build GRCh38
rs376972789 Chr.19: 55159612-55159612 on Build GRCh38
rs781749900 Chr.19: 55159202-55159202 on Build GRCh38
rs748755474 Chr.19: 55159507-55159507 on Build GRCh38
rs200506321 Chr.19: 55159479-55159479 on Build GRCh38
rs767701157 Chr.19: 55159359-55159359 on Build GRCh38
rs746451709 Chr.19: 55159242-55159242 on Build GRCh38
rs750077313 Chr.19: 55159497-55159497 on Build GRCh38
rs746688785 Chr.19: 55159328-55159328 on Build GRCh38
rs201231901 Chr.19: 55159436-55159436 on Build GRCh38
rs755289447 Chr.19: 55159402-55159402 on Build GRCh38
rs878854280 Chr.19: 55159278-55159278 on Build GRCh38
rs748036767 Chr.19: 55159472-55159473 on Build GRCh38
rs774691685 Chr.19: 55159357-55159357 on Build GRCh38
rs759571188 Chr.19: 55159257-55159257 on Build GRCh38
rs778741276 Chr.19: 55159525-55159525 on Build GRCh38
rs780507081 Chr.19: 55159220-55159220 on Build GRCh38
rs371072227 Chr.19: 55159620-55159620 on Build GRCh38
rs746397485 Chr.19: 55159205-55159205 on Build GRCh38
rs918171964 Chr.19: 55159553-55159553 on Build GRCh38
rs987811530 Chr.19: 55159604-55159604 on Build GRCh38
rs1030647614 Chr.19: 55159514-55159514 on Build GRCh38
rs756970924 Chr.19: 55159427-55159427 on Build GRCh38
rs747747588 Chr.19: 55159221-55159221 on Build GRCh38
rs755385476 Chr.19: 55159266-55159266 on Build GRCh38
rs372062264 Chr.19: 55159445-55159445 on Build GRCh38
rs777175999 Chr.19: 55159470-55159470 on Build GRCh38
rs779003677 Chr.19: 55159518-55159518 on Build GRCh38
rs199690352 Chr.19: 55159192-55159192 on Build GRCh38
rs774354704 Chr.19: 55159449-55159449 on Build GRCh38
rs777366628 Chr.19: 55159326-55159326 on Build GRCh38
rs60176657 Chr.19: 55159621-55159621 on Build GRCh38
rs887755984 Chr.19: 55159167-55159167 on Build GRCh38
rs752844873 Chr.19: 55159179-55159179 on Build GRCh38
rs77322382 Chr.19: 55159403-55159403 on Build GRCh38
rs766839379 Chr.19: 55159363-55159363 on Build GRCh38
rs779469618 Chr.19: 55159503-55159503 on Build GRCh38
rs765106294 Chr.19: 55159177-55159177 on Build GRCh38
rs772359974 Chr.19: 55159461-55159461 on Build GRCh38
rs754792308 Chr.19: 55159261-55159262 on Build GRCh38
rs1011268850 Chr.19: 55159565-55159565 on Build GRCh38
rs546293010 Chr.19: 55159334-55159334 on Build GRCh38
rs771811729 Chr.19: 55159241-55159241 on Build GRCh38
rs28377509 Chr.19: 55159457-55159457 on Build GRCh38
rs201929981 Chr.19: 55159433-55159433 on Build GRCh38
rs111250144 Chr.19: 55159440-55159440 on Build GRCh38
rs778885305 Chr.19: 55159419-55159420 on Build GRCh38
rs747247157 Chr.19: 55159534-55159534 on Build GRCh38
rs771903581 Chr.19: 55159592-55159593 on Build GRCh38
rs200919669 Chr.19: 55159267-55159267 on Build GRCh38
rs757426979 Chr.19: 55159201-55159201 on Build GRCh38
rs368138318 Chr.19: 55159301-55159301 on Build GRCh38
rs751749516 Chr.19: 55159193-55159193 on Build GRCh38
rs749232820 Chr.19: 55159513-55159513 on Build GRCh38
rs199947168 Chr.19: 55159464-55159464 on Build GRCh38
rs766394618 Chr.19: 55159469-55159469 on Build GRCh38
rs202084011 Chr.19: 55159561-55159561 on Build GRCh38
rs754170208 Chr.19: 55159316-55159316 on Build GRCh38
rs758688329 Chr.19: 55159181-55159181 on Build GRCh38
rs967781137 Chr.19: 55159512-55159512 on Build GRCh38
rs753336733 Chr.19: 55159605-55159605 on Build GRCh38
rs760079402 Chr.19: 55159472-55159472 on Build GRCh38
rs751187170 Chr.19: 55159404-55159404 on Build GRCh38
rs200920341 Chr.19: 55159615-55159615 on Build GRCh38
rs536068494 Chr.19: 55159195-55159195 on Build GRCh38
rs886054639 Chr.19: 55159382-55159382 on Build GRCh38
rs745788397 Chr.19: 55159331-55159331 on Build GRCh38
rs751901961 Chr.19: 55159245-55159245 on Build GRCh38
rs764412803 Chr.19: 55159566-55159566 on Build GRCh38
rs765675817 Chr.19: 55159550-55159550 on Build GRCh38
rs886038424 Chr.19: 55159622-55159622 on Build GRCh38
rs375688483 Chr.19: 55159524-55159524 on Build GRCh38
rs771791101 Chr.19: 55159533-55159533 on Build GRCh38
rs747254654 Chr.19: 55159624-55159624 on Build GRCh38
rs754536872 Chr.19: 55159608-55159608 on Build GRCh38
rs372161739 Chr.19: 55159311-55159311 on Build GRCh38
rs755299958 Chr.19: 55159324-55159324 on Build GRCh38
rs768592462 Chr.19: 55159355-55159355 on Build GRCh38
rs369064646 Chr.19: 55159544-55159544 on Build GRCh38
rs755788366 Chr.19: 55159572-55159572 on Build GRCh38
rs753118183 Chr.19: 55159486-55159486 on Build GRCh38
rs373489149 Chr.19: 55159432-55159432 on Build GRCh38
rs564220260 Chr.19: 55159501-55159501 on Build GRCh38
rs779990994 Chr.19: 55159437-55159437 on Build GRCh38

View Full Product Details